Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 6349680-6349941 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACOT7
Entrez Gene ID
11332
Gene Name
acyl-CoA thioesterase 7
Gene Aliases
ACH1, ACT, BACH, CTE-II, LACH, LACH1, hBACH
UniGene
Hs.126137
Gene Chromosome Location
Chr.1: 6264272-6393766 on Build GRCh38
Transcripts
NM_007274.3 NM_181864.2 NM_181865.2 NM_181866.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4658877 c.223G>A p.A75T Chr.1: 6349787-6349787 on Build GRCh38
4658879 c.100G>A p.A34T Chr.1: 6349787-6349787 on Build GRCh38
2238769 c.148A>G p.M50V Chr.1: 6349862-6349862 on Build GRCh38
6592306 c.240G>A p.R80R Chr.1: 6349770-6349770 on Build GRCh38
6366890 c.181C>T p.H61Y Chr.1: 6349769-6349769 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775625418 Chr.1: 6349842-6349842 on Build GRCh38
rs754648000 Chr.1: 6349830-6349830 on Build GRCh38
rs200819032 Chr.1: 6349751-6349751 on Build GRCh38
rs905177413 Chr.1: 6349862-6349862 on Build GRCh38
rs773070166 Chr.1: 6349750-6349750 on Build GRCh38
rs755608954 Chr.1: 6349809-6349809 on Build GRCh38
rs1036321605 Chr.1: 6349903-6349903 on Build GRCh38
rs754957438 Chr.1: 6349731-6349731 on Build GRCh38
rs909221872 Chr.1: 6349856-6349856 on Build GRCh38
rs146057501 Chr.1: 6349772-6349772 on Build GRCh38
rs779581844 Chr.1: 6349818-6349818 on Build GRCh38
rs748928255 Chr.1: 6349709-6349709 on Build GRCh38
rs762286520 Chr.1: 6349861-6349861 on Build GRCh38
rs368642662 Chr.1: 6349725-6349725 on Build GRCh38
rs369212021 Chr.1: 6349702-6349702 on Build GRCh38
rs765865616 Chr.1: 6349912-6349912 on Build GRCh38
rs146476044 Chr.1: 6349836-6349836 on Build GRCh38
rs773892923 Chr.1: 6349881-6349881 on Build GRCh38
rs767949225 Chr.1: 6349871-6349871 on Build GRCh38
rs750908241 Chr.1: 6349800-6349800 on Build GRCh38
rs146315441 Chr.1: 6349841-6349841 on Build GRCh38
rs140259918 Chr.1: 6349788-6349788 on Build GRCh38
rs376851666 Chr.1: 6349752-6349752 on Build GRCh38
rs754263561 Chr.1: 6349705-6349705 on Build GRCh38
rs761220171 Chr.1: 6349892-6349892 on Build GRCh38
rs562460126 Chr.1: 6349824-6349824 on Build GRCh38
rs763150711 Chr.1: 6349845-6349845 on Build GRCh38
rs371975249 Chr.1: 6349733-6349733 on Build GRCh38
rs751965758 Chr.1: 6349779-6349779 on Build GRCh38
rs768151405 Chr.1: 6349799-6349799 on Build GRCh38
rs768373241 Chr.1: 6349721-6349721 on Build GRCh38
rs111828929 Chr.1: 6349901-6349901 on Build GRCh38
rs755589640 Chr.1: 6349701-6349701 on Build GRCh38
rs757726898 Chr.1: 6349736-6349736 on Build GRCh38
rs17029376 Chr.1: 6349919-6349919 on Build GRCh38
rs978376483 Chr.1: 6349823-6349823 on Build GRCh38
rs940563368 Chr.1: 6349896-6349896 on Build GRCh38
rs771996901 Chr.1: 6349744-6349744 on Build GRCh38
rs1046695237 Chr.1: 6349857-6349857 on Build GRCh38
rs753387191 Chr.1: 6349916-6349916 on Build GRCh38
rs778750851 Chr.1: 6349831-6349831 on Build GRCh38
rs763204698 Chr.1: 6349770-6349770 on Build GRCh38
rs577490040 Chr.1: 6349835-6349835 on Build GRCh38
rs985165374 Chr.1: 6349848-6349848 on Build GRCh38
rs768926964 Chr.1: 6349859-6349859 on Build GRCh38

View Full Product Details