Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 150999286-150999559 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM63A
Entrez Gene ID
55793
Gene Name
family with sequence similarity 63 member A
Gene Aliases
-
UniGene
Hs.743952
Gene Chromosome Location
Chr.1: 150996737-151008378 on Build GRCh38
Transcripts
NM_001040217.2 NM_001163258.1 NM_001163259.1 NM_001163260.1 NM_001319998.1 NM_018379.4 XM_005245321.3 XM_005245325.3 XM_005245327.3 XM_005245328.3 XM_011509757.2 XM_011509759.2 XM_017001770.1 XM_017001771.1 XM_017001772.1 XM_017001773.1 XM_017001774.1 XM_017001775.1 XM_017001776.1 XM_017001777.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
895665 c.962G>T p.S321I Chr.1: 150999388-150999388 on Build GRCh38
6297957 c.1033C>A p.H345N Chr.1: 150999461-150999461 on Build GRCh38
6659922 c.1030T>C p.Y344H Chr.1: 150999464-150999464 on Build GRCh38
6396763 c.848C>T p.A283V Chr.1: 150999502-150999502 on Build GRCh38
675510 c.946C>T p.R316* Chr.1: 150999404-150999404 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762341074 Chr.1: 150999418-150999418 on Build GRCh38
rs587761368 Chr.1: 150999318-150999318 on Build GRCh38
rs778907961 Chr.1: 150999462-150999462 on Build GRCh38
rs767537672 Chr.1: 150999526-150999526 on Build GRCh38
rs200192131 Chr.1: 150999402-150999402 on Build GRCh38
rs201570673 Chr.1: 150999364-150999364 on Build GRCh38
rs374869298 Chr.1: 150999414-150999414 on Build GRCh38
rs375355944 Chr.1: 150999338-150999338 on Build GRCh38
rs758387166 Chr.1: 150999354-150999354 on Build GRCh38
rs587598007 Chr.1: 150999419-150999419 on Build GRCh38
rs767749987 Chr.1: 150999310-150999310 on Build GRCh38
rs371523428 Chr.1: 150999441-150999441 on Build GRCh38
rs754892523 Chr.1: 150999426-150999426 on Build GRCh38
rs374995576 Chr.1: 150999459-150999459 on Build GRCh38
rs369386016 Chr.1: 150999478-150999478 on Build GRCh38
rs139059568 Chr.1: 150999403-150999403 on Build GRCh38
rs771507332 Chr.1: 150999396-150999396 on Build GRCh38
rs751070204 Chr.1: 150999511-150999511 on Build GRCh38
rs756365955 Chr.1: 150999449-150999450 on Build GRCh38
rs587677126 Chr.1: 150999332-150999332 on Build GRCh38
rs200349479 Chr.1: 150999508-150999508 on Build GRCh38
rs138246511 Chr.1: 150999473-150999473 on Build GRCh38
rs769865387 Chr.1: 150999375-150999375 on Build GRCh38
rs761487426 Chr.1: 150999411-150999411 on Build GRCh38
rs761009150 Chr.1: 150999479-150999479 on Build GRCh38
rs759566643 Chr.1: 150999517-150999517 on Build GRCh38
rs587607974 Chr.1: 150999477-150999477 on Build GRCh38
rs756429078 Chr.1: 150999351-150999351 on Build GRCh38
rs751532553 Chr.1: 150999422-150999422 on Build GRCh38
rs779836205 Chr.1: 150999359-150999359 on Build GRCh38
rs765808077 Chr.1: 150999498-150999498 on Build GRCh38
rs780350510 Chr.1: 150999309-150999309 on Build GRCh38
rs755855761 Chr.1: 150999527-150999527 on Build GRCh38
rs1021406385 Chr.1: 150999532-150999532 on Build GRCh38
rs773360327 Chr.1: 150999383-150999383 on Build GRCh38
rs996238951 Chr.1: 150999501-150999501 on Build GRCh38
rs587720999 Chr.1: 150999413-150999413 on Build GRCh38
rs767973476 Chr.1: 150999350-150999351 on Build GRCh38
rs368632463 Chr.1: 150999340-150999340 on Build GRCh38
rs901793980 Chr.1: 150999483-150999483 on Build GRCh38
rs754819337 Chr.1: 150999362-150999362 on Build GRCh38
rs150431391 Chr.1: 150999404-150999404 on Build GRCh38
rs928919032 Chr.1: 150999369-150999369 on Build GRCh38
rs756406352 Chr.1: 150999446-150999446 on Build GRCh38
rs777824543 Chr.1: 150999454-150999454 on Build GRCh38
rs376639048 Chr.1: 150999339-150999339 on Build GRCh38
rs762585378 Chr.1: 150999497-150999497 on Build GRCh38
rs749451114 Chr.1: 150999458-150999458 on Build GRCh38
rs772740181 Chr.1: 150999491-150999491 on Build GRCh38
rs949875930 Chr.1: 150999305-150999305 on Build GRCh38
rs587631784 Chr.1: 150999385-150999385 on Build GRCh38
rs587661375 Chr.1: 150999480-150999480 on Build GRCh38
rs753270702 Chr.1: 150999362-150999363 on Build GRCh38
rs955753160 Chr.1: 150999519-150999519 on Build GRCh38
rs938686806 Chr.1: 150999371-150999371 on Build GRCh38
rs113120419 Chr.1: 150999466-150999466 on Build GRCh38
rs746845026 Chr.1: 150999361-150999361 on Build GRCh38
rs781162463 Chr.1: 150999427-150999427 on Build GRCh38
rs111942686 Chr.1: 150999380-150999380 on Build GRCh38
rs748381848 Chr.1: 150999365-150999365 on Build GRCh38
rs749861533 Chr.1: 150999347-150999347 on Build GRCh38
rs778114266 Chr.1: 150999531-150999531 on Build GRCh38
rs763801641 Chr.1: 150999325-150999325 on Build GRCh38

View Full Product Details