Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 43992498-43992737 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC24
Entrez Gene ID
149473
Gene Name
coiled-coil domain containing 24
Gene Aliases
-
UniGene
Hs.632394
Gene Chromosome Location
Chr.1: 43991247-43996528 on Build GRCh38
Transcripts
NM_152499.2 XM_017000423.1 XM_017000424.1 XM_017000425.1 XM_017000426.1 XM_017000427.1 XM_017000428.1 XM_017000429.1 XM_017000430.1 XM_017000431.1 XM_017000432.1 XM_017000433.1 XM_017000434.1 XM_017000435.1 XM_017000436.1 XM_017000437.1 XM_017000438.1 XM_017000439.1 XM_017000440.1 XM_017000441.1 XM_017000442.1 XM_017000443.1 XM_017000444.1 XM_017000445.1 XM_017000446.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6317273 c.409G>T p.G137C Chr.1: 43992629-43992629 on Build GRCh38
7284662 c.380C>A p.P127Q Chr.1: 43992600-43992600 on Build GRCh38
5773592 c.402G>A p.M134I Chr.1: 43992622-43992622 on Build GRCh38
426330 c.346C>T p.H116Y Chr.1: 43992566-43992566 on Build GRCh38
7288002 c.384A>T p.E128D Chr.1: 43992604-43992604 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765993845 Chr.1: 43992600-43992600 on Build GRCh38
rs375533554 Chr.1: 43992537-43992537 on Build GRCh38
rs1031172633 Chr.1: 43992608-43992608 on Build GRCh38
rs551341617 Chr.1: 43992685-43992685 on Build GRCh38
rs754286000 Chr.1: 43992616-43992616 on Build GRCh38
rs996682403 Chr.1: 43992573-43992573 on Build GRCh38
rs746279527 Chr.1: 43992622-43992622 on Build GRCh38
rs143854845 Chr.1: 43992604-43992604 on Build GRCh38
rs139084253 Chr.1: 43992589-43992589 on Build GRCh38
rs749832206 Chr.1: 43992538-43992538 on Build GRCh38
rs549535510 Chr.1: 43992523-43992523 on Build GRCh38
rs766614024 Chr.1: 43992559-43992560 on Build GRCh38
rs757827373 Chr.1: 43992617-43992617 on Build GRCh38
rs772322010 Chr.1: 43992558-43992558 on Build GRCh38
rs773809446 Chr.1: 43992672-43992672 on Build GRCh38
rs188098124 Chr.1: 43992544-43992544 on Build GRCh38
rs779307476 Chr.1: 43992547-43992547 on Build GRCh38
rs750033743 Chr.1: 43992519-43992519 on Build GRCh38
rs758179358 Chr.1: 43992521-43992521 on Build GRCh38
rs746087056 Chr.1: 43992549-43992549 on Build GRCh38
rs142803641 Chr.1: 43992571-43992571 on Build GRCh38
rs768814597 Chr.1: 43992583-43992583 on Build GRCh38
rs776369501 Chr.1: 43992584-43992584 on Build GRCh38
rs762660047 Chr.1: 43992598-43992598 on Build GRCh38
rs763290594 Chr.1: 43992530-43992531 on Build GRCh38
rs747472099 Chr.1: 43992639-43992639 on Build GRCh38
rs187616405 Chr.1: 43992629-43992629 on Build GRCh38
rs776834078 Chr.1: 43992635-43992635 on Build GRCh38
rs777863685 Chr.1: 43992636-43992636 on Build GRCh38
rs144458666 Chr.1: 43992614-43992614 on Build GRCh38
rs761859315 Chr.1: 43992587-43992587 on Build GRCh38
rs751067996 Chr.1: 43992526-43992526 on Build GRCh38
rs769896048 Chr.1: 43992637-43992637 on Build GRCh38
rs756620709 Chr.1: 43992527-43992527 on Build GRCh38
rs775619431 Chr.1: 43992568-43992568 on Build GRCh38
rs927095447 Chr.1: 43992677-43992677 on Build GRCh38
rs771218197 Chr.1: 43992546-43992546 on Build GRCh38
rs773121666 Chr.1: 43992649-43992649 on Build GRCh38
rs545179189 Chr.1: 43992594-43992594 on Build GRCh38
rs980159287 Chr.1: 43992676-43992676 on Build GRCh38
rs766230094 Chr.1: 43992669-43992669 on Build GRCh38
rs754405515 Chr.1: 43992611-43992611 on Build GRCh38
rs762788875 Chr.1: 43992663-43992663 on Build GRCh38
rs780096688 Chr.1: 43992627-43992627 on Build GRCh38
rs779217176 Chr.1: 43992618-43992618 on Build GRCh38
rs987985609 Chr.1: 43992696-43992696 on Build GRCh38

View Full Product Details