Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 124417388-124417649 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNTNAP5
Entrez Gene ID
129684
Gene Name
contactin associated protein like 5
Gene Aliases
caspr5
UniGene
Hs.660653
Gene Chromosome Location
Chr.2: 124025287-124921201 on Build GRCh38
Transcripts
NM_130773.3 XM_006712258.1 XM_017003316.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5804048 c.382-2A>T p.? Chr.2: 124417441-124417441 on Build GRCh38
5025703 c.529+2T>C p.? Chr.2: 124417592-124417592 on Build GRCh38
4806487 c.465G>A p.V155V Chr.2: 124417526-124417526 on Build GRCh38
5795223 c.458G>T p.R153L Chr.2: 124417519-124417519 on Build GRCh38
6584251 c.507C>G p.V169V Chr.2: 124417568-124417568 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774904439 Chr.2: 124417444-124417444 on Build GRCh38
rs755160553 Chr.2: 124417559-124417559 on Build GRCh38
rs372513895 Chr.2: 124417606-124417606 on Build GRCh38
rs374763163 Chr.2: 124417476-124417476 on Build GRCh38
rs765263584 Chr.2: 124417473-124417473 on Build GRCh38
rs757899515 Chr.2: 124417596-124417596 on Build GRCh38
rs763713984 Chr.2: 124417467-124417467 on Build GRCh38
rs747572014 Chr.2: 124417607-124417607 on Build GRCh38
rs760393085 Chr.2: 124417519-124417519 on Build GRCh38
rs1050617076 Chr.2: 124417432-124417432 on Build GRCh38
rs191027725 Chr.2: 124417544-124417544 on Build GRCh38
rs770399256 Chr.2: 124417437-124417437 on Build GRCh38
rs751786808 Chr.2: 124417494-124417494 on Build GRCh38
rs547539440 Chr.2: 124417510-124417510 on Build GRCh38
rs776252735 Chr.2: 124417531-124417531 on Build GRCh38
rs375524808 Chr.2: 124417472-124417472 on Build GRCh38
rs761439127 Chr.2: 124417532-124417532 on Build GRCh38
rs779417866 Chr.2: 124417597-124417597 on Build GRCh38
rs773234872 Chr.2: 124417542-124417542 on Build GRCh38
rs972094847 Chr.2: 124417631-124417631 on Build GRCh38
rs768492902 Chr.2: 124417526-124417526 on Build GRCh38
rs188441641 Chr.2: 124417419-124417419 on Build GRCh38
rs760192883 Chr.2: 124417466-124417466 on Build GRCh38
rs749813678 Chr.2: 124417586-124417586 on Build GRCh38
rs775311503 Chr.2: 124417458-124417458 on Build GRCh38
rs777663745 Chr.2: 124417428-124417431 on Build GRCh38
rs368252422 Chr.2: 124417577-124417577 on Build GRCh38
rs568679567 Chr.2: 124417480-124417480 on Build GRCh38
rs769352086 Chr.2: 124417609-124417609 on Build GRCh38
rs766342995 Chr.2: 124417617-124417617 on Build GRCh38
rs775223081 Chr.2: 124417513-124417513 on Build GRCh38
rs765036259 Chr.2: 124417533-124417533 on Build GRCh38
rs751422237 Chr.2: 124417552-124417552 on Build GRCh38
rs774108562 Chr.2: 124417620-124417620 on Build GRCh38
rs755250149 Chr.2: 124417500-124417500 on Build GRCh38
rs762636188 Chr.2: 124417603-124417603 on Build GRCh38
rs748024411 Chr.2: 124417555-124417555 on Build GRCh38
rs769619622 Chr.2: 124417568-124417568 on Build GRCh38
rs746278745 Chr.2: 124417602-124417602 on Build GRCh38
rs961284651 Chr.2: 124417618-124417618 on Build GRCh38
rs781243750 Chr.2: 124417502-124417502 on Build GRCh38
rs959923552 Chr.2: 124417427-124417427 on Build GRCh38
rs750163321 Chr.2: 124417475-124417475 on Build GRCh38
rs767776332 Chr.2: 124417565-124417565 on Build GRCh38
rs753459439 Chr.2: 124417469-124417469 on Build GRCh38
rs771572008 Chr.2: 124417445-124417445 on Build GRCh38
rs766146641 Chr.2: 124417545-124417545 on Build GRCh38
rs778361514 Chr.2: 124417506-124417506 on Build GRCh38
rs200969624 Chr.2: 124417512-124417512 on Build GRCh38
rs770178341 Chr.2: 124417505-124417505 on Build GRCh38
rs889258426 Chr.2: 124417450-124417450 on Build GRCh38
rs369026856 Chr.2: 124417572-124417572 on Build GRCh38
rs370500496 Chr.2: 124417438-124417438 on Build GRCh38
rs772789509 Chr.2: 124417578-124417578 on Build GRCh38
rs183701800 Chr.2: 124417615-124417615 on Build GRCh38
rs142053498 Chr.2: 124417504-124417504 on Build GRCh38
rs772966795 Chr.2: 124417613-124417613 on Build GRCh38

View Full Product Details