Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 132450668-132450899 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJC13
Entrez Gene ID
23317
Gene Name
DnaJ heat shock protein family (Hsp40) member C13
Gene Aliases
PARK21, RME8
UniGene
Hs.12707
Gene Chromosome Location
Chr.3: 132417527-132539032 on Build GRCh38
Transcripts
NM_015268.3 XM_005247245.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6504146 c.403G>A p.G135R Chr.3: 132450713-132450713 on Build GRCh38
222852 c.499G>A p.G167R Chr.3: 132450809-132450809 on Build GRCh38
4883487 c.488C>T p.S163L Chr.3: 132450798-132450798 on Build GRCh38
1693473 c.455A>G p.D152G Chr.3: 132450765-132450765 on Build GRCh38
3121831 c.419T>G p.I140S Chr.3: 132450729-132450729 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1045395543 Chr.3: 132450701-132450701 on Build GRCh38
rs761733112 Chr.3: 132450783-132450783 on Build GRCh38
rs770797343 Chr.3: 132450857-132450858 on Build GRCh38
rs200307667 Chr.3: 132450858-132450858 on Build GRCh38
rs936510202 Chr.3: 132450770-132450770 on Build GRCh38
rs766088625 Chr.3: 132450796-132450796 on Build GRCh38
rs375141561 Chr.3: 132450777-132450777 on Build GRCh38
rs774305258 Chr.3: 132450858-132450861 on Build GRCh38
rs796549071 Chr.3: 132450862-132450863 on Build GRCh38
rs770039601 Chr.3: 132450734-132450734 on Build GRCh38
rs771261509 Chr.3: 132450704-132450704 on Build GRCh38
rs530036442 Chr.3: 132450792-132450792 on Build GRCh38
rs80328158 Chr.3: 132450861-132450861 on Build GRCh38
rs376841896 Chr.3: 132450821-132450821 on Build GRCh38
rs371204623 Chr.3: 132450769-132450769 on Build GRCh38
rs1051518804 Chr.3: 132450815-132450815 on Build GRCh38
rs767254248 Chr.3: 132450861-132450862 on Build GRCh38
rs760338262 Chr.3: 132450794-132450794 on Build GRCh38
rs768662545 Chr.3: 132450773-132450773 on Build GRCh38
rs752180538 Chr.3: 132450829-132450829 on Build GRCh38
rs149480465 Chr.3: 132450732-132450732 on Build GRCh38
rs759359528 Chr.3: 132450860-132450861 on Build GRCh38
rs754551314 Chr.3: 132450805-132450805 on Build GRCh38
rs34198565 Chr.3: 132450862-132450863 on Build GRCh38
rs757755351 Chr.3: 132450834-132450834 on Build GRCh38
rs753475492 Chr.3: 132450803-132450803 on Build GRCh38
rs781618558 Chr.3: 132450714-132450714 on Build GRCh38
rs989605527 Chr.3: 132450703-132450703 on Build GRCh38
rs559364378 Chr.3: 132450744-132450744 on Build GRCh38
rs200011659 Chr.3: 132450862-132450862 on Build GRCh38
rs112154025 Chr.3: 132450862-132450862 on Build GRCh38

View Full Product Details