Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 144842471-144842742 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COMMD5
Entrez Gene ID
28991
Gene Name
COMM domain containing 5
Gene Aliases
HCARG, HT002
UniGene
Hs.493218
Gene Chromosome Location
Chr.8: 144837978-144853578 on Build GRCh38
Transcripts
NM_001282795.1 NM_001282796.1 NM_001282797.1 NM_003416.3 XM_006716654.2 XM_006716656.3 XM_011517291.2 XM_011517292.2 XM_011517293.2 XM_011517294.2 XM_011517295.2 XM_011517296.2 XM_011517297.2 XM_017013813.1 XM_017013814.1 XM_017013815.1 XM_017013816.1 XM_017013817.1

Gene Symbol
ZNF7
Entrez Gene ID
7553
Gene Name
zinc finger protein 7
Gene Aliases
HF.16, KOX4, zf30
UniGene
Hs.493218
Gene Chromosome Location
Chr.8: 144827500-144848070 on Build GRCh38
Transcripts
NM_001282795.1 NM_001282796.1 NM_001282797.1 NM_003416.3 XM_006716654.2 XM_006716656.3 XM_011517291.2 XM_011517292.2 XM_011517293.2 XM_011517294.2 XM_011517295.2 XM_011517296.2 XM_011517297.2 XM_017013813.1 XM_017013814.1 XM_017013815.1 XM_017013816.1 XM_017013817.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5459706 c.1465C>T p.R489* Chr.8: 144842572-144842572 on Build GRCh38
6832527 c.1591G>A p.G531R Chr.8: 144842698-144842698 on Build GRCh38
1455728 c.1382G>T p.R461I Chr.8: 144842489-144842489 on Build GRCh38
1097328 c.1520G>T p.S507I Chr.8: 144842627-144842627 on Build GRCh38
1455730 c.1604G>T p.S535I Chr.8: 144842711-144842711 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs34385444 Chr.8: 144842711-144842711 on Build GRCh38
rs757080895 Chr.8: 144842664-144842664 on Build GRCh38
rs372508675 Chr.8: 144842705-144842705 on Build GRCh38
rs746869135 Chr.8: 144842573-144842573 on Build GRCh38
rs762152302 Chr.8: 144842509-144842509 on Build GRCh38
rs374481400 Chr.8: 144842597-144842597 on Build GRCh38
rs747568869 Chr.8: 144842698-144842698 on Build GRCh38
rs765758845 Chr.8: 144842532-144842532 on Build GRCh38
rs779928022 Chr.8: 144842580-144842580 on Build GRCh38
rs572997816 Chr.8: 144842620-144842620 on Build GRCh38
rs771107932 Chr.8: 144842620-144842621 on Build GRCh38
rs749394485 Chr.8: 144842675-144842675 on Build GRCh38
rs201846657 Chr.8: 144842697-144842697 on Build GRCh38
rs763939395 Chr.8: 144842637-144842637 on Build GRCh38
rs779156102 Chr.8: 144842683-144842683 on Build GRCh38
rs574491846 Chr.8: 144842598-144842598 on Build GRCh38
rs773732807 Chr.8: 144842601-144842601 on Build GRCh38
rs749537107 Chr.8: 144842582-144842582 on Build GRCh38
rs142485502 Chr.8: 144842547-144842547 on Build GRCh38
rs748321934 Chr.8: 144842689-144842689 on Build GRCh38
rs781175829 Chr.8: 144842673-144842673 on Build GRCh38
rs765141390 Chr.8: 144842557-144842557 on Build GRCh38
rs774639361 Chr.8: 144842496-144842496 on Build GRCh38
rs777903102 Chr.8: 144842572-144842572 on Build GRCh38
rs759078046 Chr.8: 144842614-144842614 on Build GRCh38
rs145861591 Chr.8: 144842513-144842513 on Build GRCh38
rs564102152 Chr.8: 144842565-144842565 on Build GRCh38
rs768924132 Chr.8: 144842590-144842590 on Build GRCh38
rs997657853 Chr.8: 144842537-144842537 on Build GRCh38
rs752879403 Chr.8: 144842525-144842525 on Build GRCh38
rs774549474 Chr.8: 144842593-144842593 on Build GRCh38
rs759160918 Chr.8: 144842717-144842717 on Build GRCh38
rs765918704 Chr.8: 144842603-144842603 on Build GRCh38
rs753559832 Chr.8: 144842534-144842534 on Build GRCh38
rs751515165 Chr.8: 144842644-144842644 on Build GRCh38
rs752370306 Chr.8: 144842621-144842621 on Build GRCh38
rs748550751 Chr.8: 144842596-144842596 on Build GRCh38
rs777122818 Chr.8: 144842646-144842649 on Build GRCh38
rs367564747 Chr.8: 144842608-144842608 on Build GRCh38
rs946662077 Chr.8: 144842524-144842524 on Build GRCh38
rs760284010 Chr.8: 144842528-144842528 on Build GRCh38
rs886549305 Chr.8: 144842594-144842594 on Build GRCh38
rs756244783 Chr.8: 144842577-144842577 on Build GRCh38
rs758287767 Chr.8: 144842630-144842630 on Build GRCh38
rs368317181 Chr.8: 144842701-144842701 on Build GRCh38
rs1006602328 Chr.8: 144842670-144842670 on Build GRCh38
rs769065860 Chr.8: 144842493-144842493 on Build GRCh38
rs372104925 Chr.8: 144842694-144842694 on Build GRCh38
rs148588579 Chr.8: 144842529-144842529 on Build GRCh38
rs2975302 Chr.8: 144842682-144842682 on Build GRCh38

View Full Product Details