Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 25742743-25743000 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASXL2
Entrez Gene ID
55252
Gene Name
additional sex combs like 2, transcriptional regulator
Gene Aliases
ASXH2
UniGene
Hs.119815
Gene Chromosome Location
Chr.2: 25733758-25878485 on Build GRCh38
Transcripts
NM_018263.4 XM_006712039.3 XM_006712040.2 XM_011532950.2 XM_011532951.2 XM_017004429.1 XM_017004430.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
418775 c.1926G>C p.L642F Chr.2: 25742860-25742860 on Build GRCh38
6888416 c.1796C>T p.S599F Chr.2: 25742990-25742990 on Build GRCh38
4776611 c.3532G>C p.E1178Q Chr.2: 25742805-25742805 on Build GRCh38
4093291 c.3396C>T p.G1132G Chr.2: 25742941-25742941 on Build GRCh38
5807507 c.3521G>A p.S1174N Chr.2: 25742816-25742816 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369840548 Chr.2: 25742780-25742780 on Build GRCh38
rs559319074 Chr.2: 25742799-25742799 on Build GRCh38
rs756706843 Chr.2: 25742927-25742927 on Build GRCh38
rs766551006 Chr.2: 25742858-25742858 on Build GRCh38
rs746779573 Chr.2: 25742937-25742937 on Build GRCh38
rs879044390 Chr.2: 25742853-25742853 on Build GRCh38
rs777213228 Chr.2: 25742781-25742781 on Build GRCh38
rs753079173 Chr.2: 25742895-25742895 on Build GRCh38
rs759907363 Chr.2: 25742950-25742950 on Build GRCh38
rs773618990 Chr.2: 25742846-25742846 on Build GRCh38
rs774517028 Chr.2: 25742949-25742949 on Build GRCh38
rs745874693 Chr.2: 25742810-25742810 on Build GRCh38
rs771706903 Chr.2: 25742813-25742813 on Build GRCh38
rs377352572 Chr.2: 25742892-25742892 on Build GRCh38
rs752000777 Chr.2: 25742873-25742873 on Build GRCh38
rs778498247 Chr.2: 25742928-25742928 on Build GRCh38
rs373106531 Chr.2: 25742851-25742851 on Build GRCh38
rs745724932 Chr.2: 25742930-25742930 on Build GRCh38
rs748771560 Chr.2: 25742791-25742791 on Build GRCh38
rs749004587 Chr.2: 25742902-25742902 on Build GRCh38
rs773567574 Chr.2: 25742943-25742943 on Build GRCh38
rs983776205 Chr.2: 25742842-25742842 on Build GRCh38
rs775377109 Chr.2: 25742936-25742936 on Build GRCh38
rs765581039 Chr.2: 25742844-25742844 on Build GRCh38
rs777370673 Chr.2: 25742901-25742901 on Build GRCh38
rs747728489 Chr.2: 25742775-25742775 on Build GRCh38
rs756517436 Chr.2: 25742899-25742899 on Build GRCh38
rs938580887 Chr.2: 25742910-25742910 on Build GRCh38
rs775430297 Chr.2: 25742814-25742814 on Build GRCh38
rs766316873 Chr.2: 25742947-25742947 on Build GRCh38
rs530281362 Chr.2: 25742935-25742935 on Build GRCh38
rs764182016 Chr.2: 25742944-25742944 on Build GRCh38
rs768613233 Chr.2: 25742939-25742939 on Build GRCh38
rs774914918 Chr.2: 25742801-25742803 on Build GRCh38
rs774150572 Chr.2: 25742802-25742802 on Build GRCh38

View Full Product Details