Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 151092989-151093254 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GABPB2
Entrez Gene ID
126626
Gene Name
GA binding protein transcription factor beta subunit 2
Gene Aliases
GABPB-2
UniGene
Hs.15671
Gene Chromosome Location
Chr.1: 151070180-151125545 on Build GRCh38
Transcripts
NM_001323906.1 NM_001323907.1 NM_001323908.1 NM_001323909.1 NM_001323910.1 NM_001323911.1 NM_001323912.1 NM_001323913.1 NM_144618.2 XM_017000246.1 XM_017000247.1 XM_017000248.1 XM_017000249.1 XM_017000250.1 XM_017000251.1 XM_017000252.1 XM_017000253.1 XM_017000254.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6476638 c.338A>G p.E113G Chr.1: 151093253-151093253 on Build GRCh38
1295021 c.291G>T p.V97V Chr.1: 151093206-151093206 on Build GRCh38
6326019 c.277-1G>A p.? Chr.1: 151093191-151093191 on Build GRCh38
6345259 c.329G>T p.W110L Chr.1: 151093244-151093244 on Build GRCh38
6392639 c.295G>A p.A99T Chr.1: 151093210-151093210 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756457147 Chr.1: 151093232-151093232 on Build GRCh38
rs1007420877 Chr.1: 151093019-151093019 on Build GRCh38
rs371525284 Chr.1: 151093146-151093146 on Build GRCh38
rs1000287154 Chr.1: 151093129-151093129 on Build GRCh38
rs587761079 Chr.1: 151093018-151093018 on Build GRCh38
rs765578971 Chr.1: 151093150-151093150 on Build GRCh38
rs747860758 Chr.1: 151093147-151093149 on Build GRCh38
rs587619339 Chr.1: 151093131-151093131 on Build GRCh38
rs367812321 Chr.1: 151093141-151093141 on Build GRCh38
rs748477841 Chr.1: 151093223-151093223 on Build GRCh38
rs750634304 Chr.1: 151093157-151093157 on Build GRCh38
rs937486318 Chr.1: 151093175-151093175 on Build GRCh38
rs956243036 Chr.1: 151093204-151093204 on Build GRCh38
rs755438708 Chr.1: 151093196-151093196 on Build GRCh38
rs376497930 Chr.1: 151093115-151093115 on Build GRCh38
rs587743802 Chr.1: 151093077-151093077 on Build GRCh38
rs769277749 Chr.1: 151093168-151093169 on Build GRCh38
rs766543478 Chr.1: 151093169-151093169 on Build GRCh38
rs1042811337 Chr.1: 151093080-151093080 on Build GRCh38
rs141995070 Chr.1: 151093070-151093070 on Build GRCh38
rs904531717 Chr.1: 151093087-151093087 on Build GRCh38
rs758589650 Chr.1: 151093158-151093158 on Build GRCh38
rs781675381 Chr.1: 151093222-151093222 on Build GRCh38
rs752171961 Chr.1: 151093185-151093185 on Build GRCh38

View Full Product Details