Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 91942812-91943075 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBLN5
Entrez Gene ID
10516
Gene Name
fibulin 5
Gene Aliases
ADCL2, ARCL1A, ARMD3, DANCE, EVEC, FIBL-5, HNARMD, UP50
UniGene
Hs.332708
Gene Chromosome Location
Chr.14: 91869411-91947702 on Build GRCh38
Transcripts
NM_006329.3 XM_005267267.3 XM_011536356.1 XM_011536357.1 XM_011536358.1 XM_017020929.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6214940 c.72+5G>C p.? Chr.14: 91942902-91942902 on Build GRCh38
6425161 c.62G>A p.G21E Chr.14: 91942917-91942917 on Build GRCh38
4959658 c.59C>T p.P20L Chr.14: 91942920-91942920 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs570162560 Chr.14: 91942851-91942851 on Build GRCh38
rs986108144 Chr.14: 91942843-91942843 on Build GRCh38
rs770470522 Chr.14: 91942992-91942992 on Build GRCh38
rs1015326450 Chr.14: 91943032-91943032 on Build GRCh38
rs778999697 Chr.14: 91942918-91942918 on Build GRCh38
rs373067056 Chr.14: 91943031-91943031 on Build GRCh38
rs113167395 Chr.14: 91942881-91942881 on Build GRCh38
rs772595054 Chr.14: 91942972-91942972 on Build GRCh38
rs1002873645 Chr.14: 91943006-91943006 on Build GRCh38
rs569683263 Chr.14: 91942916-91942916 on Build GRCh38
rs748040891 Chr.14: 91942925-91942925 on Build GRCh38
rs10135192 Chr.14: 91943048-91943048 on Build GRCh38
rs561218063 Chr.14: 91942833-91942833 on Build GRCh38
rs149904034 Chr.14: 91943042-91943042 on Build GRCh38
rs923895835 Chr.14: 91943046-91943046 on Build GRCh38
rs896157646 Chr.14: 91943015-91943015 on Build GRCh38
rs1016427895 Chr.14: 91942984-91942984 on Build GRCh38
rs953952718 Chr.14: 91943053-91943053 on Build GRCh38
rs973948359 Chr.14: 91942837-91942837 on Build GRCh38
rs950994101 Chr.14: 91942933-91942933 on Build GRCh38
rs184502226 Chr.14: 91942838-91942838 on Build GRCh38
rs751863037 Chr.14: 91942872-91942872 on Build GRCh38
rs896984600 Chr.14: 91943037-91943037 on Build GRCh38
rs914056629 Chr.14: 91942957-91942957 on Build GRCh38
rs961216486 Chr.14: 91942832-91942832 on Build GRCh38
rs565761187 Chr.14: 91942882-91942882 on Build GRCh38
rs532141720 Chr.14: 91942841-91942841 on Build GRCh38
rs150715038 Chr.14: 91942902-91942902 on Build GRCh38
rs978037296 Chr.14: 91943045-91943045 on Build GRCh38
rs990880040 Chr.14: 91943004-91943004 on Build GRCh38
rs371468999 Chr.14: 91942861-91942861 on Build GRCh38
rs945558917 Chr.14: 91942974-91942974 on Build GRCh38
rs17127768 Chr.14: 91942989-91942989 on Build GRCh38
rs536696027 Chr.14: 91943026-91943026 on Build GRCh38
rs1026941629 Chr.14: 91942932-91942932 on Build GRCh38
rs904145472 Chr.14: 91942991-91942991 on Build GRCh38
rs1040501479 Chr.14: 91942889-91942889 on Build GRCh38
rs565448041 Chr.14: 91943049-91943049 on Build GRCh38
rs766688945 Chr.14: 91942873-91942873 on Build GRCh38
rs749068976 Chr.14: 91942900-91942900 on Build GRCh38
rs771672858 Chr.14: 91942969-91942969 on Build GRCh38
rs78978877 Chr.14: 91943007-91943007 on Build GRCh38

View Full Product Details