Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 110904557-110904826 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC63
Entrez Gene ID
160762
Gene Name
coiled-coil domain containing 63
Gene Aliases
ODA5
UniGene
Hs.437141
Gene Chromosome Location
Chr.12: 110844489-110907535 on Build GRCh38
Transcripts
NM_001286243.1 NM_001286244.1 NM_152591.2 XM_006719262.1 XM_006719263.2 XM_011537999.1 XM_011538001.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6478782 c.1521C>A p.D507E Chr.12: 110904766-110904766 on Build GRCh38
5590089 c.1398C>T p.I466I Chr.12: 110904643-110904643 on Build GRCh38
6420271 c.1456G>A p.G486S Chr.12: 110904701-110904701 on Build GRCh38
4526749 c.1417G>A p.E473K Chr.12: 110904662-110904662 on Build GRCh38
6537830 c.1473C>G p.L491L Chr.12: 110904718-110904718 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768700196 Chr.12: 110904622-110904636 on Build GRCh38
rs747497345 Chr.12: 110904715-110904715 on Build GRCh38
rs752575542 Chr.12: 110904656-110904656 on Build GRCh38
rs747870492 Chr.12: 110904743-110904743 on Build GRCh38
rs763268920 Chr.12: 110904780-110904780 on Build GRCh38
rs932862877 Chr.12: 110904758-110904758 on Build GRCh38
rs139596933 Chr.12: 110904686-110904686 on Build GRCh38
rs777701219 Chr.12: 110904793-110904793 on Build GRCh38
rs775250425 Chr.12: 110904781-110904781 on Build GRCh38
rs750494632 Chr.12: 110904699-110904699 on Build GRCh38
rs757358650 Chr.12: 110904610-110904610 on Build GRCh38
rs199586446 Chr.12: 110904651-110904651 on Build GRCh38
rs774187392 Chr.12: 110904772-110904772 on Build GRCh38
rs747336794 Chr.12: 110904630-110904630 on Build GRCh38
rs770306431 Chr.12: 110904721-110904721 on Build GRCh38
rs372078813 Chr.12: 110904692-110904692 on Build GRCh38
rs148758697 Chr.12: 110904775-110904775 on Build GRCh38
rs369852755 Chr.12: 110904742-110904742 on Build GRCh38
rs34300256 Chr.12: 110904754-110904754 on Build GRCh38
rs748821671 Chr.12: 110904761-110904761 on Build GRCh38
rs146312173 Chr.12: 110904639-110904639 on Build GRCh38
rs763052565 Chr.12: 110904660-110904660 on Build GRCh38
rs1024229644 Chr.12: 110904695-110904695 on Build GRCh38
rs1008614863 Chr.12: 110904606-110904606 on Build GRCh38
rs758434248 Chr.12: 110904701-110904701 on Build GRCh38
rs971639523 Chr.12: 110904689-110904689 on Build GRCh38
rs773685692 Chr.12: 110904766-110904766 on Build GRCh38
rs200583861 Chr.12: 110904594-110904594 on Build GRCh38
rs373980752 Chr.12: 110904687-110904687 on Build GRCh38
rs375752382 Chr.12: 110904705-110904705 on Build GRCh38
rs535552535 Chr.12: 110904706-110904706 on Build GRCh38
rs761841287 Chr.12: 110904698-110904699 on Build GRCh38
rs142416969 Chr.12: 110904782-110904782 on Build GRCh38
rs1051355785 Chr.12: 110904763-110904763 on Build GRCh38
rs574092598 Chr.12: 110904641-110904641 on Build GRCh38
rs762960378 Chr.12: 110904801-110904801 on Build GRCh38
rs115334501 Chr.12: 110904718-110904718 on Build GRCh38
rs914387399 Chr.12: 110904798-110904801 on Build GRCh38
rs754736027 Chr.12: 110904588-110904588 on Build GRCh38
rs766763794 Chr.12: 110904798-110904799 on Build GRCh38
rs138877502 Chr.12: 110904611-110904611 on Build GRCh38
rs769568156 Chr.12: 110904746-110904746 on Build GRCh38
rs951040033 Chr.12: 110904730-110904730 on Build GRCh38
rs116032516 Chr.12: 110904776-110904776 on Build GRCh38
rs533169835 Chr.12: 110904690-110904690 on Build GRCh38
rs766233341 Chr.12: 110904665-110904665 on Build GRCh38
rs375870661 Chr.12: 110904614-110904614 on Build GRCh38
rs201517807 Chr.12: 110904676-110904676 on Build GRCh38
rs774288568 Chr.12: 110904675-110904675 on Build GRCh38
rs374279689 Chr.12: 110904794-110904794 on Build GRCh38
rs573564509 Chr.12: 110904590-110904590 on Build GRCh38
rs141920499 Chr.12: 110904622-110904622 on Build GRCh38
rs115635364 Chr.12: 110904791-110904791 on Build GRCh38
rs772838782 Chr.12: 110904657-110904657 on Build GRCh38
rs769004212 Chr.12: 110904634-110904634 on Build GRCh38

View Full Product Details