Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 207712092-207712327 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCNYL1
Entrez Gene ID
151195
Gene Name
cyclin Y like 1
Gene Aliases
-
UniGene
Hs.471234
Gene Chromosome Location
Chr.2: 207711540-207756174 on Build GRCh38
Transcripts
NM_001142300.1 NM_152523.2 XM_005246343.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4427670 c.7delG p.E3fs*3 Chr.2: 207712113-207712113 on Build GRCh38
4427669 c.217delG p.E73fs*3 Chr.2: 207712113-207712113 on Build GRCh38
6623130 c.1A>G p.M1V Chr.2: 207712107-207712107 on Build GRCh38
6528711 c.7G>C p.E3Q Chr.2: 207712113-207712113 on Build GRCh38
6623129 c.211A>G p.M71V Chr.2: 207712107-207712107 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs564264026 Chr.2: 207712285-207712285 on Build GRCh38
rs1031181833 Chr.2: 207712164-207712164 on Build GRCh38
rs762371251 Chr.2: 207712153-207712153 on Build GRCh38
rs200665125 Chr.2: 207712129-207712129 on Build GRCh38
rs980854228 Chr.2: 207712218-207712218 on Build GRCh38
rs190940419 Chr.2: 207712300-207712300 on Build GRCh38
rs929649621 Chr.2: 207712266-207712266 on Build GRCh38
rs767617903 Chr.2: 207712154-207712154 on Build GRCh38
rs1053906164 Chr.2: 207712220-207712220 on Build GRCh38
rs892214461 Chr.2: 207712222-207712222 on Build GRCh38
rs1024563827 Chr.2: 207712233-207712233 on Build GRCh38
rs758172966 Chr.2: 207712219-207712219 on Build GRCh38
rs773210102 Chr.2: 207712160-207712160 on Build GRCh38
rs866833226 Chr.2: 207712248-207712248 on Build GRCh38
rs766538547 Chr.2: 207712163-207712163 on Build GRCh38
rs59050344 Chr.2: 207712210-207712210 on Build GRCh38
rs760918911 Chr.2: 207712161-207712161 on Build GRCh38
rs1050700609 Chr.2: 207712184-207712184 on Build GRCh38
rs1025082322 Chr.2: 207712173-207712173 on Build GRCh38
rs756833163 Chr.2: 207712122-207712122 on Build GRCh38
rs769105976 Chr.2: 207712142-207712142 on Build GRCh38
rs974019741 Chr.2: 207712227-207712227 on Build GRCh38
rs752491068 Chr.2: 207712166-207712166 on Build GRCh38
rs528538954 Chr.2: 207712238-207712238 on Build GRCh38
rs923329768 Chr.2: 207712174-207712174 on Build GRCh38
rs542971429 Chr.2: 207712170-207712170 on Build GRCh38
rs775897174 Chr.2: 207712136-207712136 on Build GRCh38
rs376226089 Chr.2: 207712180-207712180 on Build GRCh38
rs752498507 Chr.2: 207712162-207712162 on Build GRCh38
rs375999071 Chr.2: 207712134-207712134 on Build GRCh38
rs774841379 Chr.2: 207712148-207712148 on Build GRCh38
rs866178406 Chr.2: 207712168-207712168 on Build GRCh38
rs369009815 Chr.2: 207712150-207712150 on Build GRCh38
rs962502401 Chr.2: 207712225-207712225 on Build GRCh38
rs940778108 Chr.2: 207712151-207712151 on Build GRCh38
rs1039417548 Chr.2: 207712303-207712303 on Build GRCh38
rs764363949 Chr.2: 207712245-207712245 on Build GRCh38
rs952766742 Chr.2: 207712288-207712288 on Build GRCh38
rs906201382 Chr.2: 207712240-207712240 on Build GRCh38
rs545566445 Chr.2: 207712253-207712253 on Build GRCh38
rs200116272 Chr.2: 207712130-207712130 on Build GRCh38
rs749687477 Chr.2: 207712137-207712137 on Build GRCh38
rs758371445 Chr.2: 207712109-207712109 on Build GRCh38
rs904039856 Chr.2: 207712305-207712305 on Build GRCh38
rs543689322 Chr.2: 207712112-207712112 on Build GRCh38
rs986857592 Chr.2: 207712282-207712282 on Build GRCh38
rs369686216 Chr.2: 207712111-207712111 on Build GRCh38

View Full Product Details