Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 140672369-140672642 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DND1
Entrez Gene ID
373863
Gene Name
DND microRNA-mediated repression inhibitor 1
Gene Aliases
RBMS4
UniGene
Hs.591262
Gene Chromosome Location
Chr.5: 140670796-140673586 on Build GRCh38
Transcripts
NM_194249.2 XM_005268469.2 XM_017009600.1

Gene Symbol
WDR55
Entrez Gene ID
54853
Gene Name
WD repeat domain 55
Gene Aliases
-
UniGene
Hs.286261
Gene Chromosome Location
Chr.5: 140664799-140674119 on Build GRCh38
Transcripts
NM_194249.2 XM_005268469.2 XM_017009600.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1292030 c.535G>T p.G179W Chr.5: 140672514-140672514 on Build GRCh38
7301339 c.430G>T p.G144C Chr.5: 140672619-140672619 on Build GRCh38
6403596 c.461T>C p.L154P Chr.5: 140672588-140672588 on Build GRCh38
4606998 c.407A>G p.E136G Chr.5: 140672642-140672642 on Build GRCh38
6408630 c.444T>A p.N148K Chr.5: 140672605-140672605 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs199725201 Chr.5: 140672619-140672619 on Build GRCh38
rs1008578743 Chr.5: 140672506-140672506 on Build GRCh38
rs374524765 Chr.5: 140672482-140672482 on Build GRCh38
rs751456989 Chr.5: 140672399-140672399 on Build GRCh38
rs769284305 Chr.5: 140672557-140672557 on Build GRCh38
rs752612450 Chr.5: 140672589-140672589 on Build GRCh38
rs375656093 Chr.5: 140672465-140672465 on Build GRCh38
rs761457502 Chr.5: 140672485-140672485 on Build GRCh38
rs766056441 Chr.5: 140672570-140672570 on Build GRCh38
rs762290232 Chr.5: 140672416-140672416 on Build GRCh38
rs767554500 Chr.5: 140672514-140672514 on Build GRCh38
rs778003256 Chr.5: 140672522-140672522 on Build GRCh38
rs201909457 Chr.5: 140672613-140672613 on Build GRCh38
rs752706118 Chr.5: 140672519-140672519 on Build GRCh38
rs749478856 Chr.5: 140672526-140672526 on Build GRCh38
rs754021893 Chr.5: 140672604-140672604 on Build GRCh38
rs527897702 Chr.5: 140672609-140672609 on Build GRCh38
rs762567505 Chr.5: 140672562-140672562 on Build GRCh38
rs867373147 Chr.5: 140672598-140672598 on Build GRCh38
rs756034452 Chr.5: 140672590-140672590 on Build GRCh38
rs751402051 Chr.5: 140672502-140672502 on Build GRCh38
rs757578310 Chr.5: 140672528-140672528 on Build GRCh38
rs761021121 Chr.5: 140672488-140672488 on Build GRCh38
rs764223305 Chr.5: 140672591-140672591 on Build GRCh38
rs754836948 Chr.5: 140672504-140672504 on Build GRCh38
rs774889948 Chr.5: 140672466-140672466 on Build GRCh38
rs776126223 Chr.5: 140672545-140672545 on Build GRCh38
rs143867711 Chr.5: 140672565-140672565 on Build GRCh38
rs35212498 Chr.5: 140672591-140672591 on Build GRCh38
rs759432597 Chr.5: 140672577-140672577 on Build GRCh38
rs371022226 Chr.5: 140672521-140672521 on Build GRCh38
rs779400219 Chr.5: 140672536-140672536 on Build GRCh38
rs768077931 Chr.5: 140672428-140672428 on Build GRCh38
rs774124510 Chr.5: 140672571-140672571 on Build GRCh38
rs754963510 Chr.5: 140672414-140672414 on Build GRCh38
rs779345075 Chr.5: 140672612-140672612 on Build GRCh38
rs2530243 Chr.5: 140672398-140672398 on Build GRCh38
rs781107278 Chr.5: 140672417-140672417 on Build GRCh38
rs374440658 Chr.5: 140672556-140672556 on Build GRCh38
rs373809626 Chr.5: 140672464-140672464 on Build GRCh38
rs370271469 Chr.5: 140672439-140672439 on Build GRCh38
rs978440615 Chr.5: 140672599-140672599 on Build GRCh38
rs772684319 Chr.5: 140672479-140672479 on Build GRCh38
rs371133972 Chr.5: 140672477-140672477 on Build GRCh38
rs567008499 Chr.5: 140672579-140672579 on Build GRCh38
rs747486447 Chr.5: 140672552-140672552 on Build GRCh38
rs772456914 Chr.5: 140672542-140672542 on Build GRCh38
rs772727815 Chr.5: 140672559-140672559 on Build GRCh38
rs377110867 Chr.5: 140672421-140672421 on Build GRCh38
rs368366843 Chr.5: 140672500-140672500 on Build GRCh38
rs552100499 Chr.5: 140672469-140672469 on Build GRCh38

View Full Product Details