Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 38403771-38404012 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EGFLAM
Entrez Gene ID
133584
Gene Name
EGF like, fibronectin type III and laminin G domains
Gene Aliases
AGRINL, AGRNL, PIKA
UniGene
Hs.20103
Gene Chromosome Location
Chr.5: 38258409-38465480 on Build GRCh38
Transcripts
NM_001205301.1 NM_152403.3 NM_182798.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7186952 c.10+3A>G p.? Chr.5: 38403894-38403894 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs764738644 Chr.5: 38403882-38403882 on Build GRCh38
rs749898074 Chr.5: 38403886-38403886 on Build GRCh38
rs145217235 Chr.5: 38403907-38403907 on Build GRCh38
rs771962133 Chr.5: 38403910-38403910 on Build GRCh38
rs780558273 Chr.5: 38403875-38403880 on Build GRCh38
rs772023275 Chr.5: 38403839-38403839 on Build GRCh38
rs185324186 Chr.5: 38403849-38403849 on Build GRCh38
rs779445311 Chr.5: 38403897-38403897 on Build GRCh38
rs776575626 Chr.5: 38403935-38403935 on Build GRCh38
rs1025667277 Chr.5: 38403929-38403929 on Build GRCh38
rs969431004 Chr.5: 38403979-38403979 on Build GRCh38
rs189108196 Chr.5: 38403985-38403985 on Build GRCh38
rs370654364 Chr.5: 38403856-38403856 on Build GRCh38
rs772848942 Chr.5: 38403990-38403990 on Build GRCh38
rs765629205 Chr.5: 38403866-38403866 on Build GRCh38
rs1038637015 Chr.5: 38403908-38403908 on Build GRCh38
rs368778812 Chr.5: 38403855-38403855 on Build GRCh38
rs1035607150 Chr.5: 38403874-38403874 on Build GRCh38
rs181085593 Chr.5: 38403986-38403986 on Build GRCh38
rs759688672 Chr.5: 38403862-38403862 on Build GRCh38
rs200600181 Chr.5: 38403878-38403878 on Build GRCh38
rs763489407 Chr.5: 38403879-38403879 on Build GRCh38
rs542260540 Chr.5: 38403833-38403833 on Build GRCh38
rs754660449 Chr.5: 38403905-38403905 on Build GRCh38
rs373714860 Chr.5: 38403840-38403840 on Build GRCh38
rs753466856 Chr.5: 38403902-38403902 on Build GRCh38
rs200814616 Chr.5: 38403896-38403896 on Build GRCh38
rs1008319318 Chr.5: 38403984-38403984 on Build GRCh38
rs778687231 Chr.5: 38403831-38403831 on Build GRCh38
rs747042906 Chr.5: 38403919-38403919 on Build GRCh38
rs960943481 Chr.5: 38403945-38403945 on Build GRCh38
rs777829146 Chr.5: 38403911-38403911 on Build GRCh38
rs964095041 Chr.5: 38403989-38403989 on Build GRCh38
rs769488550 Chr.5: 38403810-38403810 on Build GRCh38
rs770982251 Chr.5: 38403933-38403933 on Build GRCh38

View Full Product Details