Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 65581801-65582026 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EHBP1L1
Entrez Gene ID
254102
Gene Name
EH domain binding protein 1 like 1
Gene Aliases
-
UniGene
Hs.502867
Gene Chromosome Location
Chr.11: 65576034-65592650 on Build GRCh38
Transcripts
NM_001099409.1 XM_005273867.3 XM_005273868.3 XM_005273869.3 XM_005273870.3 XM_006718486.3 XM_006718487.3 XM_017017467.1 XM_017017468.1 XM_017017469.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6250502 c.1140G>T p.E380D Chr.11: 65581812-65581812 on Build GRCh38
5552208 c.1273G>T p.D425Y Chr.11: 65581945-65581945 on Build GRCh38
5970289 c.1132G>A p.A378T Chr.11: 65581804-65581804 on Build GRCh38
118293 c.1224A>G p.S408S Chr.11: 65581896-65581896 on Build GRCh38
3810056 c.1170T>G p.S390R Chr.11: 65581842-65581842 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749015483 Chr.11: 65581842-65581842 on Build GRCh38
rs376275465 Chr.11: 65581859-65581859 on Build GRCh38
rs751402784 Chr.11: 65581973-65581973 on Build GRCh38
rs187871743 Chr.11: 65581980-65581980 on Build GRCh38
rs759818285 Chr.11: 65581954-65581954 on Build GRCh38
rs772661889 Chr.11: 65581920-65581920 on Build GRCh38
rs779679767 Chr.11: 65581841-65581841 on Build GRCh38
rs755731960 Chr.11: 65581837-65581837 on Build GRCh38
rs765309971 Chr.11: 65581959-65581959 on Build GRCh38
rs998227406 Chr.11: 65581941-65581941 on Build GRCh38
rs368161955 Chr.11: 65581981-65581981 on Build GRCh38
rs753336840 Chr.11: 65581996-65581996 on Build GRCh38
rs778387395 Chr.11: 65581998-65581998 on Build GRCh38
rs780953961 Chr.11: 65581833-65581833 on Build GRCh38
rs749974288 Chr.11: 65581834-65581834 on Build GRCh38
rs1028290059 Chr.11: 65581962-65581962 on Build GRCh38
rs779993363 Chr.11: 65581895-65581895 on Build GRCh38
rs757225418 Chr.11: 65581829-65581829 on Build GRCh38
rs779345399 Chr.11: 65581871-65581871 on Build GRCh38
rs954286034 Chr.11: 65581972-65581972 on Build GRCh38
rs761468079 Chr.11: 65581975-65581975 on Build GRCh38
rs370393705 Chr.11: 65581997-65581997 on Build GRCh38
rs756516200 Chr.11: 65581944-65581944 on Build GRCh38
rs754919584 Chr.11: 65582001-65582002 on Build GRCh38
rs776683152 Chr.11: 65581945-65581945 on Build GRCh38
rs141669012 Chr.11: 65581921-65581921 on Build GRCh38
rs761142152 Chr.11: 65581936-65581936 on Build GRCh38
rs755742607 Chr.11: 65581973-65581974 on Build GRCh38
rs190443164 Chr.11: 65581995-65581995 on Build GRCh38
rs756115656 Chr.11: 65581982-65581982 on Build GRCh38
rs370827866 Chr.11: 65581881-65581881 on Build GRCh38
rs748564593 Chr.11: 65582005-65582005 on Build GRCh38
rs373711828 Chr.11: 65582007-65582007 on Build GRCh38
rs751255816 Chr.11: 65581826-65581826 on Build GRCh38
rs768177146 Chr.11: 65581852-65581852 on Build GRCh38

View Full Product Details