Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 152313996-152314269 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLG
Entrez Gene ID
2312
Gene Name
filaggrin
Gene Aliases
ATOD2
UniGene
Hs.654510
Gene Chromosome Location
Chr.1: 152302175-152325203 on Build GRCh38
Transcripts
NM_002016.1 NR_103778.1

Gene Symbol
FLG-AS1
Entrez Gene ID
339400
Gene Name
FLG antisense RNA 1
Gene Aliases
-
UniGene
Hs.23783
Gene Chromosome Location
Chr.1: 152313459-152366692 on Build GRCh38
Transcripts
NM_002016.1 NR_103778.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
317371 c.676A>G p.K226E Chr.1: 152314210-152314210 on Build GRCh38
5377659 c.691G>A p.G231S Chr.1: 152314195-152314195 on Build GRCh38
4684852 c.644A>G p.Y215C Chr.1: 152314242-152314242 on Build GRCh38
4554807 c.631G>A p.E211K Chr.1: 152314255-152314255 on Build GRCh38
1162060 c.694C>A p.H232N Chr.1: 152314192-152314192 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776562005 Chr.1: 152314058-152314058 on Build GRCh38
rs944553671 Chr.1: 152314128-152314128 on Build GRCh38
rs369330096 Chr.1: 152314188-152314188 on Build GRCh38
rs180813633 Chr.1: 152314162-152314162 on Build GRCh38
rs1020843420 Chr.1: 152314025-152314025 on Build GRCh38
rs199885226 Chr.1: 152314107-152314107 on Build GRCh38
rs370891418 Chr.1: 152314225-152314225 on Build GRCh38
rs755421595 Chr.1: 152314031-152314031 on Build GRCh38
rs142607263 Chr.1: 152314117-152314117 on Build GRCh38
rs775168514 Chr.1: 152314208-152314208 on Build GRCh38
rs766276354 Chr.1: 152314079-152314079 on Build GRCh38
rs761900011 Chr.1: 152314060-152314060 on Build GRCh38
rs748724298 Chr.1: 152314037-152314037 on Build GRCh38
rs762770002 Chr.1: 152314135-152314135 on Build GRCh38
rs147044321 Chr.1: 152314156-152314156 on Build GRCh38
rs145497350 Chr.1: 152314116-152314116 on Build GRCh38
rs386635460 Chr.1: 152314116-152314117 on Build GRCh38
rs758484558 Chr.1: 152314173-152314173 on Build GRCh38
rs907663577 Chr.1: 152314118-152314118 on Build GRCh38
rs770871356 Chr.1: 152314042-152314042 on Build GRCh38
rs749387670 Chr.1: 152314041-152314041 on Build GRCh38
rs751629874 Chr.1: 152314052-152314052 on Build GRCh38
rs558180193 Chr.1: 152314036-152314036 on Build GRCh38
rs767165262 Chr.1: 152314150-152314150 on Build GRCh38
rs777426932 Chr.1: 152314191-152314191 on Build GRCh38
rs773723648 Chr.1: 152314148-152314148 on Build GRCh38
rs780631118 Chr.1: 152314068-152314068 on Build GRCh38
rs749308880 Chr.1: 152314193-152314193 on Build GRCh38
rs893374458 Chr.1: 152314019-152314019 on Build GRCh38
rs747212728 Chr.1: 152314184-152314184 on Build GRCh38
rs773244478 Chr.1: 152314065-152314065 on Build GRCh38
rs377069311 Chr.1: 152314046-152314046 on Build GRCh38
rs756407428 Chr.1: 152314106-152314106 on Build GRCh38
rs756603752 Chr.1: 152314039-152314039 on Build GRCh38
rs765785917 Chr.1: 152314073-152314073 on Build GRCh38
rs372997287 Chr.1: 152314057-152314057 on Build GRCh38
rs769664239 Chr.1: 152314120-152314120 on Build GRCh38
rs756626190 Chr.1: 152314226-152314226 on Build GRCh38
rs140880252 Chr.1: 152314119-152314119 on Build GRCh38
rs755000793 Chr.1: 152314120-152314124 on Build GRCh38
rs971513367 Chr.1: 152314049-152314049 on Build GRCh38
rs577951951 Chr.1: 152314062-152314062 on Build GRCh38
rs931271894 Chr.1: 152314015-152314015 on Build GRCh38
rs759081837 Chr.1: 152314201-152314201 on Build GRCh38
rs781280495 Chr.1: 152314140-152314141 on Build GRCh38
rs368478141 Chr.1: 152314149-152314149 on Build GRCh38
rs375815215 Chr.1: 152314099-152314099 on Build GRCh38
rs1016034871 Chr.1: 152314165-152314165 on Build GRCh38
rs961814950 Chr.1: 152314140-152314140 on Build GRCh38
rs770789381 Chr.1: 152314197-152314197 on Build GRCh38
rs149599296 Chr.1: 152314212-152314212 on Build GRCh38
rs753078948 Chr.1: 152314105-152314105 on Build GRCh38
rs757787640 Chr.1: 152314017-152314017 on Build GRCh38
rs754585644 Chr.1: 152314087-152314087 on Build GRCh38
rs370142037 Chr.1: 152314047-152314047 on Build GRCh38
rs201980178 Chr.1: 152314206-152314206 on Build GRCh38
rs779105422 Chr.1: 152314163-152314163 on Build GRCh38
rs753134537 Chr.1: 152314159-152314159 on Build GRCh38
rs540577457 Chr.1: 152314055-152314055 on Build GRCh38
rs770542634 Chr.1: 152314142-152314142 on Build GRCh38
rs969698285 Chr.1: 152314125-152314125 on Build GRCh38
rs760486851 Chr.1: 152314027-152314027 on Build GRCh38
rs1010421428 Chr.1: 152314022-152314022 on Build GRCh38
rs766088521 Chr.1: 152314020-152314020 on Build GRCh38
rs780446767 Chr.1: 152314181-152314181 on Build GRCh38
rs750481871 Chr.1: 152314171-152314171 on Build GRCh38
rs754405401 Chr.1: 152314159-152314159 on Build GRCh38
rs778824842 Chr.1: 152314115-152314115 on Build GRCh38
rs752302548 Chr.1: 152314155-152314155 on Build GRCh38
rs147854910 Chr.1: 152314132-152314132 on Build GRCh38
rs774282541 Chr.1: 152314200-152314200 on Build GRCh38
rs764417267 Chr.1: 152314157-152314157 on Build GRCh38
rs757405173 Chr.1: 152314109-152314109 on Build GRCh38

View Full Product Details