Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 133468407-133468680 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EYA4
Entrez Gene ID
2070
Gene Name
EYA transcriptional coactivator and phosphatase 4
Gene Aliases
CMD1J, DFNA10
UniGene
Hs.596680
Gene Chromosome Location
Chr.6: 133240340-133532120 on Build GRCh38
Transcripts
NM_001301012.1 NM_001301013.1 NM_004100.4 NM_172103.3 NM_172105.3 XM_005266851.4 XM_005266853.4 XM_017010368.1 XM_017010369.1 XM_017010370.1 XM_017010371.1 XM_017010372.1 XM_017010373.1 XM_017010374.1 XM_017010375.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3761491 c.829G>A p.G277S Chr.6: 133468590-133468590 on Build GRCh38
1440583 c.903T>C p.D301D Chr.6: 133468664-133468664 on Build GRCh38
740040 c.811C>G p.P271A Chr.6: 133468572-133468572 on Build GRCh38
374827 c.812C>A p.P271Q Chr.6: 133468573-133468573 on Build GRCh38
5709868 c.811C>T p.P271S Chr.6: 133468572-133468572 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756672587 Chr.6: 133468652-133468652 on Build GRCh38
rs549888564 Chr.6: 133468466-133468466 on Build GRCh38
rs202030718 Chr.6: 133468599-133468599 on Build GRCh38
rs763899252 Chr.6: 133468628-133468628 on Build GRCh38
rs944091671 Chr.6: 133468501-133468501 on Build GRCh38
rs759365993 Chr.6: 133468610-133468610 on Build GRCh38
rs755455584 Chr.6: 133468648-133468648 on Build GRCh38
rs1048564742 Chr.6: 133468570-133468570 on Build GRCh38
rs750099139 Chr.6: 133468637-133468637 on Build GRCh38
rs1053782342 Chr.6: 133468661-133468661 on Build GRCh38
rs535519738 Chr.6: 133468507-133468507 on Build GRCh38
rs764985361 Chr.6: 133468636-133468636 on Build GRCh38
rs41286200 Chr.6: 133468627-133468627 on Build GRCh38
rs376151731 Chr.6: 133468609-133468609 on Build GRCh38
rs749283137 Chr.6: 133468598-133468598 on Build GRCh38
rs768010410 Chr.6: 133468649-133468649 on Build GRCh38
rs752023028 Chr.6: 133468572-133468572 on Build GRCh38
rs775461679 Chr.6: 133468622-133468622 on Build GRCh38
rs374522988 Chr.6: 133468621-133468621 on Build GRCh38
rs763676242 Chr.6: 133468515-133468515 on Build GRCh38
rs569234562 Chr.6: 133468524-133468524 on Build GRCh38
rs200092484 Chr.6: 133468656-133468656 on Build GRCh38
rs753221348 Chr.6: 133468539-133468539 on Build GRCh38
rs748654006 Chr.6: 133468585-133468585 on Build GRCh38
rs9493627 Chr.6: 133468590-133468590 on Build GRCh38
rs757252384 Chr.6: 133468660-133468660 on Build GRCh38
rs1035314735 Chr.6: 133468632-133468632 on Build GRCh38
rs570118350 Chr.6: 133468476-133468476 on Build GRCh38
rs555728674 Chr.6: 133468584-133468584 on Build GRCh38
rs950691061 Chr.6: 133468545-133468545 on Build GRCh38
rs558399007 Chr.6: 133468603-133468603 on Build GRCh38
rs1035683390 Chr.6: 133468508-133468508 on Build GRCh38
rs375862613 Chr.6: 133468554-133468554 on Build GRCh38
rs778381205 Chr.6: 133468556-133468556 on Build GRCh38
rs778089186 Chr.6: 133468654-133468654 on Build GRCh38
rs188638017 Chr.6: 133468448-133468448 on Build GRCh38
rs761206775 Chr.6: 133468634-133468634 on Build GRCh38
rs370562413 Chr.6: 133468574-133468574 on Build GRCh38

View Full Product Details