Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 110155242-110155489 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL4A1
Entrez Gene ID
1282
Gene Name
collagen type IV alpha 1 chain
Gene Aliases
BSVD, RATOR
UniGene
Hs.17441
Gene Chromosome Location
Chr.13: 110148958-110307157 on Build GRCh38
Transcripts
NM_001845.5 XM_011521048.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3467099 c.4755+1G>A p.? Chr.13: 110155282-110155282 on Build GRCh38
6633083 c.4727C>T p.S1576L Chr.13: 110155311-110155311 on Build GRCh38
4760091 c.4755+2T>C p.? Chr.13: 110155281-110155281 on Build GRCh38
4496215 c.4706C>T p.P1569L Chr.13: 110155332-110155332 on Build GRCh38
6633086 c.4704A>T p.P1568P Chr.13: 110155334-110155334 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374343679 Chr.13: 110155361-110155361 on Build GRCh38
rs75885959 Chr.13: 110155332-110155332 on Build GRCh38
rs747637623 Chr.13: 110155365-110155365 on Build GRCh38
rs895949915 Chr.13: 110155264-110155264 on Build GRCh38
rs375191949 Chr.13: 110155430-110155430 on Build GRCh38
rs760683683 Chr.13: 110155417-110155417 on Build GRCh38
rs141072338 Chr.13: 110155379-110155379 on Build GRCh38
rs750924809 Chr.13: 110155439-110155439 on Build GRCh38
rs774341334 Chr.13: 110155320-110155320 on Build GRCh38
rs927275932 Chr.13: 110155438-110155438 on Build GRCh38
rs372131402 Chr.13: 110155391-110155391 on Build GRCh38
rs752187609 Chr.13: 110155446-110155446 on Build GRCh38
rs775544336 Chr.13: 110155326-110155326 on Build GRCh38
rs764244883 Chr.13: 110155331-110155331 on Build GRCh38
rs768345784 Chr.13: 110155277-110155277 on Build GRCh38
rs754511803 Chr.13: 110155360-110155360 on Build GRCh38
rs201150281 Chr.13: 110155380-110155380 on Build GRCh38
rs146822264 Chr.13: 110155298-110155298 on Build GRCh38
rs890307300 Chr.13: 110155464-110155464 on Build GRCh38
rs766813036 Chr.13: 110155445-110155445 on Build GRCh38
rs770790869 Chr.13: 110155314-110155314 on Build GRCh38
rs188147295 Chr.13: 110155460-110155460 on Build GRCh38
rs1025847262 Chr.13: 110155274-110155274 on Build GRCh38
rs980701216 Chr.13: 110155433-110155433 on Build GRCh38
rs757699048 Chr.13: 110155337-110155337 on Build GRCh38
rs775590711 Chr.13: 110155409-110155409 on Build GRCh38
rs760910929 Chr.13: 110155327-110155327 on Build GRCh38
rs552846696 Chr.13: 110155310-110155310 on Build GRCh38
rs113994114 Chr.13: 110155300-110155300 on Build GRCh38
rs1053853583 Chr.13: 110155442-110155442 on Build GRCh38
rs200290773 Chr.13: 110155322-110155322 on Build GRCh38
rs776274526 Chr.13: 110155455-110155455 on Build GRCh38
rs370674095 Chr.13: 110155301-110155301 on Build GRCh38
rs1051434182 Chr.13: 110155456-110155456 on Build GRCh38
rs374495995 Chr.13: 110155432-110155432 on Build GRCh38
rs570472326 Chr.13: 110155280-110155280 on Build GRCh38
rs866262017 Chr.13: 110155347-110155347 on Build GRCh38
rs617111 Chr.13: 110155429-110155429 on Build GRCh38
rs183728092 Chr.13: 110155448-110155448 on Build GRCh38
rs759600848 Chr.13: 110155321-110155321 on Build GRCh38
rs764309688 Chr.13: 110155425-110155425 on Build GRCh38
rs746450625 Chr.13: 110155276-110155276 on Build GRCh38
rs749107113 Chr.13: 110155382-110155382 on Build GRCh38
rs773014501 Chr.13: 110155307-110155307 on Build GRCh38
rs758928777 Chr.13: 110155440-110155440 on Build GRCh38
rs191620431 Chr.13: 110155265-110155265 on Build GRCh38
rs555466778 Chr.13: 110155359-110155359 on Build GRCh38
rs369837146 Chr.13: 110155449-110155449 on Build GRCh38
rs779727078 Chr.13: 110155408-110155410 on Build GRCh38
rs765833500 Chr.13: 110155355-110155355 on Build GRCh38
rs75795731 Chr.13: 110155288-110155288 on Build GRCh38
rs774251351 Chr.13: 110155390-110155390 on Build GRCh38

View Full Product Details