Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 96264185-96264432 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BDKRB1
Entrez Gene ID
623
Gene Name
bradykinin receptor B1
Gene Aliases
B1BKR, B1R, BDKRB2, BKB1R, BKR1, BRADYB1
UniGene
Hs.525572
Gene Chromosome Location
Chr.14: 96256210-96264763 on Build GRCh38
Transcripts
NM_000710.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3815623 c.617T>C p.L206S Chr.14: 96264299-96264299 on Build GRCh38
6611146 c.582C>A p.P194P Chr.14: 96264264-96264264 on Build GRCh38
1640184 c.721G>T p.G241W Chr.14: 96264403-96264403 on Build GRCh38
69814 c.749C>T p.A250V Chr.14: 96264431-96264431 on Build GRCh38
5589807 c.636C>T p.L212L Chr.14: 96264318-96264318 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs952768042 Chr.14: 96264301-96264301 on Build GRCh38
rs761782388 Chr.14: 96264323-96264323 on Build GRCh38
rs753420449 Chr.14: 96264336-96264336 on Build GRCh38
rs375483074 Chr.14: 96264244-96264244 on Build GRCh38
rs753395833 Chr.14: 96264252-96264252 on Build GRCh38
rs768607326 Chr.14: 96264297-96264297 on Build GRCh38
rs748553489 Chr.14: 96264265-96264265 on Build GRCh38
rs868223942 Chr.14: 96264377-96264378 on Build GRCh38
rs774234943 Chr.14: 96264315-96264315 on Build GRCh38
rs773048836 Chr.14: 96264332-96264332 on Build GRCh38
rs755771607 Chr.14: 96264248-96264248 on Build GRCh38
rs371690606 Chr.14: 96264376-96264376 on Build GRCh38
rs758922069 Chr.14: 96264365-96264365 on Build GRCh38
rs779487244 Chr.14: 96264204-96264204 on Build GRCh38
rs772887692 Chr.14: 96264399-96264399 on Build GRCh38
rs779513626 Chr.14: 96264264-96264264 on Build GRCh38
rs112987981 Chr.14: 96264294-96264294 on Build GRCh38
rs113666900 Chr.14: 96264409-96264409 on Build GRCh38
rs145002930 Chr.14: 96264209-96264209 on Build GRCh38
rs747328339 Chr.14: 96264371-96264371 on Build GRCh38
rs147649798 Chr.14: 96264214-96264214 on Build GRCh38
rs202216190 Chr.14: 96264390-96264390 on Build GRCh38
rs61744359 Chr.14: 96264329-96264329 on Build GRCh38
rs905716111 Chr.14: 96264211-96264211 on Build GRCh38
rs140574024 Chr.14: 96264222-96264222 on Build GRCh38
rs188387870 Chr.14: 96264243-96264243 on Build GRCh38
rs200342765 Chr.14: 96264208-96264208 on Build GRCh38
rs143735378 Chr.14: 96264402-96264402 on Build GRCh38
rs757505732 Chr.14: 96264337-96264337 on Build GRCh38
rs369351922 Chr.14: 96264375-96264375 on Build GRCh38
rs368279365 Chr.14: 96264272-96264272 on Build GRCh38
rs765990196 Chr.14: 96264334-96264334 on Build GRCh38
rs979445300 Chr.14: 96264309-96264309 on Build GRCh38
rs771176101 Chr.14: 96264277-96264277 on Build GRCh38
rs753566189 Chr.14: 96264338-96264340 on Build GRCh38
rs1045664666 Chr.14: 96264362-96264362 on Build GRCh38
rs757437458 Chr.14: 96264374-96264374 on Build GRCh38
rs566659692 Chr.14: 96264266-96264266 on Build GRCh38
rs45528332 Chr.14: 96264403-96264403 on Build GRCh38
rs868023682 Chr.14: 96264279-96264279 on Build GRCh38
rs557512070 Chr.14: 96264386-96264386 on Build GRCh38
rs560358743 Chr.14: 96264333-96264333 on Build GRCh38
rs372328639 Chr.14: 96264217-96264217 on Build GRCh38
rs764894215 Chr.14: 96264406-96264406 on Build GRCh38
rs765863931 Chr.14: 96264250-96264250 on Build GRCh38
rs764807889 Chr.14: 96264347-96264347 on Build GRCh38
rs917950992 Chr.14: 96264342-96264342 on Build GRCh38
rs573449408 Chr.14: 96264410-96264410 on Build GRCh38
rs763956811 Chr.14: 96264224-96264224 on Build GRCh38
rs751373446 Chr.14: 96264232-96264232 on Build GRCh38
rs762941065 Chr.14: 96264223-96264223 on Build GRCh38
rs143823168 Chr.14: 96264253-96264253 on Build GRCh38
rs146837856 Chr.14: 96264355-96264355 on Build GRCh38
rs767266886 Chr.14: 96264328-96264328 on Build GRCh38
rs372538907 Chr.14: 96264340-96264340 on Build GRCh38
rs76887699 Chr.14: 96264241-96264241 on Build GRCh38
rs752244721 Chr.14: 96264407-96264407 on Build GRCh38
rs33925361 Chr.14: 96264381-96264381 on Build GRCh38
rs748095029 Chr.14: 96264385-96264385 on Build GRCh38
rs376804785 Chr.14: 96264370-96264370 on Build GRCh38
rs200369442 Chr.14: 96264382-96264382 on Build GRCh38

View Full Product Details