Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 91164506-91164779 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCN
Entrez Gene ID
1634
Gene Name
decorin
Gene Aliases
CSCD, DSPG2, PG40, PGII, PGS2, SLRR1B
UniGene
Hs.156316
Gene Chromosome Location
Chr.12: 91143277-91183124 on Build GRCh38
Transcripts
NM_001920.4 NM_133503.3 NM_133504.3 NM_133505.3 NM_133506.3 NM_133507.3 XM_005268693.1 XM_006719270.1 XM_017018917.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
342149 c.324+1G>T p.? Chr.12: 91164604-91164604 on Build GRCh38
4937522 c.311T>C p.L104P Chr.12: 91164618-91164618 on Build GRCh38
3466385 c.296G>A p.G99E Chr.12: 91164633-91164633 on Build GRCh38
944392 c.283G>A p.E95K Chr.12: 91164646-91164646 on Build GRCh38
6640956 c.272A>T p.N91I Chr.12: 91164657-91164657 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780406074 Chr.12: 91164727-91164727 on Build GRCh38
rs541064496 Chr.12: 91164587-91164587 on Build GRCh38
rs762795489 Chr.12: 91164708-91164708 on Build GRCh38
rs113132993 Chr.12: 91164719-91164719 on Build GRCh38
rs1043985896 Chr.12: 91164537-91164537 on Build GRCh38
rs555086929 Chr.12: 91164605-91164605 on Build GRCh38
rs767153102 Chr.12: 91164641-91164641 on Build GRCh38
rs747245434 Chr.12: 91164733-91164733 on Build GRCh38
rs146575397 Chr.12: 91164583-91164583 on Build GRCh38
rs993484273 Chr.12: 91164618-91164618 on Build GRCh38
rs752179884 Chr.12: 91164568-91164568 on Build GRCh38
rs770718112 Chr.12: 91164633-91164633 on Build GRCh38
rs778239874 Chr.12: 91164576-91164576 on Build GRCh38
rs146745555 Chr.12: 91164619-91164619 on Build GRCh38
rs766965021 Chr.12: 91164561-91164561 on Build GRCh38
rs112967859 Chr.12: 91164602-91164602 on Build GRCh38
rs186303865 Chr.12: 91164560-91164560 on Build GRCh38
rs765826074 Chr.12: 91164693-91164693 on Build GRCh38
rs199610197 Chr.12: 91164604-91164604 on Build GRCh38
rs530149040 Chr.12: 91164737-91164737 on Build GRCh38
rs376699697 Chr.12: 91164652-91164652 on Build GRCh38
rs754315861 Chr.12: 91164581-91164581 on Build GRCh38
rs113560440 Chr.12: 91164662-91164662 on Build GRCh38
rs780467336 Chr.12: 91164591-91164591 on Build GRCh38
rs757730297 Chr.12: 91164682-91164682 on Build GRCh38
rs758926209 Chr.12: 91164585-91164586 on Build GRCh38
rs3138221 Chr.12: 91164647-91164647 on Build GRCh38
rs747443101 Chr.12: 91164636-91164636 on Build GRCh38
rs750727808 Chr.12: 91164701-91164701 on Build GRCh38
rs763519278 Chr.12: 91164668-91164668 on Build GRCh38
rs201346668 Chr.12: 91164637-91164637 on Build GRCh38
rs772404315 Chr.12: 91164590-91164590 on Build GRCh38
rs754333573 Chr.12: 91164670-91164670 on Build GRCh38
rs182985515 Chr.12: 91164541-91164541 on Build GRCh38
rs575105909 Chr.12: 91164586-91164586 on Build GRCh38
rs11106024 Chr.12: 91164676-91164676 on Build GRCh38
rs530254085 Chr.12: 91164548-91164548 on Build GRCh38
rs759374285 Chr.12: 91164638-91164638 on Build GRCh38
rs749184678 Chr.12: 91164623-91164623 on Build GRCh38
rs781160196 Chr.12: 91164739-91164739 on Build GRCh38
rs191218249 Chr.12: 91164723-91164723 on Build GRCh38
rs755689229 Chr.12: 91164570-91164570 on Build GRCh38

View Full Product Details