Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 63295035-63295304 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL20A1
Entrez Gene ID
57642
Gene Name
collagen type XX alpha 1 chain
Gene Aliases
bA261N11.4
UniGene
Hs.271285
Gene Chromosome Location
Chr.20: 63293169-63334847 on Build GRCh38
Transcripts
NM_020882.2 XM_011528937.1 XM_011528938.1 XM_011528939.1 XM_011528940.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3939434 c.55G>A p.A19T Chr.20: 63295162-63295162 on Build GRCh38
5142620 c.54C>T p.G18G Chr.20: 63295161-63295161 on Build GRCh38
3770878 c.61C>T p.L21L Chr.20: 63295168-63295168 on Build GRCh38
4487533 c.31C>T p.L11F Chr.20: 63295138-63295138 on Build GRCh38
4487532 c.31C>T p.L11F Chr.20: 63295138-63295138 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370140638 Chr.20: 63295127-63295127 on Build GRCh38
rs781283646 Chr.20: 63295116-63295116 on Build GRCh38
rs1015270984 Chr.20: 63295086-63295086 on Build GRCh38
rs753809030 Chr.20: 63295063-63295063 on Build GRCh38
rs573268710 Chr.20: 63295061-63295061 on Build GRCh38
rs750316242 Chr.20: 63295206-63295206 on Build GRCh38
rs369025538 Chr.20: 63295134-63295134 on Build GRCh38
rs768092946 Chr.20: 63295171-63295171 on Build GRCh38
rs765164190 Chr.20: 63295198-63295198 on Build GRCh38
rs921951250 Chr.20: 63295231-63295231 on Build GRCh38
rs566583799 Chr.20: 63295254-63295254 on Build GRCh38
rs758976717 Chr.20: 63295155-63295155 on Build GRCh38
rs771591733 Chr.20: 63295146-63295146 on Build GRCh38
rs886593089 Chr.20: 63295067-63295067 on Build GRCh38
rs1027764080 Chr.20: 63295266-63295267 on Build GRCh38
rs773681956 Chr.20: 63295135-63295135 on Build GRCh38
rs373759197 Chr.20: 63295065-63295065 on Build GRCh38
rs183105977 Chr.20: 63295100-63295100 on Build GRCh38
rs781328211 Chr.20: 63295224-63295224 on Build GRCh38
rs879186642 Chr.20: 63295250-63295250 on Build GRCh38
rs551578950 Chr.20: 63295253-63295253 on Build GRCh38
rs372680831 Chr.20: 63295057-63295057 on Build GRCh38
rs376987277 Chr.20: 63295264-63295264 on Build GRCh38
rs376830125 Chr.20: 63295129-63295129 on Build GRCh38
rs372419078 Chr.20: 63295180-63295180 on Build GRCh38
rs975181953 Chr.20: 63295219-63295219 on Build GRCh38
rs778773127 Chr.20: 63295077-63295077 on Build GRCh38
rs923628211 Chr.20: 63295275-63295275 on Build GRCh38
rs767037555 Chr.20: 63295158-63295158 on Build GRCh38
rs984643516 Chr.20: 63295058-63295058 on Build GRCh38
rs527370752 Chr.20: 63295255-63295255 on Build GRCh38
rs562736407 Chr.20: 63295151-63295151 on Build GRCh38
rs763922288 Chr.20: 63295162-63295162 on Build GRCh38
rs898182205 Chr.20: 63295087-63295087 on Build GRCh38
rs1036746547 Chr.20: 63295269-63295269 on Build GRCh38
rs1016120608 Chr.20: 63295245-63295245 on Build GRCh38
rs753653751 Chr.20: 63295174-63295174 on Build GRCh38
rs1026285402 Chr.20: 63295216-63295216 on Build GRCh38
rs544624291 Chr.20: 63295141-63295141 on Build GRCh38
rs755917680 Chr.20: 63295058-63295058 on Build GRCh38
rs112489058 Chr.20: 63295232-63295232 on Build GRCh38
rs755072816 Chr.20: 63295212-63295212 on Build GRCh38
rs959499102 Chr.20: 63295247-63295247 on Build GRCh38
rs775067603 Chr.20: 63295161-63295161 on Build GRCh38
rs747089798 Chr.20: 63295095-63295095 on Build GRCh38
rs977814826 Chr.20: 63295274-63295274 on Build GRCh38
rs1003950123 Chr.20: 63295084-63295084 on Build GRCh38
rs540408642 Chr.20: 63295075-63295075 on Build GRCh38
rs991004388 Chr.20: 63295082-63295082 on Build GRCh38
rs908061694 Chr.20: 63295252-63295252 on Build GRCh38
rs898218155 Chr.20: 63295281-63295281 on Build GRCh38
rs868604295 Chr.20: 63295117-63295117 on Build GRCh38

View Full Product Details