Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 70894055-70894328 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
B3GAT2
Entrez Gene ID
135152
Gene Name
beta-1,3-glucuronyltransferase 2
Gene Aliases
GLCATS
UniGene
Hs.713609
Gene Chromosome Location
Chr.6: 70861366-70957085 on Build GRCh38
Transcripts
NM_080742.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4386526 c.674G>A p.G225D Chr.6: 70894190-70894190 on Build GRCh38
6609281 c.604C>T p.R202C Chr.6: 70894260-70894260 on Build GRCh38
6609282 c.651C>T p.Y217Y Chr.6: 70894213-70894213 on Build GRCh38
1176528 c.659C>T p.P220L Chr.6: 70894205-70894205 on Build GRCh38
403829 c.605G>A p.R202H Chr.6: 70894259-70894259 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766615751 Chr.6: 70894150-70894150 on Build GRCh38
rs559747245 Chr.6: 70894261-70894261 on Build GRCh38
rs756691800 Chr.6: 70894110-70894110 on Build GRCh38
rs759621574 Chr.6: 70894268-70894268 on Build GRCh38
rs746136093 Chr.6: 70894161-70894161 on Build GRCh38
rs769719606 Chr.6: 70894170-70894170 on Build GRCh38
rs753086277 Chr.6: 70894105-70894105 on Build GRCh38
rs757465499 Chr.6: 70894156-70894156 on Build GRCh38
rs776621778 Chr.6: 70894260-70894260 on Build GRCh38
rs746022699 Chr.6: 70894212-70894212 on Build GRCh38
rs773915976 Chr.6: 70894248-70894248 on Build GRCh38
rs375873781 Chr.6: 70894198-70894198 on Build GRCh38
rs757375740 Chr.6: 70894208-70894208 on Build GRCh38
rs576071209 Chr.6: 70894092-70894092 on Build GRCh38
rs750513549 Chr.6: 70894269-70894269 on Build GRCh38
rs770367885 Chr.6: 70894239-70894239 on Build GRCh38
rs772064174 Chr.6: 70894190-70894190 on Build GRCh38
rs754440723 Chr.6: 70894207-70894207 on Build GRCh38
rs780300853 Chr.6: 70894282-70894282 on Build GRCh38
rs943853919 Chr.6: 70894243-70894243 on Build GRCh38
rs987713823 Chr.6: 70894130-70894130 on Build GRCh38
rs775050972 Chr.6: 70894258-70894258 on Build GRCh38
rs760977667 Chr.6: 70894082-70894082 on Build GRCh38
rs764672977 Chr.6: 70894205-70894205 on Build GRCh38
rs751263724 Chr.6: 70894088-70894088 on Build GRCh38
rs765905195 Chr.6: 70894188-70894188 on Build GRCh38
rs934615176 Chr.6: 70894280-70894280 on Build GRCh38
rs774249812 Chr.6: 70894167-70894167 on Build GRCh38
rs543509960 Chr.6: 70894115-70894115 on Build GRCh38
rs779250933 Chr.6: 70894157-70894157 on Build GRCh38
rs767180832 Chr.6: 70894257-70894257 on Build GRCh38
rs371527631 Chr.6: 70894192-70894192 on Build GRCh38
rs779074725 Chr.6: 70894209-70894209 on Build GRCh38
rs773092539 Chr.6: 70894231-70894231 on Build GRCh38
rs533685012 Chr.6: 70894267-70894267 on Build GRCh38
rs367693161 Chr.6: 70894179-70894179 on Build GRCh38
rs552142097 Chr.6: 70894283-70894283 on Build GRCh38
rs138042804 Chr.6: 70894234-70894234 on Build GRCh38
rs749246454 Chr.6: 70894235-70894235 on Build GRCh38
rs541631037 Chr.6: 70894217-70894217 on Build GRCh38
rs890567324 Chr.6: 70894233-70894233 on Build GRCh38
rs762418161 Chr.6: 70894180-70894180 on Build GRCh38
rs773063307 Chr.6: 70894274-70894275 on Build GRCh38
rs780210019 Chr.6: 70894213-70894213 on Build GRCh38
rs561877361 Chr.6: 70894138-70894138 on Build GRCh38
rs368880277 Chr.6: 70894272-70894272 on Build GRCh38
rs538444071 Chr.6: 70894226-70894226 on Build GRCh38
rs867463178 Chr.6: 70894250-70894250 on Build GRCh38
rs759031870 Chr.6: 70894249-70894249 on Build GRCh38
rs773823914 Chr.6: 70894191-70894191 on Build GRCh38
rs201425180 Chr.6: 70894204-70894204 on Build GRCh38
rs940986185 Chr.6: 70894109-70894109 on Build GRCh38
rs778486662 Chr.6: 70894126-70894126 on Build GRCh38
rs374592671 Chr.6: 70894168-70894168 on Build GRCh38
rs748064015 Chr.6: 70894182-70894182 on Build GRCh38
rs867150619 Chr.6: 70894160-70894160 on Build GRCh38

View Full Product Details