Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 77164075-77164344 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCNG2
Entrez Gene ID
901
Gene Name
cyclin G2
Gene Aliases
-
UniGene
Hs.740456
Gene Chromosome Location
Chr.4: 77157204-77170060 on Build GRCh38
Transcripts
NM_004354.2 XM_011532398.1 XM_011532399.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4005804 c.712G>T p.D238Y Chr.4: 77164280-77164280 on Build GRCh38
6623030 c.717T>C p.T239T Chr.4: 77164285-77164285 on Build GRCh38
6623033 c.770C>A p.S257Y Chr.4: 77164338-77164338 on Build GRCh38
4343140 c.723C>A p.F241L Chr.4: 77164291-77164291 on Build GRCh38
4343142 c.738G>T p.E246D Chr.4: 77164306-77164306 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs899493230 Chr.4: 77164160-77164160 on Build GRCh38
rs758895798 Chr.4: 77164284-77164284 on Build GRCh38
rs1003981055 Chr.4: 77164227-77164227 on Build GRCh38
rs766489205 Chr.4: 77164265-77164265 on Build GRCh38
rs890836622 Chr.4: 77164196-77164196 on Build GRCh38
rs768629171 Chr.4: 77164255-77164255 on Build GRCh38
rs757770502 Chr.4: 77164262-77164262 on Build GRCh38
rs574484644 Chr.4: 77164261-77164262 on Build GRCh38
rs770537416 Chr.4: 77164239-77164239 on Build GRCh38
rs946605765 Chr.4: 77164154-77164155 on Build GRCh38
rs757589588 Chr.4: 77164296-77164296 on Build GRCh38
rs376445357 Chr.4: 77164245-77164245 on Build GRCh38
rs533572928 Chr.4: 77164241-77164241 on Build GRCh38
rs772560078 Chr.4: 77164156-77164156 on Build GRCh38
rs147604582 Chr.4: 77164294-77164294 on Build GRCh38
rs753514383 Chr.4: 77164261-77164261 on Build GRCh38
rs956670911 Chr.4: 77164109-77164109 on Build GRCh38
rs927237718 Chr.4: 77164145-77164145 on Build GRCh38
rs372283071 Chr.4: 77164280-77164280 on Build GRCh38
rs1034996746 Chr.4: 77164310-77164310 on Build GRCh38
rs764480461 Chr.4: 77164291-77164291 on Build GRCh38
rs780735377 Chr.4: 77164312-77164312 on Build GRCh38
rs992215253 Chr.4: 77164110-77164110 on Build GRCh38
rs753219931 Chr.4: 77164281-77164281 on Build GRCh38
rs988351013 Chr.4: 77164128-77164131 on Build GRCh38
rs932345882 Chr.4: 77164173-77164173 on Build GRCh38
rs139497489 Chr.4: 77164274-77164274 on Build GRCh38
rs980703105 Chr.4: 77164289-77164289 on Build GRCh38
rs4150085 Chr.4: 77164236-77164236 on Build GRCh38
rs749177845 Chr.4: 77164251-77164251 on Build GRCh38
rs763716061 Chr.4: 77164119-77164119 on Build GRCh38
rs116948162 Chr.4: 77164189-77164189 on Build GRCh38
rs759569928 Chr.4: 77164278-77164278 on Build GRCh38
rs971228139 Chr.4: 77164120-77164120 on Build GRCh38
rs758770961 Chr.4: 77164228-77164228 on Build GRCh38
rs775293041 Chr.4: 77164249-77164249 on Build GRCh38
rs548947776 Chr.4: 77164168-77164168 on Build GRCh38
rs979947310 Chr.4: 77164127-77164127 on Build GRCh38
rs761019183 Chr.4: 77164263-77164263 on Build GRCh38
rs774285178 Chr.4: 77164258-77164258 on Build GRCh38
rs769307109 Chr.4: 77164246-77164246 on Build GRCh38

View Full Product Details