Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 132075668-132075935 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSF2
Entrez Gene ID
1437
Gene Name
colony stimulating factor 2
Gene Aliases
GMCSF
UniGene
Hs.1349
Gene Chromosome Location
Chr.5: 132073792-132076170 on Build GRCh38
Transcripts
NM_000758.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6636038 c.332C>T p.T111I Chr.5: 132075749-132075749 on Build GRCh38
4383985 c.388T>C p.F130L Chr.5: 132075805-132075805 on Build GRCh38
3760908 c.350T>C p.I117T Chr.5: 132075767-132075767 on Build GRCh38
7362792 c.416G>C p.W139S Chr.5: 132075833-132075833 on Build GRCh38
3918612 c.365G>A p.S122N Chr.5: 132075782-132075782 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374836396 Chr.5: 132075857-132075857 on Build GRCh38
rs2069640 Chr.5: 132075761-132075761 on Build GRCh38
rs780237469 Chr.5: 132075894-132075894 on Build GRCh38
rs748213403 Chr.5: 132075792-132075792 on Build GRCh38
rs751251297 Chr.5: 132075872-132075872 on Build GRCh38
rs371799279 Chr.5: 132075739-132075739 on Build GRCh38
rs779033001 Chr.5: 132075781-132075781 on Build GRCh38
rs25882 Chr.5: 132075767-132075767 on Build GRCh38
rs923174222 Chr.5: 132075888-132075888 on Build GRCh38
rs778087300 Chr.5: 132075825-132075825 on Build GRCh38
rs755412035 Chr.5: 132075773-132075773 on Build GRCh38
rs752247728 Chr.5: 132075727-132075727 on Build GRCh38
rs756143117 Chr.5: 132075734-132075734 on Build GRCh38
rs759609965 Chr.5: 132075851-132075851 on Build GRCh38
rs776500407 Chr.5: 132075849-132075849 on Build GRCh38
rs373703815 Chr.5: 132075695-132075695 on Build GRCh38
rs759487007 Chr.5: 132075701-132075701 on Build GRCh38
rs577891805 Chr.5: 132075734-132075734 on Build GRCh38
rs750789448 Chr.5: 132075746-132075746 on Build GRCh38
rs770771439 Chr.5: 132075845-132075845 on Build GRCh38
rs772232572 Chr.5: 132075807-132075807 on Build GRCh38
rs1028953004 Chr.5: 132075703-132075703 on Build GRCh38
rs760496931 Chr.5: 132075694-132075694 on Build GRCh38
rs781248668 Chr.5: 132075720-132075721 on Build GRCh38
rs763776262 Chr.5: 132075741-132075741 on Build GRCh38
rs564912678 Chr.5: 132075843-132075843 on Build GRCh38
rs187262170 Chr.5: 132075880-132075880 on Build GRCh38
rs2069630 Chr.5: 132075879-132075879 on Build GRCh38
rs952750895 Chr.5: 132075704-132075704 on Build GRCh38
rs554218352 Chr.5: 132075692-132075692 on Build GRCh38
rs769838857 Chr.5: 132075853-132075853 on Build GRCh38
rs747150863 Chr.5: 132075835-132075835 on Build GRCh38
rs758526563 Chr.5: 132075903-132075903 on Build GRCh38
rs373535761 Chr.5: 132075891-132075891 on Build GRCh38
rs940112572 Chr.5: 132075789-132075789 on Build GRCh38
rs754359508 Chr.5: 132075768-132075768 on Build GRCh38
rs764842866 Chr.5: 132075712-132075712 on Build GRCh38
rs949177975 Chr.5: 132075841-132075841 on Build GRCh38
rs776731418 Chr.5: 132075697-132075697 on Build GRCh38
rs775184590 Chr.5: 132075856-132075856 on Build GRCh38
rs752761208 Chr.5: 132075896-132075896 on Build GRCh38
rs745838061 Chr.5: 132075733-132075734 on Build GRCh38

View Full Product Details