Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 127807611-127807846 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FPGS
Entrez Gene ID
2356
Gene Name
folylpolyglutamate synthase
Gene Aliases
-
UniGene
Hs.335084
Gene Chromosome Location
Chr.9: 127802858-127814520 on Build GRCh38
Transcripts
NM_001018078.2 NM_001288803.1 NM_004957.5 NR_110170.1 XM_005251864.3 XM_011518439.2 XM_017014565.1 XM_017014566.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3904305 c.679G>A p.D227N Chr.9: 127807623-127807623 on Build GRCh38
3654514 c.670C>T p.L224F Chr.9: 127807614-127807614 on Build GRCh38
4921878 c.701A>G p.D234G Chr.9: 127807645-127807645 on Build GRCh38
4940839 c.698G>T p.G233V Chr.9: 127807642-127807642 on Build GRCh38
752667 c.700G>A p.D234N Chr.9: 127807644-127807644 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761029201 Chr.9: 127807739-127807739 on Build GRCh38
rs377414586 Chr.9: 127807708-127807708 on Build GRCh38
rs749039023 Chr.9: 127807704-127807704 on Build GRCh38
rs754234653 Chr.9: 127807648-127807648 on Build GRCh38
rs979498555 Chr.9: 127807634-127807634 on Build GRCh38
rs138057415 Chr.9: 127807650-127807650 on Build GRCh38
rs773329644 Chr.9: 127807735-127807735 on Build GRCh38
rs752640884 Chr.9: 127807645-127807645 on Build GRCh38
rs138442786 Chr.9: 127807799-127807799 on Build GRCh38
rs749305433 Chr.9: 127807820-127807820 on Build GRCh38
rs756655583 Chr.9: 127807673-127807673 on Build GRCh38
rs112925235 Chr.9: 127807763-127807763 on Build GRCh38
rs768817305 Chr.9: 127807824-127807824 on Build GRCh38
rs529852737 Chr.9: 127807720-127807720 on Build GRCh38
rs141628617 Chr.9: 127807806-127807806 on Build GRCh38
rs1017299473 Chr.9: 127807780-127807780 on Build GRCh38
rs184155766 Chr.9: 127807813-127807813 on Build GRCh38
rs772113700 Chr.9: 127807721-127807721 on Build GRCh38
rs560807113 Chr.9: 127807693-127807693 on Build GRCh38
rs751093805 Chr.9: 127807662-127807662 on Build GRCh38
rs375974912 Chr.9: 127807676-127807676 on Build GRCh38
rs967159607 Chr.9: 127807741-127807741 on Build GRCh38
rs779861417 Chr.9: 127807701-127807701 on Build GRCh38
rs758390457 Chr.9: 127807673-127807674 on Build GRCh38
rs745577142 Chr.9: 127807685-127807685 on Build GRCh38
rs572353784 Chr.9: 127807649-127807649 on Build GRCh38
rs540968727 Chr.9: 127807661-127807661 on Build GRCh38
rs766678221 Chr.9: 127807740-127807740 on Build GRCh38
rs542408761 Chr.9: 127807812-127807812 on Build GRCh38
rs777753909 Chr.9: 127807676-127807677 on Build GRCh38

View Full Product Details