Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 45136854-45137115 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FANCM
Entrez Gene ID
57697
Gene Name
Fanconi anemia complementation group M
Gene Aliases
FAAP250, KIAA1596
UniGene
Hs.509229
Gene Chromosome Location
Chr.14: 45135939-45200890 on Build GRCh38
Transcripts
NM_001308133.1 NM_001308134.1 NM_020937.3 XM_011537034.2 XM_011537035.2 XM_017021523.1 XM_017021524.1 XM_017021525.1 XM_017021527.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1188809 c.523T>A p.S175T Chr.14: 45137083-45137083 on Build GRCh38
7234345 c.524C>T p.S175F Chr.14: 45137084-45137084 on Build GRCh38
5470239 c.543G>A p.W181* Chr.14: 45137103-45137103 on Build GRCh38
5019675 c.527C>T p.T176I Chr.14: 45137087-45137087 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs772770014 Chr.14: 45137006-45137006 on Build GRCh38
rs1050572153 Chr.14: 45136906-45136906 on Build GRCh38
rs1010690835 Chr.14: 45137033-45137033 on Build GRCh38
rs745424852 Chr.14: 45137075-45137075 on Build GRCh38
rs763793953 Chr.14: 45137027-45137027 on Build GRCh38
rs1041347930 Chr.14: 45136963-45136963 on Build GRCh38
rs77374493 Chr.14: 45137087-45137087 on Build GRCh38
rs753514728 Chr.14: 45137044-45137044 on Build GRCh38
rs573519429 Chr.14: 45136921-45136921 on Build GRCh38
rs1008383699 Chr.14: 45136930-45136930 on Build GRCh38
rs10138997 Chr.14: 45137084-45137084 on Build GRCh38
rs756896619 Chr.14: 45137052-45137052 on Build GRCh38
rs756581728 Chr.14: 45137023-45137023 on Build GRCh38
rs889978741 Chr.14: 45136915-45136915 on Build GRCh38
rs758296940 Chr.14: 45137080-45137080 on Build GRCh38
rs931760543 Chr.14: 45136901-45136901 on Build GRCh38
rs898787734 Chr.14: 45136895-45136895 on Build GRCh38
rs753046957 Chr.14: 45137022-45137022 on Build GRCh38
rs560644773 Chr.14: 45137059-45137059 on Build GRCh38
rs1021627007 Chr.14: 45137079-45137079 on Build GRCh38
rs767925823 Chr.14: 45137020-45137020 on Build GRCh38
rs979529277 Chr.14: 45137069-45137069 on Build GRCh38
rs779858649 Chr.14: 45137083-45137083 on Build GRCh38

View Full Product Details