Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 100020559-100020820 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERP44
Entrez Gene ID
23071
Gene Name
endoplasmic reticulum protein 44
Gene Aliases
PDIA10, TXNDC4
UniGene
Hs.154023
Gene Chromosome Location
Chr.9: 99979179-100099052 on Build GRCh38
Transcripts
NM_015051.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3166862 c.539G>A p.R180Q Chr.9: 100020664-100020664 on Build GRCh38
6417421 c.473G>T p.R158L Chr.9: 100020730-100020730 on Build GRCh38
6479586 c.502G>C p.E168Q Chr.9: 100020701-100020701 on Build GRCh38
461385 c.494G>C p.G165A Chr.9: 100020709-100020709 on Build GRCh38
6656270 c.582A>G p.A194A Chr.9: 100020621-100020621 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756976665 Chr.9: 100020623-100020623 on Build GRCh38
rs759978577 Chr.9: 100020699-100020699 on Build GRCh38
rs755835966 Chr.9: 100020608-100020608 on Build GRCh38
rs905359579 Chr.9: 100020635-100020635 on Build GRCh38
rs145422046 Chr.9: 100020675-100020675 on Build GRCh38
rs369068577 Chr.9: 100020743-100020743 on Build GRCh38
rs971040381 Chr.9: 100020682-100020682 on Build GRCh38
rs761998029 Chr.9: 100020769-100020770 on Build GRCh38
rs771231030 Chr.9: 100020687-100020687 on Build GRCh38
rs150455027 Chr.9: 100020655-100020655 on Build GRCh38
rs774733634 Chr.9: 100020688-100020688 on Build GRCh38
rs984559635 Chr.9: 100020744-100020744 on Build GRCh38
rs55827103 Chr.9: 100020760-100020760 on Build GRCh38
rs374566095 Chr.9: 100020768-100020768 on Build GRCh38
rs752325297 Chr.9: 100020721-100020721 on Build GRCh38
rs149213852 Chr.9: 100020677-100020677 on Build GRCh38
rs375903014 Chr.9: 100020724-100020724 on Build GRCh38
rs371595032 Chr.9: 100020664-100020664 on Build GRCh38
rs373797701 Chr.9: 100020730-100020730 on Build GRCh38
rs866848699 Chr.9: 100020674-100020674 on Build GRCh38
rs535434128 Chr.9: 100020783-100020783 on Build GRCh38
rs777565786 Chr.9: 100020611-100020611 on Build GRCh38
rs753742172 Chr.9: 100020615-100020615 on Build GRCh38
rs531744364 Chr.9: 100020587-100020587 on Build GRCh38
rs753506440 Chr.9: 100020734-100020734 on Build GRCh38
rs41273941 Chr.9: 100020741-100020741 on Build GRCh38
rs77408644 Chr.9: 100020708-100020708 on Build GRCh38
rs867981269 Chr.9: 100020746-100020746 on Build GRCh38
rs768743894 Chr.9: 100020657-100020657 on Build GRCh38
rs778946012 Chr.9: 100020646-100020646 on Build GRCh38
rs757366760 Chr.9: 100020696-100020696 on Build GRCh38
rs561203722 Chr.9: 100020716-100020716 on Build GRCh38
rs766962467 Chr.9: 100020586-100020586 on Build GRCh38
rs1019880612 Chr.9: 100020747-100020747 on Build GRCh38
rs768073902 Chr.9: 100020715-100020715 on Build GRCh38

View Full Product Details