Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 111789746-111790015 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALDH2
Entrez Gene ID
217
Gene Name
aldehyde dehydrogenase 2 family (mitochondrial)
Gene Aliases
ALDH-E2, ALDHI, ALDM
UniGene
Hs.604551
Gene Chromosome Location
Chr.12: 111766887-111809985 on Build GRCh38
Transcripts
NM_000690.3 NM_001204889.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1947911 c.528G>A p.V176V Chr.12: 111789910-111789910 on Build GRCh38
6228963 c.450C>T p.A150A Chr.12: 111789832-111789832 on Build GRCh38
7424323 c.446A>G p.Y149C Chr.12: 111789828-111789828 on Build GRCh38
4953506 c.549T>G p.I183M Chr.12: 111789931-111789931 on Build GRCh38
6846121 c.479C>T p.T160I Chr.12: 111789861-111789861 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753125040 Chr.12: 111789829-111789829 on Build GRCh38
rs533704716 Chr.12: 111789874-111789874 on Build GRCh38
rs756650696 Chr.12: 111789831-111789831 on Build GRCh38
rs190463190 Chr.12: 111789806-111789806 on Build GRCh38
rs765448984 Chr.12: 111789934-111789934 on Build GRCh38
rs769168320 Chr.12: 111789971-111789971 on Build GRCh38
rs907230062 Chr.12: 111789909-111789910 on Build GRCh38
rs754896704 Chr.12: 111789962-111789962 on Build GRCh38
rs747615625 Chr.12: 111789969-111789969 on Build GRCh38
rs779941847 Chr.12: 111789867-111789867 on Build GRCh38
rs748608292 Chr.12: 111789963-111789975 on Build GRCh38
rs758715136 Chr.12: 111789958-111789958 on Build GRCh38
rs751889386 Chr.12: 111789800-111789800 on Build GRCh38
rs768023125 Chr.12: 111789814-111789814 on Build GRCh38
rs753847903 Chr.12: 111789922-111789922 on Build GRCh38
rs781313528 Chr.12: 111789968-111789968 on Build GRCh38
rs141629803 Chr.12: 111789830-111789830 on Build GRCh38
rs781757716 Chr.12: 111789774-111789775 on Build GRCh38
rs776223807 Chr.12: 111789885-111789885 on Build GRCh38
rs762718195 Chr.12: 111789897-111789897 on Build GRCh38
rs368356402 Chr.12: 111789896-111789896 on Build GRCh38
rs1054678169 Chr.12: 111789768-111789768 on Build GRCh38
rs951845127 Chr.12: 111789950-111789950 on Build GRCh38
rs77384237 Chr.12: 111789993-111789993 on Build GRCh38
rs748886318 Chr.12: 111789986-111789986 on Build GRCh38
rs144375294 Chr.12: 111789875-111789875 on Build GRCh38
rs753972907 Chr.12: 111789847-111789847 on Build GRCh38
rs191240032 Chr.12: 111789933-111789933 on Build GRCh38
rs866683603 Chr.12: 111789928-111789928 on Build GRCh38
rs768251860 Chr.12: 111789772-111789772 on Build GRCh38
rs1009289495 Chr.12: 111789846-111789846 on Build GRCh38
rs772821114 Chr.12: 111789975-111789975 on Build GRCh38
rs764594848 Chr.12: 111789920-111789920 on Build GRCh38
rs776347741 Chr.12: 111789774-111789774 on Build GRCh38
rs746814335 Chr.12: 111789960-111789960 on Build GRCh38
rs375708006 Chr.12: 111789893-111789893 on Build GRCh38
rs779701360 Chr.12: 111789959-111789959 on Build GRCh38
rs758149074 Chr.12: 111789866-111789866 on Build GRCh38
rs976336262 Chr.12: 111789972-111789972 on Build GRCh38
rs201725825 Chr.12: 111789919-111789919 on Build GRCh38
rs892067750 Chr.12: 111789797-111789797 on Build GRCh38
rs767739457 Chr.12: 111789905-111789905 on Build GRCh38
rs746940858 Chr.12: 111789870-111789870 on Build GRCh38
rs775896862 Chr.12: 111789913-111789913 on Build GRCh38
rs764810743 Chr.12: 111789796-111789796 on Build GRCh38
rs747670612 Chr.12: 111789889-111789889 on Build GRCh38
rs199847950 Chr.12: 111789935-111789935 on Build GRCh38
rs575953094 Chr.12: 111789853-111789853 on Build GRCh38
rs779006912 Chr.12: 111789854-111789854 on Build GRCh38
rs372921830 Chr.12: 111789832-111789832 on Build GRCh38

View Full Product Details