Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 207899784-207900049 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD34
Entrez Gene ID
947
Gene Name
CD34 molecule
Gene Aliases
-
UniGene
Hs.374990
Gene Chromosome Location
Chr.1: 207886538-207911338 on Build GRCh38
Transcripts
NM_001025109.1 NM_001773.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3482805 c.202G>A p.G68R Chr.1: 207899881-207899881 on Build GRCh38
5024461 c.154G>A p.V52I Chr.1: 207899929-207899929 on Build GRCh38
3482806 c.97C>T p.L33F Chr.1: 207899986-207899986 on Build GRCh38
5992995 c.92T>C p.M31T Chr.1: 207899991-207899991 on Build GRCh38
678833 c.114T>A p.T38T Chr.1: 207899969-207899969 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs954687741 Chr.1: 207899824-207899824 on Build GRCh38
rs766045452 Chr.1: 207899840-207899840 on Build GRCh38
rs766205855 Chr.1: 207899957-207899957 on Build GRCh38
rs200225798 Chr.1: 207899844-207899844 on Build GRCh38
rs988032193 Chr.1: 207899959-207899959 on Build GRCh38
rs760459179 Chr.1: 207899942-207899942 on Build GRCh38
rs746310748 Chr.1: 207899867-207899867 on Build GRCh38
rs764284174 Chr.1: 207899847-207899847 on Build GRCh38
rs978892034 Chr.1: 207899892-207899892 on Build GRCh38
rs35359362 Chr.1: 207899860-207899860 on Build GRCh38
rs756637939 Chr.1: 207900015-207900015 on Build GRCh38
rs760543727 Chr.1: 207899839-207899839 on Build GRCh38
rs373533705 Chr.1: 207899992-207899992 on Build GRCh38
rs759370976 Chr.1: 207899974-207899974 on Build GRCh38
rs776343546 Chr.1: 207899970-207899970 on Build GRCh38
rs750366745 Chr.1: 207899836-207899836 on Build GRCh38
rs761408025 Chr.1: 207899827-207899827 on Build GRCh38
rs925093528 Chr.1: 207899900-207899900 on Build GRCh38
rs780607094 Chr.1: 207900016-207900016 on Build GRCh38
rs376656432 Chr.1: 207899861-207899861 on Build GRCh38
rs747713193 Chr.1: 207899898-207899898 on Build GRCh38
rs777207314 Chr.1: 207899889-207899889 on Build GRCh38
rs764339451 Chr.1: 207899975-207899975 on Build GRCh38
rs751704997 Chr.1: 207899854-207899854 on Build GRCh38
rs778631125 Chr.1: 207899982-207899982 on Build GRCh38
rs113112247 Chr.1: 207900004-207900004 on Build GRCh38
rs756508294 Chr.1: 207899870-207899870 on Build GRCh38
rs529148972 Chr.1: 207899995-207899995 on Build GRCh38
rs1046481057 Chr.1: 207900010-207900010 on Build GRCh38
rs369149182 Chr.1: 207899907-207899907 on Build GRCh38
rs768260843 Chr.1: 207899882-207899882 on Build GRCh38
rs762447505 Chr.1: 207899813-207899813 on Build GRCh38
rs767607306 Chr.1: 207899834-207899834 on Build GRCh38
rs768227484 Chr.1: 207899817-207899817 on Build GRCh38
rs111712412 Chr.1: 207900005-207900005 on Build GRCh38
rs773316312 Chr.1: 207899912-207899912 on Build GRCh38
rs913820873 Chr.1: 207899978-207899978 on Build GRCh38
rs1020831010 Chr.1: 207899818-207899818 on Build GRCh38
rs757295859 Chr.1: 207899857-207899857 on Build GRCh38
rs201694419 Chr.1: 207899842-207899842 on Build GRCh38
rs774063710 Chr.1: 207899820-207899820 on Build GRCh38
rs138486900 Chr.1: 207899988-207899988 on Build GRCh38
rs749652320 Chr.1: 207899874-207899874 on Build GRCh38
rs774120895 Chr.1: 207899895-207899895 on Build GRCh38
rs780226626 Chr.1: 207899872-207899872 on Build GRCh38
rs137941074 Chr.1: 207899985-207899985 on Build GRCh38
rs932746492 Chr.1: 207899918-207899918 on Build GRCh38
rs926350274 Chr.1: 207900011-207900011 on Build GRCh38

View Full Product Details