Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 145064425-145064652 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANAPC10
Entrez Gene ID
10393
Gene Name
anaphase promoting complex subunit 10
Gene Aliases
APC10, DOC1
UniGene
Hs.480876
Gene Chromosome Location
Chr.4: 144967117-145098541 on Build GRCh38
Transcripts
NM_001256706.1 NM_001256707.1 NM_001256708.1 NM_001256709.1 NM_001256710.1 NM_001256711.1 NM_001256712.1 NM_001318367.1 NM_014885.4 NR_046345.1 XM_011531524.2 XM_011531525.2 XM_011531526.2 XM_011531527.2 XM_011531529.2 XM_011531530.2 XM_011531531.2 XM_011531532.2 XM_017007638.1 XM_017007639.1 XM_017007640.1 XM_017007641.1 XM_017007642.1 XM_017007643.1 XM_017007644.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7312795 c.269C>T p.T90I Chr.4: 145064630-145064630 on Build GRCh38
1427343 c.267T>C p.Y89Y Chr.4: 145064632-145064632 on Build GRCh38
136444 c.307C>T p.H103Y Chr.4: 145064592-145064592 on Build GRCh38
3600626 c.254C>T p.S85F Chr.4: 145064645-145064645 on Build GRCh38
4637945 c.309C>T p.H103H Chr.4: 145064590-145064590 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs954909755 Chr.4: 145064603-145064603 on Build GRCh38
rs749794176 Chr.4: 145064538-145064538 on Build GRCh38
rs35754508 Chr.4: 145064464-145064464 on Build GRCh38
rs765089127 Chr.4: 145064605-145064605 on Build GRCh38
rs1000270711 Chr.4: 145064582-145064582 on Build GRCh38
rs759790944 Chr.4: 145064559-145064559 on Build GRCh38
rs372401997 Chr.4: 145064590-145064590 on Build GRCh38
rs750440669 Chr.4: 145064607-145064607 on Build GRCh38
rs778279410 Chr.4: 145064532-145064532 on Build GRCh38
rs761610901 Chr.4: 145064600-145064600 on Build GRCh38
rs1037400798 Chr.4: 145064473-145064473 on Build GRCh38
rs748499295 Chr.4: 145064524-145064524 on Build GRCh38
rs369778610 Chr.4: 145064588-145064588 on Build GRCh38
rs766028459 Chr.4: 145064620-145064620 on Build GRCh38
rs895661070 Chr.4: 145064519-145064519 on Build GRCh38
rs1023065805 Chr.4: 145064545-145064546 on Build GRCh38
rs969879082 Chr.4: 145064610-145064610 on Build GRCh38
rs536139076 Chr.4: 145064470-145064470 on Build GRCh38
rs567515853 Chr.4: 145064467-145064467 on Build GRCh38
rs929250095 Chr.4: 145064492-145064492 on Build GRCh38
rs781441929 Chr.4: 145064521-145064521 on Build GRCh38
rs34600335 Chr.4: 145064579-145064580 on Build GRCh38
rs34647238 Chr.4: 145064472-145064472 on Build GRCh38
rs905963724 Chr.4: 145064572-145064572 on Build GRCh38
rs780429965 Chr.4: 145064528-145064528 on Build GRCh38
rs770221015 Chr.4: 145064528-145064528 on Build GRCh38
rs762962943 Chr.4: 145064612-145064612 on Build GRCh38
rs376827906 Chr.4: 145064540-145064540 on Build GRCh38
rs569862410 Chr.4: 145064498-145064498 on Build GRCh38
rs898826583 Chr.4: 145064488-145064488 on Build GRCh38
rs1047641622 Chr.4: 145064517-145064517 on Build GRCh38
rs774390312 Chr.4: 145064544-145064544 on Build GRCh38

View Full Product Details