Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 38254614-38254859 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLEC14A
Entrez Gene ID
161198
Gene Name
C-type lectin domain family 14 member A
Gene Aliases
C14orf27, CEG1, EGFR-5
UniGene
Hs.525307
Gene Chromosome Location
Chr.14: 38254000-38256370 on Build GRCh38
Transcripts
NM_175060.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5382250 c.1306G>A p.E436K Chr.14: 38254717-38254717 on Build GRCh38
5382249 c.1387G>A p.G463R Chr.14: 38254636-38254636 on Build GRCh38
7328202 c.1199T>C p.F400S Chr.14: 38254824-38254824 on Build GRCh38
3793693 c.1245G>C p.M415I Chr.14: 38254778-38254778 on Build GRCh38
279924 c.1398C>T p.V466V Chr.14: 38254625-38254625 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755930300 Chr.14: 38254704-38254704 on Build GRCh38
rs559027633 Chr.14: 38254784-38254784 on Build GRCh38
rs1009956918 Chr.14: 38254634-38254634 on Build GRCh38
rs759225727 Chr.14: 38254676-38254676 on Build GRCh38
rs770308576 Chr.14: 38254638-38254638 on Build GRCh38
rs755172731 Chr.14: 38254740-38254740 on Build GRCh38
rs773883500 Chr.14: 38254768-38254768 on Build GRCh38
rs1040140814 Chr.14: 38254826-38254826 on Build GRCh38
rs34471237 Chr.14: 38254739-38254739 on Build GRCh38
rs780176144 Chr.14: 38254722-38254722 on Build GRCh38
rs375642755 Chr.14: 38254658-38254658 on Build GRCh38
rs369867645 Chr.14: 38254833-38254833 on Build GRCh38
rs781344993 Chr.14: 38254745-38254745 on Build GRCh38
rs770417486 Chr.14: 38254763-38254763 on Build GRCh38
rs372611166 Chr.14: 38254831-38254831 on Build GRCh38
rs758464635 Chr.14: 38254721-38254721 on Build GRCh38
rs752560289 Chr.14: 38254691-38254691 on Build GRCh38
rs753783862 Chr.14: 38254810-38254810 on Build GRCh38
rs775213023 Chr.14: 38254782-38254782 on Build GRCh38
rs972768480 Chr.14: 38254811-38254811 on Build GRCh38
rs775279333 Chr.14: 38254638-38254641 on Build GRCh38
rs867156025 Chr.14: 38254679-38254679 on Build GRCh38
rs764127595 Chr.14: 38254710-38254710 on Build GRCh38
rs758918675 Chr.14: 38254774-38254774 on Build GRCh38
rs78287081 Chr.14: 38254838-38254838 on Build GRCh38
rs749793261 Chr.14: 38254762-38254762 on Build GRCh38
rs1006134019 Chr.14: 38254737-38254737 on Build GRCh38
rs112144257 Chr.14: 38254708-38254708 on Build GRCh38
rs753751121 Chr.14: 38254715-38254715 on Build GRCh38
rs763798779 Chr.14: 38254796-38254797 on Build GRCh38
rs1035916478 Chr.14: 38254637-38254637 on Build GRCh38
rs747234597 Chr.14: 38254734-38254734 on Build GRCh38
rs951947364 Chr.14: 38254733-38254733 on Build GRCh38
rs770077651 Chr.14: 38254760-38254760 on Build GRCh38
rs910482314 Chr.14: 38254803-38254803 on Build GRCh38
rs1045894939 Chr.14: 38254648-38254648 on Build GRCh38
rs760522759 Chr.14: 38254741-38254741 on Build GRCh38
rs767466452 Chr.14: 38254681-38254681 on Build GRCh38
rs368598659 Chr.14: 38254772-38254772 on Build GRCh38
rs530380827 Chr.14: 38254836-38254838 on Build GRCh38
rs977270113 Chr.14: 38254730-38254730 on Build GRCh38
rs751685062 Chr.14: 38254832-38254832 on Build GRCh38
rs572756352 Chr.14: 38254805-38254805 on Build GRCh38
rs767305073 Chr.14: 38254781-38254781 on Build GRCh38
rs150923168 Chr.14: 38254664-38254664 on Build GRCh38
rs757186521 Chr.14: 38254829-38254829 on Build GRCh38
rs182049277 Chr.14: 38254652-38254652 on Build GRCh38

View Full Product Details