Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 123937618-123937857 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC92
Entrez Gene ID
80212
Gene Name
coiled-coil domain containing 92
Gene Aliases
-
UniGene
Hs.114111
Gene Chromosome Location
Chr.12: 123936409-123972985 on Build GRCh38
Transcripts
NM_001304957.1 NM_001304958.1 NM_001304959.1 NM_001304960.1 NM_001304961.1 NM_025140.2 XM_005253624.2 XM_017019984.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7185176 c.225_226insTAGGAG p.G75_D76ins* Chr.12: 123937828-123937829 on Build GRCh38
6622683 c.377G>A p.R126Q Chr.12: 123937677-123937677 on Build GRCh38
7126974 c.413G>A p.S138N Chr.12: 123937641-123937641 on Build GRCh38
5553737 c.270G>T p.E90D Chr.12: 123937784-123937784 on Build GRCh38
5508696 c.274G>A p.E92K Chr.12: 123937780-123937780 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs143608456 Chr.12: 123937691-123937691 on Build GRCh38
rs763400066 Chr.12: 123937826-123937826 on Build GRCh38
rs756831339 Chr.12: 123937728-123937728 on Build GRCh38
rs760855449 Chr.12: 123937711-123937711 on Build GRCh38
rs148048206 Chr.12: 123937703-123937703 on Build GRCh38
rs202226542 Chr.12: 123937714-123937714 on Build GRCh38
rs766744858 Chr.12: 123937712-123937712 on Build GRCh38
rs771043483 Chr.12: 123937794-123937794 on Build GRCh38
rs761013340 Chr.12: 123937796-123937799 on Build GRCh38
rs780064664 Chr.12: 123937673-123937673 on Build GRCh38
rs778675070 Chr.12: 123937773-123937773 on Build GRCh38
rs543085766 Chr.12: 123937802-123937802 on Build GRCh38
rs937340391 Chr.12: 123937688-123937688 on Build GRCh38
rs745427235 Chr.12: 123937637-123937637 on Build GRCh38
rs370285111 Chr.12: 123937723-123937723 on Build GRCh38
rs200055938 Chr.12: 123937760-123937760 on Build GRCh38
rs762009006 Chr.12: 123937705-123937705 on Build GRCh38
rs777805548 Chr.12: 123937686-123937686 on Build GRCh38
rs140699606 Chr.12: 123937805-123937805 on Build GRCh38
rs147257055 Chr.12: 123937754-123937754 on Build GRCh38
rs201517947 Chr.12: 123937799-123937799 on Build GRCh38
rs184378821 Chr.12: 123937753-123937753 on Build GRCh38
rs573154431 Chr.12: 123937784-123937784 on Build GRCh38
rs753183790 Chr.12: 123937726-123937726 on Build GRCh38
rs201632527 Chr.12: 123937766-123937766 on Build GRCh38
rs765888240 Chr.12: 123937821-123937821 on Build GRCh38
rs747976096 Chr.12: 123937776-123937776 on Build GRCh38
rs865975725 Chr.12: 123937704-123937704 on Build GRCh38
rs775978105 Chr.12: 123937825-123937825 on Build GRCh38
rs747018104 Chr.12: 123937787-123937787 on Build GRCh38
rs749238387 Chr.12: 123937769-123937769 on Build GRCh38
rs772402768 Chr.12: 123937706-123937706 on Build GRCh38
rs200549175 Chr.12: 123937741-123937741 on Build GRCh38
rs777304210 Chr.12: 123937713-123937713 on Build GRCh38
rs200385849 Chr.12: 123937777-123937777 on Build GRCh38
rs773134124 Chr.12: 123937710-123937710 on Build GRCh38
rs200883084 Chr.12: 123937772-123937772 on Build GRCh38
rs1000003607 Chr.12: 123937694-123937694 on Build GRCh38
rs769598142 Chr.12: 123937665-123937665 on Build GRCh38
rs35794481 Chr.12: 123937715-123937715 on Build GRCh38
rs767039955 Chr.12: 123937834-123937834 on Build GRCh38
rs768445428 Chr.12: 123937692-123937692 on Build GRCh38
rs1019836472 Chr.12: 123937817-123937817 on Build GRCh38

View Full Product Details