Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 116720219-116720492 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C12orf49
Entrez Gene ID
79794
Gene Name
chromosome 12 open reading frame 49
Gene Aliases
-
UniGene
Hs.592011
Gene Chromosome Location
Chr.12: 116713320-116738070 on Build GRCh38
Transcripts
NM_024738.2 XM_005253937.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1706337 c.382T>A p.Y128N Chr.12: 116720334-116720334 on Build GRCh38
7080473 c.326C>T p.P109L Chr.12: 116720390-116720390 on Build GRCh38
1717107 c.294G>C p.L98F Chr.12: 116720422-116720422 on Build GRCh38
4575070 c.243G>A p.T81T Chr.12: 116720383-116720383 on Build GRCh38
2173352 c.406C>A p.L136M Chr.12: 116720310-116720310 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780903919 Chr.12: 116720272-116720272 on Build GRCh38
rs751817746 Chr.12: 116720350-116720350 on Build GRCh38
rs766567103 Chr.12: 116720345-116720345 on Build GRCh38
rs777290744 Chr.12: 116720288-116720288 on Build GRCh38
rs766590101 Chr.12: 116720253-116720253 on Build GRCh38
rs766049370 Chr.12: 116720455-116720456 on Build GRCh38
rs376518938 Chr.12: 116720395-116720395 on Build GRCh38
rs756002577 Chr.12: 116720279-116720279 on Build GRCh38
rs763132487 Chr.12: 116720252-116720252 on Build GRCh38
rs576717796 Chr.12: 116720246-116720246 on Build GRCh38
rs756781540 Chr.12: 116720401-116720401 on Build GRCh38
rs763084366 Chr.12: 116720344-116720344 on Build GRCh38
rs904984197 Chr.12: 116720322-116720322 on Build GRCh38
rs773463635 Chr.12: 116720337-116720337 on Build GRCh38
rs770533929 Chr.12: 116720310-116720310 on Build GRCh38
rs756942210 Chr.12: 116720300-116720300 on Build GRCh38
rs745539360 Chr.12: 116720415-116720415 on Build GRCh38
rs774252952 Chr.12: 116720450-116720450 on Build GRCh38
rs762443477 Chr.12: 116720333-116720333 on Build GRCh38
rs770401818 Chr.12: 116720336-116720336 on Build GRCh38
rs750491583 Chr.12: 116720250-116720250 on Build GRCh38
rs752437374 Chr.12: 116720384-116720384 on Build GRCh38
rs201676406 Chr.12: 116720437-116720437 on Build GRCh38
rs375002609 Chr.12: 116720429-116720429 on Build GRCh38
rs769078292 Chr.12: 116720327-116720327 on Build GRCh38
rs778627083 Chr.12: 116720316-116720316 on Build GRCh38
rs752633472 Chr.12: 116720273-116720273 on Build GRCh38
rs747392112 Chr.12: 116720320-116720320 on Build GRCh38
rs373559755 Chr.12: 116720338-116720338 on Build GRCh38
rs76041009 Chr.12: 116720383-116720383 on Build GRCh38
rs201073590 Chr.12: 116720470-116720470 on Build GRCh38
rs753360322 Chr.12: 116720396-116720396 on Build GRCh38
rs771043506 Chr.12: 116720444-116720444 on Build GRCh38
rs751873889 Chr.12: 116720457-116720458 on Build GRCh38
rs916987290 Chr.12: 116720349-116720349 on Build GRCh38
rs778229935 Chr.12: 116720410-116720410 on Build GRCh38
rs759824137 Chr.12: 116720451-116720451 on Build GRCh38
rs141835393 Chr.12: 116720421-116720421 on Build GRCh38
rs139514585 Chr.12: 116720443-116720443 on Build GRCh38
rs762829170 Chr.12: 116720448-116720449 on Build GRCh38
rs146281512 Chr.12: 116720442-116720442 on Build GRCh38
rs781708387 Chr.12: 116720425-116720425 on Build GRCh38
rs748811812 Chr.12: 116720294-116720294 on Build GRCh38
rs751376631 Chr.12: 116720269-116720269 on Build GRCh38
rs777071119 Chr.12: 116720331-116720331 on Build GRCh38
rs369548022 Chr.12: 116720270-116720270 on Build GRCh38
rs756096562 Chr.12: 116720394-116720394 on Build GRCh38
rs750271935 Chr.12: 116720258-116720258 on Build GRCh38
rs558278615 Chr.12: 116720436-116720436 on Build GRCh38
rs376281087 Chr.12: 116720247-116720247 on Build GRCh38

View Full Product Details