Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 46781517-46781748 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ESD
Entrez Gene ID
2098
Gene Name
esterase D
Gene Aliases
FGH
UniGene
Hs.432491
Gene Chromosome Location
Chr.13: 46771256-46797681 on Build GRCh38
Transcripts
NM_001984.1 XM_005266278.2 XM_011534954.1 XM_017020414.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3688741 c.441C>T p.G147G Chr.13: 46781556-46781556 on Build GRCh38
6001057 c.386C>A p.P129H Chr.13: 46781611-46781611 on Build GRCh38
3885393 c.419C>T p.P140L Chr.13: 46781578-46781578 on Build GRCh38
947746 c.397A>C p.N133H Chr.13: 46781600-46781600 on Build GRCh38
6656308 c.421delC p.Q141fs*16 Chr.13: 46781576-46781576 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780715105 Chr.13: 46781620-46781620 on Build GRCh38
rs1015097466 Chr.13: 46781649-46781649 on Build GRCh38
rs7989468 Chr.13: 46781707-46781707 on Build GRCh38
rs775752401 Chr.13: 46781545-46781545 on Build GRCh38
rs754689056 Chr.13: 46781618-46781618 on Build GRCh38
rs766702005 Chr.13: 46781603-46781603 on Build GRCh38
rs768011202 Chr.13: 46781588-46781588 on Build GRCh38
rs565787210 Chr.13: 46781667-46781667 on Build GRCh38
rs549690200 Chr.13: 46781538-46781538 on Build GRCh38
rs35178907 Chr.13: 46781620-46781620 on Build GRCh38
rs963351190 Chr.13: 46781652-46781652 on Build GRCh38
rs370424297 Chr.13: 46781547-46781547 on Build GRCh38
rs746767554 Chr.13: 46781539-46781539 on Build GRCh38
rs149029670 Chr.13: 46781549-46781549 on Build GRCh38
rs538728073 Chr.13: 46781593-46781593 on Build GRCh38
rs113789969 Chr.13: 46781641-46781641 on Build GRCh38
rs987033243 Chr.13: 46781666-46781666 on Build GRCh38
rs780911252 Chr.13: 46781648-46781648 on Build GRCh38
rs761989853 Chr.13: 46781565-46781565 on Build GRCh38
rs951585627 Chr.13: 46781637-46781637 on Build GRCh38
rs764260255 Chr.13: 46781548-46781548 on Build GRCh38
rs746286682 Chr.13: 46781619-46781619 on Build GRCh38
rs770740917 Chr.13: 46781644-46781644 on Build GRCh38
rs564338173 Chr.13: 46781626-46781626 on Build GRCh38
rs146164024 Chr.13: 46781564-46781564 on Build GRCh38
rs761069005 Chr.13: 46781596-46781596 on Build GRCh38
rs757923497 Chr.13: 46781631-46781631 on Build GRCh38
rs777572363 Chr.13: 46781657-46781657 on Build GRCh38
rs1030899389 Chr.13: 46781553-46781553 on Build GRCh38
rs927429490 Chr.13: 46781700-46781700 on Build GRCh38
rs770666625 Chr.13: 46781542-46781542 on Build GRCh38
rs753515959 Chr.13: 46781610-46781610 on Build GRCh38
rs553862652 Chr.13: 46781640-46781640 on Build GRCh38
rs533644943 Chr.13: 46781703-46781703 on Build GRCh38
rs898917227 Chr.13: 46781583-46781583 on Build GRCh38
rs749517506 Chr.13: 46781655-46781655 on Build GRCh38
rs778538153 Chr.13: 46781619-46781619 on Build GRCh38

View Full Product Details