Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 43129184-43129409 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BIK
Entrez Gene ID
638
Gene Name
BCL2 interacting killer
Gene Aliases
BIP1, BP4, NBK
UniGene
Hs.475055
Gene Chromosome Location
Chr.22: 43110748-43129712 on Build GRCh38
Transcripts
NM_001197.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3062071 c.458G>A p.G153E Chr.22: 43129280-43129280 on Build GRCh38
301430 c.426_427GC>AA p.L142>? Chr.22: 43129248-43129249 on Build GRCh38
6297333 c.400G>T p.E134* Chr.22: 43129222-43129222 on Build GRCh38
4665715 c.447G>T p.P149P Chr.22: 43129269-43129269 on Build GRCh38
4665714 c.429_434delGCTGGC p.A145_L146delAL Chr.22: 43129251-43129256 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs560282375 Chr.22: 43129281-43129281 on Build GRCh38
rs985261903 Chr.22: 43129217-43129217 on Build GRCh38
rs529029498 Chr.22: 43129311-43129311 on Build GRCh38
rs759887547 Chr.22: 43129229-43129246 on Build GRCh38
rs201004577 Chr.22: 43129237-43129237 on Build GRCh38
rs758385246 Chr.22: 43129238-43129267 on Build GRCh38
rs145876995 Chr.22: 43129250-43129250 on Build GRCh38
rs750496593 Chr.22: 43129275-43129275 on Build GRCh38
rs770706686 Chr.22: 43129292-43129292 on Build GRCh38
rs751517650 Chr.22: 43129265-43129266 on Build GRCh38
rs753912642 Chr.22: 43129230-43129256 on Build GRCh38
rs780977608 Chr.22: 43129308-43129308 on Build GRCh38
rs11574527 Chr.22: 43129265-43129265 on Build GRCh38
rs768945630 Chr.22: 43129280-43129280 on Build GRCh38
rs565847493 Chr.22: 43129316-43129316 on Build GRCh38
rs780961178 Chr.22: 43129239-43129247 on Build GRCh38
rs769377877 Chr.22: 43129242-43129256 on Build GRCh38
rs1009261413 Chr.22: 43129261-43129261 on Build GRCh38
rs762372547 Chr.22: 43129247-43129248 on Build GRCh38
rs1039226514 Chr.22: 43129238-43129238 on Build GRCh38
rs759526038 Chr.22: 43129210-43129210 on Build GRCh38
rs770689042 Chr.22: 43129239-43129259 on Build GRCh38
rs766452936 Chr.22: 43129355-43129355 on Build GRCh38
rs866956508 Chr.22: 43129226-43129226 on Build GRCh38
rs755054456 Chr.22: 43129256-43129257 on Build GRCh38
rs950752987 Chr.22: 43129310-43129310 on Build GRCh38
rs201603625 Chr.22: 43129325-43129325 on Build GRCh38
rs762089105 Chr.22: 43129321-43129321 on Build GRCh38
rs921259818 Chr.22: 43129263-43129264 on Build GRCh38
rs768176790 Chr.22: 43129248-43129268 on Build GRCh38
rs965427251 Chr.22: 43129229-43129258 on Build GRCh38
rs867355694 Chr.22: 43129255-43129255 on Build GRCh38
rs760992548 Chr.22: 43129253-43129254 on Build GRCh38
rs773647520 Chr.22: 43129251-43129268 on Build GRCh38
rs542081559 Chr.22: 43129239-43129239 on Build GRCh38
rs545887424 Chr.22: 43129268-43129268 on Build GRCh38
rs62637687 Chr.22: 43129247-43129247 on Build GRCh38
rs4988441 Chr.22: 43129315-43129315 on Build GRCh38
rs62640385 Chr.22: 43129272-43129272 on Build GRCh38
rs201234434 Chr.22: 43129278-43129278 on Build GRCh38
rs760336241 Chr.22: 43129267-43129267 on Build GRCh38
rs777532003 Chr.22: 43129239-43129241 on Build GRCh38
rs376511782 Chr.22: 43129324-43129324 on Build GRCh38
rs975840608 Chr.22: 43129254-43129255 on Build GRCh38
rs767480690 Chr.22: 43129212-43129212 on Build GRCh38
rs762638652 Chr.22: 43129246-43129246 on Build GRCh38
rs150958238 Chr.22: 43129326-43129326 on Build GRCh38
rs755791685 Chr.22: 43129313-43129314 on Build GRCh38
rs140238366 Chr.22: 43129236-43129236 on Build GRCh38
rs757446636 Chr.22: 43129271-43129271 on Build GRCh38
rs527805900 Chr.22: 43129264-43129264 on Build GRCh38
rs762326871 Chr.22: 43129244-43129245 on Build GRCh38
rs148407250 Chr.22: 43129256-43129256 on Build GRCh38
rs777795806 Chr.22: 43129221-43129221 on Build GRCh38
rs753983450 Chr.22: 43129255-43129256 on Build GRCh38
rs752427877 Chr.22: 43129214-43129214 on Build GRCh38
rs892112683 Chr.22: 43129257-43129257 on Build GRCh38
rs770618715 Chr.22: 43129249-43129249 on Build GRCh38
rs781601891 Chr.22: 43129282-43129282 on Build GRCh38
rs199601984 Chr.22: 43129279-43129279 on Build GRCh38
rs1048903862 Chr.22: 43129332-43129333 on Build GRCh38
rs766661995 Chr.22: 43129256-43129267 on Build GRCh38
rs923479366 Chr.22: 43129242-43129242 on Build GRCh38
rs759617722 Chr.22: 43129259-43129259 on Build GRCh38
rs773998673 Chr.22: 43129296-43129296 on Build GRCh38
rs756361244 Chr.22: 43129218-43129218 on Build GRCh38
rs367976515 Chr.22: 43129309-43129309 on Build GRCh38
rs772701491 Chr.22: 43129243-43129243 on Build GRCh38
rs750736679 Chr.22: 43129227-43129244 on Build GRCh38
rs775247531 Chr.22: 43129312-43129312 on Build GRCh38
rs950774622 Chr.22: 43129216-43129216 on Build GRCh38
rs757149765 Chr.22: 43129306-43129330 on Build GRCh38
rs765345235 Chr.22: 43129264-43129268 on Build GRCh38
rs745609997 Chr.22: 43129310-43129311 on Build GRCh38
rs756403552 Chr.22: 43129226-43129227 on Build GRCh38
rs756855144 Chr.22: 43129283-43129283 on Build GRCh38
rs371566011 Chr.22: 43129314-43129314 on Build GRCh38
rs754045953 Chr.22: 43129269-43129269 on Build GRCh38
rs775000855 Chr.22: 43129245-43129256 on Build GRCh38
rs111513951 Chr.22: 43129365-43129365 on Build GRCh38
rs774766775 Chr.22: 43129248-43129256 on Build GRCh38
rs532663391 Chr.22: 43129222-43129222 on Build GRCh38
rs201062941 Chr.22: 43129225-43129225 on Build GRCh38
rs746935217 Chr.22: 43129238-43129239 on Build GRCh38
rs768065717 Chr.22: 43129250-43129251 on Build GRCh38
rs544964298 Chr.22: 43129223-43129223 on Build GRCh38

View Full Product Details