Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 224401129-224401366 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM124B
Entrez Gene ID
79843
Gene Name
family with sequence similarity 124 member B
Gene Aliases
-
UniGene
Hs.147585
Gene Chromosome Location
Chr.2: 224378698-224401994 on Build GRCh38
Transcripts
NM_001122779.1 NM_024785.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
224218 c.487G>A p.E163K Chr.2: 224401282-224401282 on Build GRCh38
6090764 c.438T>A p.F146L Chr.2: 224401331-224401331 on Build GRCh38
3578268 c.404C>T p.S135F Chr.2: 224401365-224401365 on Build GRCh38
1669805 c.446A>G p.Y149C Chr.2: 224401323-224401323 on Build GRCh38
1017102 c.408G>T p.E136D Chr.2: 224401361-224401361 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750775324 Chr.2: 224401173-224401173 on Build GRCh38
rs746749968 Chr.2: 224401231-224401231 on Build GRCh38
rs371374350 Chr.2: 224401161-224401161 on Build GRCh38
rs769064165 Chr.2: 224401322-224401323 on Build GRCh38
rs756690006 Chr.2: 224401190-224401190 on Build GRCh38
rs761176688 Chr.2: 224401305-224401305 on Build GRCh38
rs752187289 Chr.2: 224401329-224401329 on Build GRCh38
rs761291964 Chr.2: 224401308-224401309 on Build GRCh38
rs780648923 Chr.2: 224401193-224401193 on Build GRCh38
rs766918271 Chr.2: 224401246-224401246 on Build GRCh38
rs151315340 Chr.2: 224401169-224401169 on Build GRCh38
rs747779894 Chr.2: 224401214-224401214 on Build GRCh38
rs928020765 Chr.2: 224401179-224401179 on Build GRCh38
rs370141388 Chr.2: 224401218-224401218 on Build GRCh38
rs982165858 Chr.2: 224401174-224401174 on Build GRCh38
rs375219740 Chr.2: 224401237-224401237 on Build GRCh38
rs769909652 Chr.2: 224401195-224401195 on Build GRCh38
rs758001941 Chr.2: 224401339-224401339 on Build GRCh38
rs764234329 Chr.2: 224401160-224401160 on Build GRCh38
rs571017410 Chr.2: 224401192-224401192 on Build GRCh38
rs754512892 Chr.2: 224401211-224401211 on Build GRCh38
rs768671588 Chr.2: 224401283-224401283 on Build GRCh38
rs776166992 Chr.2: 224401343-224401343 on Build GRCh38
rs138469734 Chr.2: 224401172-224401172 on Build GRCh38
rs565742600 Chr.2: 224401286-224401286 on Build GRCh38
rs754381768 Chr.2: 224401207-224401207 on Build GRCh38
rs755464550 Chr.2: 224401316-224401316 on Build GRCh38
rs373140603 Chr.2: 224401300-224401300 on Build GRCh38
rs777463842 Chr.2: 224401346-224401346 on Build GRCh38
rs200113424 Chr.2: 224401217-224401217 on Build GRCh38
rs147930186 Chr.2: 224401176-224401176 on Build GRCh38
rs1001233131 Chr.2: 224401341-224401342 on Build GRCh38
rs142573389 Chr.2: 224401310-224401310 on Build GRCh38
rs376596730 Chr.2: 224401303-224401303 on Build GRCh38
rs754436618 Chr.2: 224401313-224401313 on Build GRCh38
rs200639231 Chr.2: 224401164-224401164 on Build GRCh38
rs369547761 Chr.2: 224401239-224401239 on Build GRCh38
rs767708285 Chr.2: 224401301-224401301 on Build GRCh38
rs956920973 Chr.2: 224401242-224401242 on Build GRCh38
rs150435753 Chr.2: 224401323-224401323 on Build GRCh38
rs778455233 Chr.2: 224401213-224401213 on Build GRCh38

View Full Product Details