Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 21516626-21516853 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SKIDA1
Entrez Gene ID
387640
Gene Name
SKI/DACH domain containing 1
Gene Aliases
C10orf140, DLN-1
UniGene
Hs.350848
Gene Chromosome Location
Chr.10: 21513475-21525682 on Build GRCh38
Transcripts
NM_207371.3 XM_005252450.3 XM_011519480.2 XM_017016225.1 XM_017016226.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6237901 c.1017C>T p.H339H Chr.10: 21516806-21516806 on Build GRCh38
6212191 c.1139G>T p.S380I Chr.10: 21516684-21516684 on Build GRCh38
2132885 c.1161C>A p.S387S Chr.10: 21516662-21516662 on Build GRCh38
4841316 c.1035C>T p.H345H Chr.10: 21516788-21516788 on Build GRCh38
6324064 c.990C>T p.N330N Chr.10: 21516833-21516833 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs577009784 Chr.10: 21516820-21516820 on Build GRCh38
rs780812690 Chr.10: 21516756-21516756 on Build GRCh38
rs764002766 Chr.10: 21516769-21516792 on Build GRCh38
rs900156272 Chr.10: 21516792-21516803 on Build GRCh38
rs751132666 Chr.10: 21516738-21516738 on Build GRCh38
rs755480366 Chr.10: 21516818-21516818 on Build GRCh38
rs975370491 Chr.10: 21516680-21516680 on Build GRCh38
rs992865629 Chr.10: 21516758-21516758 on Build GRCh38
rs776570974 Chr.10: 21516649-21516649 on Build GRCh38
rs748453245 Chr.10: 21516825-21516825 on Build GRCh38
rs964269672 Chr.10: 21516669-21516669 on Build GRCh38
rs759429214 Chr.10: 21516690-21516690 on Build GRCh38
rs752476329 Chr.10: 21516719-21516719 on Build GRCh38
rs1040481054 Chr.10: 21516769-21516777 on Build GRCh38
rs765029583 Chr.10: 21516708-21516708 on Build GRCh38
rs763624692 Chr.10: 21516725-21516725 on Build GRCh38
rs908328759 Chr.10: 21516737-21516737 on Build GRCh38
rs867443373 Chr.10: 21516735-21516735 on Build GRCh38
rs1008797878 Chr.10: 21516765-21516765 on Build GRCh38
rs753515224 Chr.10: 21516792-21516800 on Build GRCh38
rs772376815 Chr.10: 21516827-21516827 on Build GRCh38
rs996209554 Chr.10: 21516802-21516802 on Build GRCh38
rs762796727 Chr.10: 21516769-21516795 on Build GRCh38
rs758137659 Chr.10: 21516722-21516722 on Build GRCh38
rs767418492 Chr.10: 21516792-21516794 on Build GRCh38
rs759594340 Chr.10: 21516727-21516738 on Build GRCh38
rs915589163 Chr.10: 21516767-21516767 on Build GRCh38
rs982516203 Chr.10: 21516736-21516736 on Build GRCh38
rs376037638 Chr.10: 21516750-21516750 on Build GRCh38
rs1027211677 Chr.10: 21516710-21516710 on Build GRCh38
rs775892481 Chr.10: 21516768-21516769 on Build GRCh38
rs765978784 Chr.10: 21516792-21516797 on Build GRCh38
rs770117846 Chr.10: 21516769-21516771 on Build GRCh38
rs761810986 Chr.10: 21516782-21516783 on Build GRCh38
rs749821145 Chr.10: 21516773-21516773 on Build GRCh38
rs749932127 Chr.10: 21516791-21516792 on Build GRCh38

View Full Product Details