Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 47090055-47090328 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C2orf61
Entrez Gene ID
285051
Gene Name
chromosome 2 open reading frame 61
Gene Aliases
-
UniGene
Hs.531575
Gene Chromosome Location
Chr.2: 47086991-47155378 on Build GRCh38
Transcripts
NM_001163561.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1021228 c.614C>T p.P205L Chr.2: 47090280-47090280 on Build GRCh38
3185814 c.600C>T p.V200V Chr.2: 47090294-47090294 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs149486583 Chr.2: 47090219-47090219 on Build GRCh38
rs565808980 Chr.2: 47090215-47090215 on Build GRCh38
rs992813270 Chr.2: 47090277-47090277 on Build GRCh38
rs569475075 Chr.2: 47090228-47090228 on Build GRCh38
rs79866388 Chr.2: 47090240-47090240 on Build GRCh38
rs995756970 Chr.2: 47090122-47090122 on Build GRCh38
rs769151469 Chr.2: 47090247-47090247 on Build GRCh38
rs372377905 Chr.2: 47090229-47090229 on Build GRCh38
rs940022667 Chr.2: 47090235-47090235 on Build GRCh38
rs763995885 Chr.2: 47090163-47090163 on Build GRCh38
rs765516152 Chr.2: 47090290-47090290 on Build GRCh38
rs1017496861 Chr.2: 47090104-47090104 on Build GRCh38
rs1002403707 Chr.2: 47090148-47090148 on Build GRCh38
rs767563593 Chr.2: 47090259-47090259 on Build GRCh38
rs979796749 Chr.2: 47090280-47090280 on Build GRCh38
rs775464907 Chr.2: 47090241-47090241 on Build GRCh38
rs541454105 Chr.2: 47090289-47090289 on Build GRCh38
rs559827743 Chr.2: 47090097-47090097 on Build GRCh38
rs184627300 Chr.2: 47090175-47090175 on Build GRCh38
rs867849619 Chr.2: 47090129-47090129 on Build GRCh38
rs993167607 Chr.2: 47090080-47090080 on Build GRCh38
rs745497155 Chr.2: 47090181-47090181 on Build GRCh38
rs921130097 Chr.2: 47090201-47090201 on Build GRCh38
rs542487053 Chr.2: 47090124-47090124 on Build GRCh38
rs988175107 Chr.2: 47090137-47090137 on Build GRCh38
rs115340807 Chr.2: 47090273-47090273 on Build GRCh38
rs541042211 Chr.2: 47090085-47090085 on Build GRCh38
rs28679288 Chr.2: 47090111-47090111 on Build GRCh38
rs77701325 Chr.2: 47090218-47090218 on Build GRCh38
rs1030161486 Chr.2: 47090120-47090120 on Build GRCh38
rs981451183 Chr.2: 47090173-47090173 on Build GRCh38
rs1031535505 Chr.2: 47090245-47090245 on Build GRCh38
rs997324418 Chr.2: 47090114-47090114 on Build GRCh38
rs531239162 Chr.2: 47090159-47090159 on Build GRCh38
rs138016561 Chr.2: 47090165-47090165 on Build GRCh38
rs35043478 Chr.2: 47090160-47090161 on Build GRCh38
rs754008121 Chr.2: 47090288-47090288 on Build GRCh38
rs189459316 Chr.2: 47090182-47090182 on Build GRCh38
rs774827446 Chr.2: 47090262-47090262 on Build GRCh38
rs774444518 Chr.2: 47090249-47090249 on Build GRCh38
rs756722532 Chr.2: 47090221-47090221 on Build GRCh38
rs374218594 Chr.2: 47090260-47090260 on Build GRCh38
rs745565246 Chr.2: 47090237-47090237 on Build GRCh38
rs887434020 Chr.2: 47090092-47090092 on Build GRCh38
rs757333101 Chr.2: 47090279-47090279 on Build GRCh38
rs558450503 Chr.2: 47090253-47090253 on Build GRCh38
rs960372088 Chr.2: 47090170-47090170 on Build GRCh38
rs547063773 Chr.2: 47090198-47090198 on Build GRCh38
rs753210831 Chr.2: 47090299-47090299 on Build GRCh38
rs942986047 Chr.2: 47090238-47090238 on Build GRCh38
rs12467763 Chr.2: 47090126-47090126 on Build GRCh38
rs770701353 Chr.2: 47090246-47090246 on Build GRCh38
rs749128889 Chr.2: 47090244-47090244 on Build GRCh38
rs1051179185 Chr.2: 47090078-47090078 on Build GRCh38
rs563824597 Chr.2: 47090157-47090157 on Build GRCh38

View Full Product Details