Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 43453547-43453808 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM267
Entrez Gene ID
64417
Gene Name
transmembrane protein 267
Gene Aliases
C5orf28
UniGene
Hs.732093
Gene Chromosome Location
Chr.5: 43444252-43484660 on Build GRCh38
Transcripts
NM_022483.4 XM_005248333.4 XM_005248335.3 XM_006714486.3 XM_011514074.2 XM_011514075.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
738717 c.209G>C p.G70A Chr.5: 43453761-43453761 on Build GRCh38
4155831 c.200T>A p.V67E Chr.5: 43453770-43453770 on Build GRCh38
5555824 c.182G>C p.G61A Chr.5: 43453788-43453788 on Build GRCh38
5900891 c.299C>T p.S100F Chr.5: 43453671-43453671 on Build GRCh38
5030308 c.312+8G>A p.? Chr.5: 43453650-43453650 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764564128 Chr.5: 43453761-43453761 on Build GRCh38
rs776722443 Chr.5: 43453718-43453718 on Build GRCh38
rs745352757 Chr.5: 43453672-43453672 on Build GRCh38
rs754613044 Chr.5: 43453774-43453774 on Build GRCh38
rs758851091 Chr.5: 43453648-43453648 on Build GRCh38
rs753068208 Chr.5: 43453748-43453748 on Build GRCh38
rs780713965 Chr.5: 43453669-43453669 on Build GRCh38
rs755293659 Chr.5: 43453611-43453611 on Build GRCh38
rs886313782 Chr.5: 43453671-43453671 on Build GRCh38
rs758945061 Chr.5: 43453758-43453758 on Build GRCh38
rs765540114 Chr.5: 43453740-43453740 on Build GRCh38
rs150067423 Chr.5: 43453690-43453690 on Build GRCh38
rs753155963 Chr.5: 43453618-43453618 on Build GRCh38
rs749993631 Chr.5: 43453771-43453771 on Build GRCh38
rs1006452354 Chr.5: 43453695-43453695 on Build GRCh38
rs772100712 Chr.5: 43453698-43453698 on Build GRCh38
rs766690171 Chr.5: 43453717-43453717 on Build GRCh38
rs759744689 Chr.5: 43453729-43453729 on Build GRCh38
rs931208291 Chr.5: 43453689-43453689 on Build GRCh38
rs1047476545 Chr.5: 43453628-43453628 on Build GRCh38
rs778610303 Chr.5: 43453778-43453778 on Build GRCh38
rs1036650852 Chr.5: 43453601-43453601 on Build GRCh38
rs779155225 Chr.5: 43453589-43453589 on Build GRCh38
rs779559729 Chr.5: 43453686-43453686 on Build GRCh38
rs201366361 Chr.5: 43453772-43453772 on Build GRCh38
rs774141821 Chr.5: 43453691-43453691 on Build GRCh38
rs201364406 Chr.5: 43453614-43453614 on Build GRCh38
rs540944830 Chr.5: 43453773-43453773 on Build GRCh38
rs537091081 Chr.5: 43453709-43453709 on Build GRCh38
rs1044472860 Chr.5: 43453579-43453579 on Build GRCh38
rs760820309 Chr.5: 43453716-43453716 on Build GRCh38
rs748120615 Chr.5: 43453697-43453697 on Build GRCh38
rs760611335 Chr.5: 43453747-43453749 on Build GRCh38
rs773471558 Chr.5: 43453706-43453706 on Build GRCh38
rs749178169 Chr.5: 43453687-43453687 on Build GRCh38
rs755813985 Chr.5: 43453674-43453674 on Build GRCh38

View Full Product Details