Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 73435112-73435375 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FZD9
Entrez Gene ID
8326
Gene Name
frizzled class receptor 9
Gene Aliases
CD349, FZD3
UniGene
Hs.647029
Gene Chromosome Location
Chr.7: 73433779-73436120 on Build GRCh38
Transcripts
NM_003508.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5749870 c.1348G>T p.V450F Chr.7: 73435355-73435355 on Build GRCh38
4461733 c.1218C>T p.L406L Chr.7: 73435225-73435225 on Build GRCh38
5407851 c.1263C>T p.F421F Chr.7: 73435270-73435270 on Build GRCh38
138619 c.1330C>T p.L444F Chr.7: 73435337-73435337 on Build GRCh38
6668232 c.1167C>T p.Y389Y Chr.7: 73435174-73435174 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782450675 Chr.7: 73435231-73435231 on Build GRCh38
rs782243057 Chr.7: 73435275-73435275 on Build GRCh38
rs782813094 Chr.7: 73435328-73435328 on Build GRCh38
rs782278676 Chr.7: 73435165-73435165 on Build GRCh38
rs114980092 Chr.7: 73435298-73435298 on Build GRCh38
rs782272876 Chr.7: 73435306-73435306 on Build GRCh38
rs782088568 Chr.7: 73435310-73435310 on Build GRCh38
rs782786443 Chr.7: 73435336-73435336 on Build GRCh38
rs201104999 Chr.7: 73435302-73435302 on Build GRCh38
rs782381628 Chr.7: 73435171-73435171 on Build GRCh38
rs782381088 Chr.7: 73435307-73435307 on Build GRCh38
rs17856756 Chr.7: 73435349-73435349 on Build GRCh38
rs61749862 Chr.7: 73435186-73435186 on Build GRCh38
rs547599913 Chr.7: 73435271-73435271 on Build GRCh38
rs370867927 Chr.7: 73435305-73435305 on Build GRCh38
rs202037150 Chr.7: 73435203-73435203 on Build GRCh38
rs115041747 Chr.7: 73435333-73435333 on Build GRCh38
rs537364650 Chr.7: 73435327-73435327 on Build GRCh38
rs199498649 Chr.7: 73435309-73435309 on Build GRCh38
rs199961288 Chr.7: 73435223-73435223 on Build GRCh38
rs782509182 Chr.7: 73435136-73435136 on Build GRCh38
rs781982258 Chr.7: 73435173-73435173 on Build GRCh38
rs201244210 Chr.7: 73435296-73435296 on Build GRCh38
rs782152078 Chr.7: 73435279-73435279 on Build GRCh38
rs781826044 Chr.7: 73435219-73435219 on Build GRCh38
rs139225216 Chr.7: 73435133-73435133 on Build GRCh38
rs781874253 Chr.7: 73435340-73435340 on Build GRCh38
rs782493264 Chr.7: 73435290-73435290 on Build GRCh38
rs782738625 Chr.7: 73435132-73435132 on Build GRCh38
rs532621192 Chr.7: 73435239-73435239 on Build GRCh38
rs782741814 Chr.7: 73435218-73435218 on Build GRCh38
rs145921800 Chr.7: 73435217-73435217 on Build GRCh38
rs782278695 Chr.7: 73435139-73435139 on Build GRCh38
rs781928707 Chr.7: 73435196-73435196 on Build GRCh38
rs781906732 Chr.7: 73435242-73435242 on Build GRCh38
rs373485212 Chr.7: 73435137-73435137 on Build GRCh38
rs782569037 Chr.7: 73435149-73435149 on Build GRCh38
rs1046610661 Chr.7: 73435147-73435147 on Build GRCh38
rs116090716 Chr.7: 73435204-73435204 on Build GRCh38
rs762475044 Chr.7: 73435162-73435162 on Build GRCh38
rs377542235 Chr.7: 73435354-73435354 on Build GRCh38
rs782534847 Chr.7: 73435264-73435264 on Build GRCh38
rs202155373 Chr.7: 73435320-73435320 on Build GRCh38
rs550169152 Chr.7: 73435141-73435141 on Build GRCh38

View Full Product Details