Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 18058726-18058999 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASAH1
Entrez Gene ID
427
Gene Name
N-acylsphingosine amidohydrolase 1
Gene Aliases
AC, ACDase, ASAH, PHP, PHP32, SMAPME
UniGene
Hs.527412
Gene Chromosome Location
Chr.8: 18056299-18084998 on Build GRCh38
Transcripts
NM_001127505.2 NM_004315.5 NM_177924.4 XM_005273504.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1699772 c.1031C>T p.S344L Chr.8: 18058884-18058884 on Build GRCh38
1699771 c.1049C>T p.S350L Chr.8: 18058884-18058884 on Build GRCh38
1699770 c.1097C>T p.S366L Chr.8: 18058884-18058884 on Build GRCh38
6376809 c.1146G>T p.K382N Chr.8: 18058835-18058835 on Build GRCh38
5601050 c.1095C>T p.I365I Chr.8: 18058886-18058886 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745748620 Chr.8: 18058798-18058798 on Build GRCh38
rs997097428 Chr.8: 18058960-18058960 on Build GRCh38
rs776667602 Chr.8: 18058806-18058806 on Build GRCh38
rs761250571 Chr.8: 18058946-18058946 on Build GRCh38
rs367837859 Chr.8: 18058909-18058909 on Build GRCh38
rs762466247 Chr.8: 18058894-18058895 on Build GRCh38
rs764840305 Chr.8: 18058930-18058930 on Build GRCh38
rs761522522 Chr.8: 18058921-18058921 on Build GRCh38
rs768114708 Chr.8: 18058895-18058896 on Build GRCh38
rs777271052 Chr.8: 18058785-18058785 on Build GRCh38
rs757058563 Chr.8: 18058857-18058857 on Build GRCh38
rs768969765 Chr.8: 18058800-18058800 on Build GRCh38
rs763842677 Chr.8: 18058834-18058834 on Build GRCh38
rs964224037 Chr.8: 18058839-18058839 on Build GRCh38
rs758370923 Chr.8: 18058862-18058862 on Build GRCh38
rs763456028 Chr.8: 18058915-18058915 on Build GRCh38
rs573339747 Chr.8: 18058782-18058782 on Build GRCh38
rs370218098 Chr.8: 18058931-18058931 on Build GRCh38
rs750855024 Chr.8: 18058928-18058929 on Build GRCh38
rs969999614 Chr.8: 18058793-18058793 on Build GRCh38
rs757009719 Chr.8: 18058797-18058797 on Build GRCh38
rs763391097 Chr.8: 18058900-18058900 on Build GRCh38
rs766965374 Chr.8: 18058819-18058819 on Build GRCh38
rs901535648 Chr.8: 18058945-18058945 on Build GRCh38
rs913948326 Chr.8: 18058894-18058894 on Build GRCh38
rs1056111066 Chr.8: 18058752-18058752 on Build GRCh38
rs747134165 Chr.8: 18058873-18058873 on Build GRCh38
rs755833109 Chr.8: 18058784-18058784 on Build GRCh38
rs773907933 Chr.8: 18058888-18058889 on Build GRCh38
rs748318714 Chr.8: 18058892-18058892 on Build GRCh38
rs190476632 Chr.8: 18058816-18058816 on Build GRCh38
rs773776313 Chr.8: 18058899-18058899 on Build GRCh38
rs139709919 Chr.8: 18058827-18058827 on Build GRCh38
rs778740336 Chr.8: 18058859-18058859 on Build GRCh38
rs185572418 Chr.8: 18058761-18058761 on Build GRCh38
rs564936875 Chr.8: 18058936-18058936 on Build GRCh38
rs567086714 Chr.8: 18058879-18058879 on Build GRCh38
rs755533709 Chr.8: 18058828-18058828 on Build GRCh38
rs771591421 Chr.8: 18058901-18058901 on Build GRCh38
rs371520705 Chr.8: 18058854-18058854 on Build GRCh38
rs113778837 Chr.8: 18058799-18058799 on Build GRCh38
rs746637157 Chr.8: 18058756-18058756 on Build GRCh38
rs781477468 Chr.8: 18058890-18058890 on Build GRCh38
rs779831936 Chr.8: 18058872-18058872 on Build GRCh38
rs770125895 Chr.8: 18058895-18058895 on Build GRCh38
rs745659071 Chr.8: 18058861-18058861 on Build GRCh38
rs535825543 Chr.8: 18058794-18058794 on Build GRCh38
rs374547906 Chr.8: 18058908-18058908 on Build GRCh38
rs534249736 Chr.8: 18058760-18058760 on Build GRCh38
rs376635519 Chr.8: 18058817-18058817 on Build GRCh38
rs1055760842 Chr.8: 18058944-18058944 on Build GRCh38
rs867544027 Chr.8: 18058765-18058765 on Build GRCh38
rs143850063 Chr.8: 18058883-18058883 on Build GRCh38

View Full Product Details