Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 57341202-57341463 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAAH2
Entrez Gene ID
158584
Gene Name
fatty acid amide hydrolase 2
Gene Aliases
AMDD
UniGene
Hs.496205
Gene Chromosome Location
Chr.X: 57121861-57489196 on Build GRCh38
Transcripts
NM_174912.3 XM_005261982.1 XM_005261985.2 XM_005261986.3 XM_005261987.3 XM_011530767.2 XM_017029290.1 XM_017029291.1 XM_017029292.1 XM_017029293.1 XM_017029294.1

SNP Detail

SNP ID Chromosome Location
rs774654577 Chr.X: 57341358-57341358 on Build GRCh38
rs201267649 Chr.X: 57341286-57341286 on Build GRCh38
rs766737067 Chr.X: 57341372-57341372 on Build GRCh38
rs755479954 Chr.X: 57341295-57341295 on Build GRCh38
rs769621605 Chr.X: 57341316-57341316 on Build GRCh38
rs370657252 Chr.X: 57341241-57341241 on Build GRCh38
rs761970184 Chr.X: 57341278-57341278 on Build GRCh38
rs902863297 Chr.X: 57341297-57341297 on Build GRCh38
rs910608848 Chr.X: 57341412-57341412 on Build GRCh38
rs762858325 Chr.X: 57341268-57341268 on Build GRCh38
rs935963549 Chr.X: 57341373-57341373 on Build GRCh38
rs779764694 Chr.X: 57341338-57341338 on Build GRCh38
rs745676551 Chr.X: 57341255-57341255 on Build GRCh38
rs952507338 Chr.X: 57341381-57341381 on Build GRCh38
rs983301495 Chr.X: 57341401-57341401 on Build GRCh38
rs770739388 Chr.X: 57341249-57341249 on Build GRCh38
rs775496620 Chr.X: 57341264-57341264 on Build GRCh38
rs373173279 Chr.X: 57341251-57341251 on Build GRCh38
rs746643188 Chr.X: 57341247-57341247 on Build GRCh38
rs760361862 Chr.X: 57341261-57341261 on Build GRCh38
rs958579959 Chr.X: 57341337-57341337 on Build GRCh38
rs374743678 Chr.X: 57341304-57341304 on Build GRCh38
rs371507715 Chr.X: 57341273-57341273 on Build GRCh38
rs773407850 Chr.X: 57341365-57341365 on Build GRCh38
rs1050950365 Chr.X: 57341391-57341391 on Build GRCh38
rs767735654 Chr.X: 57341283-57341283 on Build GRCh38
rs757910652 Chr.X: 57341243-57341243 on Build GRCh38
rs777298575 Chr.X: 57341301-57341301 on Build GRCh38
rs370316835 Chr.X: 57341403-57341403 on Build GRCh38
rs376344650 Chr.X: 57341259-57341259 on Build GRCh38
rs754187037 Chr.X: 57341379-57341379 on Build GRCh38
rs754345982 Chr.X: 57341291-57341291 on Build GRCh38
rs140554148 Chr.X: 57341311-57341311 on Build GRCh38
rs774427834 Chr.X: 57341350-57341350 on Build GRCh38
rs756826787 Chr.X: 57341402-57341402 on Build GRCh38
rs958802646 Chr.X: 57341240-57341240 on Build GRCh38
rs760933935 Chr.X: 57341371-57341371 on Build GRCh38
rs765952886 Chr.X: 57341363-57341363 on Build GRCh38
rs768747063 Chr.X: 57341269-57341269 on Build GRCh38
rs777199360 Chr.X: 57341246-57341246 on Build GRCh38
rs749206936 Chr.X: 57341343-57341343 on Build GRCh38
rs530184486 Chr.X: 57341396-57341396 on Build GRCh38
rs765675475 Chr.X: 57341392-57341392 on Build GRCh38
rs945997269 Chr.X: 57341329-57341329 on Build GRCh38
rs761882094 Chr.X: 57341356-57341356 on Build GRCh38
rs144496006 Chr.X: 57341364-57341364 on Build GRCh38
rs367663989 Chr.X: 57341387-57341387 on Build GRCh38
rs768497842 Chr.X: 57341347-57341347 on Build GRCh38
rs766710234 Chr.X: 57341290-57341290 on Build GRCh38
rs771442773 Chr.X: 57341313-57341313 on Build GRCh38

View Full Product Details