Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 24826913-24827186 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLIC4
Entrez Gene ID
25932
Gene Name
chloride intracellular channel 4
Gene Aliases
CLIC4L, H1, MTCLIC, huH1, p64H1
UniGene
Hs.440544
Gene Chromosome Location
Chr.1: 24745269-24844324 on Build GRCh38
Transcripts
NM_013943.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3977530 c.309-10T>C p.? Chr.1: 24827000-24827000 on Build GRCh38
907453 c.375C>A p.F125L Chr.1: 24827076-24827076 on Build GRCh38
399044 c.395C>G p.S132* Chr.1: 24827096-24827096 on Build GRCh38
6389395 c.355A>G p.M119V Chr.1: 24827056-24827056 on Build GRCh38
5499971 c.380C>T p.A127V Chr.1: 24827081-24827081 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs563875078 Chr.1: 24826949-24826949 on Build GRCh38
rs199769441 Chr.1: 24827138-24827138 on Build GRCh38
rs761846011 Chr.1: 24827039-24827039 on Build GRCh38
rs755279429 Chr.1: 24827080-24827080 on Build GRCh38
rs183272052 Chr.1: 24827157-24827157 on Build GRCh38
rs763150022 Chr.1: 24826994-24826994 on Build GRCh38
rs577806553 Chr.1: 24826988-24826989 on Build GRCh38
rs769731551 Chr.1: 24826988-24826988 on Build GRCh38
rs4648877 Chr.1: 24827156-24827156 on Build GRCh38
rs756703083 Chr.1: 24827124-24827124 on Build GRCh38
rs200646277 Chr.1: 24826996-24826996 on Build GRCh38
rs531189541 Chr.1: 24826990-24826990 on Build GRCh38
rs764645616 Chr.1: 24827040-24827040 on Build GRCh38
rs955688622 Chr.1: 24827125-24827125 on Build GRCh38
rs769177029 Chr.1: 24826965-24826965 on Build GRCh38
rs760420796 Chr.1: 24827010-24827010 on Build GRCh38
rs763971573 Chr.1: 24827020-24827020 on Build GRCh38
rs764089312 Chr.1: 24826991-24826991 on Build GRCh38
rs750008934 Chr.1: 24827045-24827045 on Build GRCh38
rs1030035223 Chr.1: 24827120-24827120 on Build GRCh38
rs781653580 Chr.1: 24827082-24827082 on Build GRCh38
rs763354050 Chr.1: 24826985-24826985 on Build GRCh38
rs377354681 Chr.1: 24826993-24826993 on Build GRCh38
rs753696941 Chr.1: 24827029-24827029 on Build GRCh38
rs751282908 Chr.1: 24827056-24827056 on Build GRCh38
rs973859549 Chr.1: 24827102-24827102 on Build GRCh38
rs373018745 Chr.1: 24826959-24826959 on Build GRCh38
rs747317080 Chr.1: 24826963-24826963 on Build GRCh38
rs748598324 Chr.1: 24827098-24827098 on Build GRCh38
rs751597017 Chr.1: 24827130-24827130 on Build GRCh38
rs779816492 Chr.1: 24827046-24827046 on Build GRCh38
rs748742408 Chr.1: 24826983-24826983 on Build GRCh38
rs781506958 Chr.1: 24826977-24826977 on Build GRCh38
rs749301691 Chr.1: 24827151-24827151 on Build GRCh38
rs774559398 Chr.1: 24827004-24827004 on Build GRCh38

View Full Product Details