Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 207091585-207091850 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C4BPB
Entrez Gene ID
725
Gene Name
complement component 4 binding protein beta
Gene Aliases
C4BP
UniGene
Hs.99886
Gene Chromosome Location
Chr.1: 207088826-207099993 on Build GRCh38
Transcripts
NM_000716.3 NM_001017364.1 NM_001017365.2 NM_001017366.2 NM_001017367.1 XM_005273254.4 XM_005273255.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4643641 c.255G>A p.G85G Chr.1: 207091696-207091696 on Build GRCh38
6435269 c.244G>A p.D82N Chr.1: 207091658-207091658 on Build GRCh38
7251450 c.217G>T p.D73Y Chr.1: 207091658-207091658 on Build GRCh38
4230261 c.314G>A p.G105D Chr.1: 207091755-207091755 on Build GRCh38
7169929 c.248C>T p.S83F Chr.1: 207091689-207091689 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754180318 Chr.1: 207091621-207091621 on Build GRCh38
rs767670680 Chr.1: 207091706-207091706 on Build GRCh38
rs1031404545 Chr.1: 207091795-207091795 on Build GRCh38
rs150433138 Chr.1: 207091651-207091651 on Build GRCh38
rs139041322 Chr.1: 207091720-207091720 on Build GRCh38
rs142909654 Chr.1: 207091763-207091763 on Build GRCh38
rs777244001 Chr.1: 207091756-207091756 on Build GRCh38
rs758087191 Chr.1: 207091751-207091751 on Build GRCh38
rs747812176 Chr.1: 207091635-207091635 on Build GRCh38
rs777331831 Chr.1: 207091643-207091643 on Build GRCh38
rs770951563 Chr.1: 207091663-207091663 on Build GRCh38
rs201260667 Chr.1: 207091731-207091731 on Build GRCh38
rs144805017 Chr.1: 207091723-207091723 on Build GRCh38
rs761658324 Chr.1: 207091761-207091761 on Build GRCh38
rs199537875 Chr.1: 207091714-207091714 on Build GRCh38
rs769720121 Chr.1: 207091678-207091678 on Build GRCh38
rs757890802 Chr.1: 207091638-207091638 on Build GRCh38
rs548763600 Chr.1: 207091624-207091624 on Build GRCh38
rs890008916 Chr.1: 207091700-207091700 on Build GRCh38
rs775461181 Chr.1: 207091786-207091786 on Build GRCh38
rs778470528 Chr.1: 207091630-207091630 on Build GRCh38
rs779009976 Chr.1: 207091769-207091769 on Build GRCh38
rs754303721 Chr.1: 207091722-207091722 on Build GRCh38
rs369264157 Chr.1: 207091789-207091789 on Build GRCh38
rs762188342 Chr.1: 207091697-207091697 on Build GRCh38
rs966570922 Chr.1: 207091801-207091801 on Build GRCh38
rs776559668 Chr.1: 207091759-207091760 on Build GRCh38
rs56258224 Chr.1: 207091715-207091715 on Build GRCh38
rs752343117 Chr.1: 207091734-207091734 on Build GRCh38
rs774661287 Chr.1: 207091682-207091682 on Build GRCh38
rs376995946 Chr.1: 207091674-207091674 on Build GRCh38
rs191950896 Chr.1: 207091671-207091671 on Build GRCh38
rs756682463 Chr.1: 207091606-207091606 on Build GRCh38
rs766628247 Chr.1: 207091609-207091609 on Build GRCh38
rs907003573 Chr.1: 207091677-207091677 on Build GRCh38
rs746891305 Chr.1: 207091645-207091645 on Build GRCh38
rs1019117802 Chr.1: 207091799-207091799 on Build GRCh38
rs1028605139 Chr.1: 207091627-207091627 on Build GRCh38
rs773386935 Chr.1: 207091709-207091709 on Build GRCh38
rs772368503 Chr.1: 207091821-207091821 on Build GRCh38
rs769703116 Chr.1: 207091764-207091764 on Build GRCh38

View Full Product Details