Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 112080336-112080605 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C11orf57
Entrez Gene ID
55216
Gene Name
chromosome 11 open reading frame 57
Gene Aliases
-
UniGene
Hs.195060
Gene Chromosome Location
Chr.11: 112074244-112085150 on Build GRCh38
Transcripts
NM_001082969.1 NM_001082970.1 NM_001301017.1 NM_001301019.1 NM_001301021.1 NM_018195.3 NR_103469.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5625456 c.267G>C p.K89N Chr.11: 112080505-112080505 on Build GRCh38
5475464 c.179G>A p.R60H Chr.11: 112080417-112080417 on Build GRCh38
1627778 c.273A>G p.K91K Chr.11: 112080511-112080511 on Build GRCh38
1297658 c.263T>C p.L88P Chr.11: 112080501-112080501 on Build GRCh38
2102739 c.178C>T p.R60C Chr.11: 112080416-112080416 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs146242663 Chr.11: 112080417-112080417 on Build GRCh38
rs369214550 Chr.11: 112080416-112080416 on Build GRCh38
rs565496995 Chr.11: 112080448-112080448 on Build GRCh38
rs758533601 Chr.11: 112080554-112080554 on Build GRCh38
rs769300528 Chr.11: 112080383-112080383 on Build GRCh38
rs752439496 Chr.11: 112080380-112080380 on Build GRCh38
rs867001396 Chr.11: 112080479-112080479 on Build GRCh38
rs377186980 Chr.11: 112080508-112080508 on Build GRCh38
rs777639229 Chr.11: 112080458-112080458 on Build GRCh38
rs906779649 Chr.11: 112080428-112080428 on Build GRCh38
rs535451040 Chr.11: 112080520-112080520 on Build GRCh38
rs767304601 Chr.11: 112080379-112080379 on Build GRCh38
rs754100691 Chr.11: 112080537-112080537 on Build GRCh38
rs141894421 Chr.11: 112080411-112080411 on Build GRCh38
rs761871136 Chr.11: 112080543-112080543 on Build GRCh38
rs747431576 Chr.11: 112080445-112080445 on Build GRCh38
rs937131700 Chr.11: 112080553-112080553 on Build GRCh38
rs181464331 Chr.11: 112080480-112080480 on Build GRCh38
rs780591905 Chr.11: 112080438-112080438 on Build GRCh38
rs780751181 Chr.11: 112080578-112080578 on Build GRCh38
rs763862584 Chr.11: 112080531-112080531 on Build GRCh38
rs372455783 Chr.11: 112080476-112080476 on Build GRCh38
rs754001029 Chr.11: 112080410-112080410 on Build GRCh38
rs1022502913 Chr.11: 112080548-112080548 on Build GRCh38
rs371358736 Chr.11: 112080491-112080491 on Build GRCh38
rs200842933 Chr.11: 112080406-112080406 on Build GRCh38
rs778798368 Chr.11: 112080412-112080412 on Build GRCh38
rs770491300 Chr.11: 112080469-112080469 on Build GRCh38
rs139311432 Chr.11: 112080547-112080547 on Build GRCh38
rs748880447 Chr.11: 112080459-112080459 on Build GRCh38
rs765385923 Chr.11: 112080545-112080545 on Build GRCh38
rs201646193 Chr.11: 112080574-112080574 on Build GRCh38
rs267602692 Chr.11: 112080500-112080500 on Build GRCh38
rs376878376 Chr.11: 112080365-112080365 on Build GRCh38
rs756156654 Chr.11: 112080485-112080485 on Build GRCh38
rs533660532 Chr.11: 112080366-112080366 on Build GRCh38
rs755881792 Chr.11: 112080397-112080397 on Build GRCh38

View Full Product Details