Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 63150241-63150502 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AVPR1A
Entrez Gene ID
552
Gene Name
arginine vasopressin receptor 1A
Gene Aliases
AVPR V1a, AVPR1, V1aR
UniGene
Hs.2131
Gene Chromosome Location
Chr.12: 63142759-63152810 on Build GRCh38
Transcripts
NM_000706.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1283606 c.509T>C p.M170T Chr.12: 63150328-63150328 on Build GRCh38
942399 c.515C>T p.A172V Chr.12: 63150322-63150322 on Build GRCh38
3955051 c.513C>A p.I171I Chr.12: 63150324-63150324 on Build GRCh38
1990764 c.503G>A p.R168H Chr.12: 63150334-63150334 on Build GRCh38
4664792 c.454G>T p.A152S Chr.12: 63150383-63150383 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs143160244 Chr.12: 63150282-63150282 on Build GRCh38
rs530628313 Chr.12: 63150297-63150297 on Build GRCh38
rs768124807 Chr.12: 63150396-63150396 on Build GRCh38
rs138241660 Chr.12: 63150369-63150369 on Build GRCh38
rs181053969 Chr.12: 63150267-63150267 on Build GRCh38
rs369032173 Chr.12: 63150363-63150363 on Build GRCh38
rs773269527 Chr.12: 63150370-63150370 on Build GRCh38
rs778650138 Chr.12: 63150341-63150341 on Build GRCh38
rs763936762 Chr.12: 63150307-63150307 on Build GRCh38
rs1042615 Chr.12: 63150429-63150429 on Build GRCh38
rs762698235 Chr.12: 63150444-63150444 on Build GRCh38
rs753214363 Chr.12: 63150474-63150474 on Build GRCh38
rs563622430 Chr.12: 63150450-63150450 on Build GRCh38
rs769533111 Chr.12: 63150437-63150437 on Build GRCh38
rs375549087 Chr.12: 63150410-63150410 on Build GRCh38
rs914049115 Chr.12: 63150344-63150344 on Build GRCh38
rs745458336 Chr.12: 63150418-63150418 on Build GRCh38
rs767709671 Chr.12: 63150470-63150470 on Build GRCh38
rs756603967 Chr.12: 63150340-63150340 on Build GRCh38
rs748117343 Chr.12: 63150434-63150434 on Build GRCh38
rs945631623 Chr.12: 63150345-63150345 on Build GRCh38
rs180760072 Chr.12: 63150439-63150439 on Build GRCh38
rs761284955 Chr.12: 63150477-63150477 on Build GRCh38
rs537394626 Chr.12: 63150318-63150318 on Build GRCh38
rs776384659 Chr.12: 63150265-63150265 on Build GRCh38
rs775085135 Chr.12: 63150351-63150351 on Build GRCh38
rs776488571 Chr.12: 63150430-63150430 on Build GRCh38
rs899668332 Chr.12: 63150347-63150347 on Build GRCh38
rs761542657 Chr.12: 63150366-63150366 on Build GRCh38
rs759630291 Chr.12: 63150452-63150452 on Build GRCh38
rs753269775 Chr.12: 63150399-63150399 on Build GRCh38
rs369710823 Chr.12: 63150403-63150403 on Build GRCh38
rs758345632 Chr.12: 63150290-63150290 on Build GRCh38
rs751963924 Chr.12: 63150300-63150300 on Build GRCh38
rs763262198 Chr.12: 63150374-63150374 on Build GRCh38
rs745537191 Chr.12: 63150348-63150348 on Build GRCh38
rs748519112 Chr.12: 63150335-63150335 on Build GRCh38
rs377107751 Chr.12: 63150380-63150380 on Build GRCh38
rs779608829 Chr.12: 63150422-63150422 on Build GRCh38
rs750338761 Chr.12: 63150356-63150357 on Build GRCh38
rs976250964 Chr.12: 63150315-63150315 on Build GRCh38
rs758045345 Chr.12: 63150420-63150420 on Build GRCh38
rs1037287168 Chr.12: 63150388-63150388 on Build GRCh38
rs925524625 Chr.12: 63150352-63150352 on Build GRCh38
rs1020309705 Chr.12: 63150416-63150416 on Build GRCh38
rs148278650 Chr.12: 63150285-63150285 on Build GRCh38
rs773038860 Chr.12: 63150438-63150438 on Build GRCh38
rs755081148 Chr.12: 63150390-63150390 on Build GRCh38
rs146296879 Chr.12: 63150359-63150359 on Build GRCh38
rs553995625 Chr.12: 63150381-63150381 on Build GRCh38
rs771180379 Chr.12: 63150446-63150446 on Build GRCh38
rs2228155 Chr.12: 63150354-63150354 on Build GRCh38
rs778311693 Chr.12: 63150286-63150286 on Build GRCh38
rs781774666 Chr.12: 63150334-63150334 on Build GRCh38

View Full Product Details