Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 73926700-73926973 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DGUOK
Entrez Gene ID
1716
Gene Name
deoxyguanosine kinase
Gene Aliases
MTDPS3, dGK
UniGene
Hs.469022
Gene Chromosome Location
Chr.2: 73926826-73958961 on Build GRCh38
Transcripts
NM_001318859.1 NM_001318860.1 NM_001318861.1 NM_001318862.1 NM_001318863.1 NM_080916.2 NM_080918.2 NR_134893.1 NR_134894.1 NR_134895.1 NR_134896.1 NR_134897.1 NR_134898.1 XM_011532647.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4095694 c.8C>T p.A3V Chr.2: 73926918-73926918 on Build GRCh38
4981190 c.9G>T p.A3A Chr.2: 73926919-73926919 on Build GRCh38
443203 c.34C>T p.R12* Chr.2: 73926944-73926944 on Build GRCh38
6643223 c.49T>C p.S17P Chr.2: 73926959-73926959 on Build GRCh38
122566 c.29G>T p.R10L Chr.2: 73926939-73926939 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775590584 Chr.2: 73926898-73926898 on Build GRCh38
rs750288928 Chr.2: 73926864-73926864 on Build GRCh38
rs10186730 Chr.2: 73926863-73926863 on Build GRCh38
rs778424567 Chr.2: 73926882-73926882 on Build GRCh38
rs756028511 Chr.2: 73926906-73926906 on Build GRCh38
rs543172600 Chr.2: 73926823-73926823 on Build GRCh38
rs773299715 Chr.2: 73926951-73926951 on Build GRCh38
rs771281628 Chr.2: 73926891-73926891 on Build GRCh38
rs757178274 Chr.2: 73926860-73926860 on Build GRCh38
rs569516260 Chr.2: 73926776-73926776 on Build GRCh38
rs372855715 Chr.2: 73926927-73926927 on Build GRCh38
rs549296132 Chr.2: 73926807-73926807 on Build GRCh38
rs140284468 Chr.2: 73926932-73926932 on Build GRCh38
rs192086507 Chr.2: 73926768-73926768 on Build GRCh38
rs775406988 Chr.2: 73926923-73926924 on Build GRCh38
rs781275867 Chr.2: 73926918-73926918 on Build GRCh38
rs747744094 Chr.2: 73926911-73926911 on Build GRCh38
rs766221659 Chr.2: 73926908-73926908 on Build GRCh38
rs763189469 Chr.2: 73926930-73926930 on Build GRCh38
rs567251538 Chr.2: 73926849-73926850 on Build GRCh38
rs779916809 Chr.2: 73926876-73926876 on Build GRCh38
rs565324493 Chr.2: 73926839-73926839 on Build GRCh38
rs1035747996 Chr.2: 73926821-73926821 on Build GRCh38
rs775274523 Chr.2: 73926924-73926924 on Build GRCh38
rs369681767 Chr.2: 73926903-73926903 on Build GRCh38
rs1050641849 Chr.2: 73926727-73926727 on Build GRCh38
rs548935596 Chr.2: 73926829-73926829 on Build GRCh38
rs1039544047 Chr.2: 73926813-73926813 on Build GRCh38
rs576435998 Chr.2: 73926833-73926833 on Build GRCh38
rs765263039 Chr.2: 73926895-73926895 on Build GRCh38
rs923251700 Chr.2: 73926855-73926855 on Build GRCh38
rs561247326 Chr.2: 73926824-73926824 on Build GRCh38
rs1026158259 Chr.2: 73926866-73926866 on Build GRCh38
rs886056320 Chr.2: 73926854-73926855 on Build GRCh38
rs779947736 Chr.2: 73926844-73926844 on Build GRCh38
rs746100266 Chr.2: 73926921-73926921 on Build GRCh38
rs112541347 Chr.2: 73926850-73926850 on Build GRCh38
rs951322194 Chr.2: 73926879-73926879 on Build GRCh38
rs534297082 Chr.2: 73926913-73926913 on Build GRCh38
rs756952542 Chr.2: 73926849-73926849 on Build GRCh38
rs767017289 Chr.2: 73926873-73926887 on Build GRCh38
rs201025800 Chr.2: 73926841-73926841 on Build GRCh38
rs772519303 Chr.2: 73926886-73926886 on Build GRCh38
rs889273205 Chr.2: 73926758-73926758 on Build GRCh38
rs572587324 Chr.2: 73926822-73926822 on Build GRCh38
rs831526 Chr.2: 73926778-73926778 on Build GRCh38
rs778346110 Chr.2: 73926862-73926876 on Build GRCh38
rs759734588 Chr.2: 73926894-73926894 on Build GRCh38
rs952486466 Chr.2: 73926835-73926835 on Build GRCh38
rs753666732 Chr.2: 73926857-73926857 on Build GRCh38
rs374309908 Chr.2: 73926888-73926888 on Build GRCh38
rs866656831 Chr.2: 73926889-73926889 on Build GRCh38
rs922450558 Chr.2: 73926818-73926818 on Build GRCh38
rs753441865 Chr.2: 73926909-73926909 on Build GRCh38
rs757874547 Chr.2: 73926912-73926912 on Build GRCh38
rs894051779 Chr.2: 73926845-73926845 on Build GRCh38
rs903470517 Chr.2: 73926897-73926897 on Build GRCh38
rs977157089 Chr.2: 73926923-73926923 on Build GRCh38
rs748637196 Chr.2: 73926885-73926885 on Build GRCh38
rs754623273 Chr.2: 73926910-73926910 on Build GRCh38
rs759288670 Chr.2: 73926861-73926862 on Build GRCh38
rs754462751 Chr.2: 73926878-73926878 on Build GRCh38
rs1054034625 Chr.2: 73926928-73926928 on Build GRCh38
rs751001163 Chr.2: 73926777-73926777 on Build GRCh38
rs747033868 Chr.2: 73926933-73926933 on Build GRCh38
rs543924291 Chr.2: 73926834-73926834 on Build GRCh38
rs770100935 Chr.2: 73926926-73926926 on Build GRCh38
rs902813530 Chr.2: 73926825-73926825 on Build GRCh38
rs942633022 Chr.2: 73926782-73926782 on Build GRCh38
rs763099320 Chr.2: 73926900-73926900 on Build GRCh38
rs551732820 Chr.2: 73926893-73926893 on Build GRCh38
rs755938949 Chr.2: 73926865-73926865 on Build GRCh38
rs147551003 Chr.2: 73926914-73926914 on Build GRCh38
rs200333852 Chr.2: 73926890-73926890 on Build GRCh38
rs998455696 Chr.2: 73926779-73926779 on Build GRCh38
rs748960762 Chr.2: 73926877-73926877 on Build GRCh38
rs768840084 Chr.2: 73926937-73926937 on Build GRCh38
rs369813011 Chr.2: 73926859-73926859 on Build GRCh38

View Full Product Details