Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 150238625-150238880 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAMK2A
Entrez Gene ID
815
Gene Name
calcium/calmodulin dependent protein kinase II alpha
Gene Aliases
CAMKA
UniGene
Hs.743976
Gene Chromosome Location
Chr.5: 150219491-150289840 on Build GRCh38
Transcripts
NM_015981.3 NM_171825.2 XM_017009898.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5807190 c.1066+4G>T p.? Chr.5: 150238696-150238696 on Build GRCh38
7130996 c.1018-1G>T p.? Chr.5: 150238749-150238749 on Build GRCh38
3919361 c.1066G>A p.V356M Chr.5: 150238700-150238700 on Build GRCh38
7167858 c.1025C>T p.S342L Chr.5: 150238741-150238741 on Build GRCh38
5769338 c.1066+1G>T p.? Chr.5: 150238699-150238699 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs535223578 Chr.5: 150238678-150238678 on Build GRCh38
rs750791622 Chr.5: 150238792-150238792 on Build GRCh38
rs1003062996 Chr.5: 150238754-150238754 on Build GRCh38
rs369600793 Chr.5: 150238690-150238690 on Build GRCh38
rs968973603 Chr.5: 150238691-150238691 on Build GRCh38
rs560646214 Chr.5: 150238824-150238824 on Build GRCh38
rs751636760 Chr.5: 150238689-150238689 on Build GRCh38
rs1038754291 Chr.5: 150238663-150238663 on Build GRCh38
rs894056852 Chr.5: 150238847-150238847 on Build GRCh38
rs900231375 Chr.5: 150238861-150238861 on Build GRCh38
rs770841322 Chr.5: 150238737-150238737 on Build GRCh38
rs371219168 Chr.5: 150238774-150238774 on Build GRCh38
rs17111082 Chr.5: 150238829-150238829 on Build GRCh38
rs767052088 Chr.5: 150238652-150238652 on Build GRCh38
rs754671966 Chr.5: 150238719-150238719 on Build GRCh38
rs762363039 Chr.5: 150238782-150238782 on Build GRCh38
rs755124930 Chr.5: 150238794-150238794 on Build GRCh38
rs373289094 Chr.5: 150238759-150238759 on Build GRCh38
rs757622736 Chr.5: 150238677-150238677 on Build GRCh38
rs911846931 Chr.5: 150238830-150238830 on Build GRCh38
rs766528143 Chr.5: 150238793-150238793 on Build GRCh38
rs983338150 Chr.5: 150238770-150238770 on Build GRCh38
rs1038869552 Chr.5: 150238860-150238860 on Build GRCh38
rs780147512 Chr.5: 150238684-150238684 on Build GRCh38
rs1024265518 Chr.5: 150238706-150238706 on Build GRCh38
rs999120588 Chr.5: 150238705-150238705 on Build GRCh38
rs1053922432 Chr.5: 150238833-150238833 on Build GRCh38
rs548464639 Chr.5: 150238778-150238778 on Build GRCh38
rs575839300 Chr.5: 150238831-150238831 on Build GRCh38
rs1008561129 Chr.5: 150238647-150238647 on Build GRCh38
rs1047639541 Chr.5: 150238822-150238822 on Build GRCh38
rs774904052 Chr.5: 150238657-150238657 on Build GRCh38
rs929207080 Chr.5: 150238821-150238821 on Build GRCh38
rs557163766 Chr.5: 150238826-150238826 on Build GRCh38
rs373698136 Chr.5: 150238760-150238760 on Build GRCh38
rs765581640 Chr.5: 150238791-150238791 on Build GRCh38
rs372970789 Chr.5: 150238672-150238672 on Build GRCh38
rs777135245 Chr.5: 150238779-150238779 on Build GRCh38
rs111673898 Chr.5: 150238751-150238751 on Build GRCh38
rs909135950 Chr.5: 150238788-150238788 on Build GRCh38
rs778642886 Chr.5: 150238757-150238757 on Build GRCh38
rs1052298552 Chr.5: 150238817-150238817 on Build GRCh38
rs530438339 Chr.5: 150238674-150238674 on Build GRCh38
rs960193459 Chr.5: 150238725-150238725 on Build GRCh38
rs17111081 Chr.5: 150238673-150238673 on Build GRCh38
rs758613373 Chr.5: 150238680-150238680 on Build GRCh38
rs367609826 Chr.5: 150238796-150238796 on Build GRCh38

View Full Product Details