Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 15011186-15011441 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC105
Entrez Gene ID
126402
Gene Name
coiled-coil domain containing 105
Gene Aliases
-
UniGene
Hs.375985
Gene Chromosome Location
Chr.19: 15010727-15023271 on Build GRCh38
Transcripts
NM_173482.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3282752 c.544C>T p.R182W Chr.19: 15011369-15011369 on Build GRCh38
6053603 c.473G>A p.R158H Chr.19: 15011298-15011298 on Build GRCh38
7271270 c.472C>T p.R158C Chr.19: 15011297-15011297 on Build GRCh38
6857595 c.449G>A p.R150Q Chr.19: 15011274-15011274 on Build GRCh38
5343219 c.420C>T p.L140L Chr.19: 15011245-15011245 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs867429438 Chr.19: 15011285-15011285 on Build GRCh38
rs747286248 Chr.19: 15011227-15011227 on Build GRCh38
rs368250990 Chr.19: 15011203-15011203 on Build GRCh38
rs28705326 Chr.19: 15011243-15011243 on Build GRCh38
rs931273936 Chr.19: 15011242-15011242 on Build GRCh38
rs565114253 Chr.19: 15011290-15011290 on Build GRCh38
rs199904661 Chr.19: 15011292-15011292 on Build GRCh38
rs758335595 Chr.19: 15011339-15011339 on Build GRCh38
rs777879899 Chr.19: 15011217-15011217 on Build GRCh38
rs774672598 Chr.19: 15011394-15011394 on Build GRCh38
rs35048025 Chr.19: 15011242-15011242 on Build GRCh38
rs775883563 Chr.19: 15011246-15011246 on Build GRCh38
rs749805061 Chr.19: 15011410-15011410 on Build GRCh38
rs537607141 Chr.19: 15011269-15011269 on Build GRCh38
rs893421034 Chr.19: 15011274-15011274 on Build GRCh38
rs747083884 Chr.19: 15011368-15011368 on Build GRCh38
rs769188507 Chr.19: 15011411-15011411 on Build GRCh38
rs199536505 Chr.19: 15011252-15011252 on Build GRCh38
rs1036592507 Chr.19: 15011330-15011330 on Build GRCh38
rs758442019 Chr.19: 15011213-15011213 on Build GRCh38
rs543333642 Chr.19: 15011273-15011273 on Build GRCh38
rs934874525 Chr.19: 15011261-15011261 on Build GRCh38
rs770086115 Chr.19: 15011406-15011406 on Build GRCh38
rs775891331 Chr.19: 15011407-15011407 on Build GRCh38
rs867336188 Chr.19: 15011331-15011331 on Build GRCh38
rs781218855 Chr.19: 15011335-15011335 on Build GRCh38
rs888793942 Chr.19: 15011413-15011413 on Build GRCh38
rs753433500 Chr.19: 15011280-15011280 on Build GRCh38
rs1039224521 Chr.19: 15011314-15011314 on Build GRCh38
rs918023086 Chr.19: 15011214-15011214 on Build GRCh38
rs565643585 Chr.19: 15011390-15011390 on Build GRCh38
rs766972749 Chr.19: 15011270-15011270 on Build GRCh38
rs997265291 Chr.19: 15011363-15011363 on Build GRCh38
rs35647251 Chr.19: 15011233-15011233 on Build GRCh38
rs183990520 Chr.19: 15011241-15011241 on Build GRCh38
rs868848778 Chr.19: 15011370-15011370 on Build GRCh38
rs547530992 Chr.19: 15011382-15011382 on Build GRCh38
rs763476052 Chr.19: 15011251-15011251 on Build GRCh38
rs757340928 Chr.19: 15011372-15011372 on Build GRCh38
rs531540932 Chr.19: 15011271-15011271 on Build GRCh38
rs866516263 Chr.19: 15011378-15011378 on Build GRCh38
rs1050134170 Chr.19: 15011386-15011386 on Build GRCh38
rs771264777 Chr.19: 15011231-15011231 on Build GRCh38
rs752620327 Chr.19: 15011302-15011302 on Build GRCh38

View Full Product Details