Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 85688632-85688865 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GINS2
Entrez Gene ID
51659
Gene Name
GINS complex subunit 2
Gene Aliases
HSPC037, PSF2, Pfs2
UniGene
Hs.433180
Gene Chromosome Location
Chr.16: 85677674-85688982 on Build GRCh38
Transcripts
NM_016095.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6581299 c.78C>G p.I26M Chr.16: 85688821-85688821 on Build GRCh38
5437331 c.87C>T p.I29I Chr.16: 85688812-85688812 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs541936985 Chr.16: 85688661-85688661 on Build GRCh38
rs200675680 Chr.16: 85688781-85688781 on Build GRCh38
rs778622711 Chr.16: 85688774-85688774 on Build GRCh38
rs1031532661 Chr.16: 85688734-85688734 on Build GRCh38
rs887648050 Chr.16: 85688743-85688743 on Build GRCh38
rs149514448 Chr.16: 85688822-85688822 on Build GRCh38
rs1038369809 Chr.16: 85688675-85688675 on Build GRCh38
rs1007725831 Chr.16: 85688719-85688719 on Build GRCh38
rs956306351 Chr.16: 85688771-85688771 on Build GRCh38
rs186443043 Chr.16: 85688801-85688801 on Build GRCh38
rs890208288 Chr.16: 85688717-85688717 on Build GRCh38
rs144157425 Chr.16: 85688836-85688836 on Build GRCh38
rs2303205 Chr.16: 85688750-85688750 on Build GRCh38
rs985603956 Chr.16: 85688808-85688808 on Build GRCh38
rs1051895854 Chr.16: 85688829-85688829 on Build GRCh38
rs746534314 Chr.16: 85688793-85688793 on Build GRCh38
rs1006440098 Chr.16: 85688670-85688670 on Build GRCh38
rs1017238981 Chr.16: 85688722-85688722 on Build GRCh38
rs1048012455 Chr.16: 85688738-85688738 on Build GRCh38
rs544633986 Chr.16: 85688727-85688727 on Build GRCh38
rs753874275 Chr.16: 85688716-85688716 on Build GRCh38
rs879864415 Chr.16: 85688749-85688750 on Build GRCh38
rs143933756 Chr.16: 85688828-85688828 on Build GRCh38
rs981448397 Chr.16: 85688659-85688659 on Build GRCh38
rs140405835 Chr.16: 85688834-85688834 on Build GRCh38
rs2303204 Chr.16: 85688749-85688749 on Build GRCh38
rs540885917 Chr.16: 85688751-85688751 on Build GRCh38
rs780742906 Chr.16: 85688804-85688804 on Build GRCh38
rs777327004 Chr.16: 85688782-85688782 on Build GRCh38
rs564320518 Chr.16: 85688728-85688728 on Build GRCh38
rs766316541 Chr.16: 85688766-85688766 on Build GRCh38
rs911448997 Chr.16: 85688812-85688812 on Build GRCh38
rs905193729 Chr.16: 85688668-85688668 on Build GRCh38
rs956167739 Chr.16: 85688666-85688666 on Build GRCh38
rs554019578 Chr.16: 85688714-85688714 on Build GRCh38
rs914776111 Chr.16: 85688707-85688707 on Build GRCh38
rs762088448 Chr.16: 85688838-85688838 on Build GRCh38
rs754789705 Chr.16: 85688770-85688770 on Build GRCh38
rs949533196 Chr.16: 85688657-85688657 on Build GRCh38
rs201567976 Chr.16: 85688760-85688760 on Build GRCh38
rs181663633 Chr.16: 85688684-85688684 on Build GRCh38
rs772238729 Chr.16: 85688840-85688840 on Build GRCh38
rs749616681 Chr.16: 85688811-85688811 on Build GRCh38
rs943007490 Chr.16: 85688720-85688720 on Build GRCh38
rs767650470 Chr.16: 85688810-85688810 on Build GRCh38
rs529484630 Chr.16: 85688769-85688769 on Build GRCh38
rs1036154886 Chr.16: 85688665-85688665 on Build GRCh38
rs747807499 Chr.16: 85688776-85688776 on Build GRCh38
rs1045070116 Chr.16: 85688658-85688658 on Build GRCh38

View Full Product Details