Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 125302479-125302730 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GAPVD1
Entrez Gene ID
26130
Gene Name
GTPase activating protein and VPS9 domains 1
Gene Aliases
GAPEX5, GAPex-5, RAP6
UniGene
Hs.495134
Gene Chromosome Location
Chr.9: 125261794-125365011 on Build GRCh38
Transcripts
NM_001282679.1 NM_001282680.1 NM_001282681.1 NM_015635.3 XM_005251901.3 XM_005251904.3 XM_006717044.3 XM_011518499.2 XM_011518500.2 XM_011518502.2 XM_011518506.2 XM_011518507.2 XM_017014600.1 XM_017014601.1 XM_017014602.1 XM_017014603.1 XM_017014604.1 XM_017014605.1 XM_017014606.1 XM_017014607.1 XM_017014608.1 XM_017014609.1 XM_017014610.1 XM_017014611.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1105163 c.745G>T p.E249* Chr.9: 125302542-125302542 on Build GRCh38
4949605 c.820T>C p.Y274H Chr.9: 125302617-125302617 on Build GRCh38
3952337 c.722A>G p.D241G Chr.9: 125302519-125302519 on Build GRCh38
3654339 c.702C>T p.L234L Chr.9: 125302499-125302499 on Build GRCh38
3654340 c.869C>T p.T290I Chr.9: 125302666-125302666 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749152514 Chr.9: 125302646-125302646 on Build GRCh38
rs776742076 Chr.9: 125302689-125302689 on Build GRCh38
rs773295739 Chr.9: 125302541-125302541 on Build GRCh38
rs757773363 Chr.9: 125302634-125302634 on Build GRCh38
rs753020065 Chr.9: 125302596-125302596 on Build GRCh38
rs4437748 Chr.9: 125302569-125302569 on Build GRCh38
rs747985352 Chr.9: 125302531-125302531 on Build GRCh38
rs770645803 Chr.9: 125302558-125302558 on Build GRCh38
rs778725908 Chr.9: 125302649-125302649 on Build GRCh38
rs181982027 Chr.9: 125302548-125302548 on Build GRCh38
rs55779102 Chr.9: 125302647-125302647 on Build GRCh38
rs758271475 Chr.9: 125302666-125302666 on Build GRCh38
rs752115913 Chr.9: 125302632-125302632 on Build GRCh38
rs200721068 Chr.9: 125302600-125302600 on Build GRCh38
rs61733785 Chr.9: 125302676-125302676 on Build GRCh38
rs372739784 Chr.9: 125302517-125302517 on Build GRCh38
rs745967169 Chr.9: 125302692-125302692 on Build GRCh38
rs770821838 Chr.9: 125302677-125302677 on Build GRCh38
rs748967334 Chr.9: 125302520-125302520 on Build GRCh38
rs531525173 Chr.9: 125302582-125302582 on Build GRCh38
rs768254622 Chr.9: 125302523-125302523 on Build GRCh38
rs765485724 Chr.9: 125302593-125302593 on Build GRCh38
rs779510473 Chr.9: 125302518-125302518 on Build GRCh38
rs1054441369 Chr.9: 125302566-125302566 on Build GRCh38
rs547560589 Chr.9: 125302693-125302693 on Build GRCh38
rs776630852 Chr.9: 125302575-125302575 on Build GRCh38
rs542250668 Chr.9: 125302535-125302535 on Build GRCh38
rs777495342 Chr.9: 125302667-125302667 on Build GRCh38
rs147633449 Chr.9: 125302606-125302606 on Build GRCh38
rs1012994439 Chr.9: 125302616-125302616 on Build GRCh38
rs935942659 Chr.9: 125302553-125302553 on Build GRCh38
rs778284557 Chr.9: 125302511-125302511 on Build GRCh38
rs764531486 Chr.9: 125302619-125302619 on Build GRCh38
rs749854037 Chr.9: 125302512-125302512 on Build GRCh38
rs778815867 Chr.9: 125302528-125302528 on Build GRCh38
rs747890761 Chr.9: 125302658-125302658 on Build GRCh38

View Full Product Details